ITT Jabsco High-Purity Air-Operated Double-Diaphragm Pumps
Supplier: BPH PUMP & EQUIPMENT INC
Certificates
About this item
Unique air valve ensures a positive shift every time – pump will not stall.
- All diaphragms, liquid chambers, and manifolds are constructed
- of 100% virgin PTFE for long life and high purity
- Unique valve requires no lubrication or prepacking.
- Easily access air valve without disassembling the pump
Pumps are manufactured under class 1000 clean room conditions to guarantee purity.
- Environmentally Preferable:
Specifications
- Catalog No:
- MFLX74020-00
- Conexión de salida de aire.:
- 0.375 NPT(F)
- Conexión de entrada de aire:
- 0.25 NPT(F)
- Duty cycle continuous:
- Yes
- Inlet connection:
- 1/4" NPT, 0.25 NPT(F)
- Max. air inlet pressure:
- 6.9 bar (100 psi)
- Max. temperature:
- 80 °C (176 °F)
- Outlet connection:
- 0.25 NPT(F)
- Run dry:
- Yes
- Wetted materials:
- PTFE
- Garantía:
- 1 year