Orders LinkContactUsLinkComponent
LMI Manually Adjustable Control Metering Pumps
LMI Manually Adjustable Control Metering Pumps
LMI Manually Adjustable Control Metering Pumps
LMI Manually Adjustable Control Metering Pumps
LMI Manually Adjustable Control Metering Pumps
Manually adjustable stroke length and stroke frequency provide precise control of flow rates.
  • Clear face cover protects pump controls on low flow models (1 GPH and lower)
  • Chemical-resistant housing provides protection in harsh environments
These pumps include a four-function valve — an anti-siphon, back pressure, priming, and pressure relief valve all in one.
What's included: a four-function valve, 16-ft polyethylene tubing, foot valve/strainer, and 8-ft power cord with molded plug.
Service kits include a replacement diaphragm, seal, four pump head screws, and check valves.
  • Accuracy:
  • ±2%
  • ±2%
  • ±2%
  • ±2%
  • CSA regulatory approval:
  • Yes
  • Yes
  • Yes
  • Yes
  • UL(US) regulatory approval:
  • Yes
  • Yes
  • Yes
  • Yes
  • Inlet connection:
  • 3/8" OD tubing
  • 3/8" OD tubing
  • 3/8" OD tubing
  • 3/8" OD tubing
  • Max. pressure:
  • 100 psi
  • 60 psi
  • 100 psi
  • 100 psi
  • Max. temperature:
  • 60 °C (140 °F)
  • 121 °C (250 °F)
  • 121 °C (250 °F)
  • 121 °C (250 °F)
  • Min. flow rate:
  • 0.79 ml/min
  • 2.52 ml/min
  • 0.79 ml/min
  • 1.26 ml/min
  • Outlet connection:
  • 3/8" OD tubing
  • 3/8" OD tubing
  • 3/8" OD tubing
  • 3/8" OD tubing
  • Repeatability:
  • ±2%
  • ±2%
  • ±2%
  • Suction lift, dry:
  • 1524 mm of H₂O (60" H₂O)
  • 1524 mm of H₂O (60" H₂O)
  • 1524 mm of H₂O (60" H₂O)
  • 1524 mm of H₂O (60" H₂O)
  • Wetted materials:
  • Ceramic, Fluorofilm®, PVC, PVDF/Polyprel®
  • Ceramic, Fluorofilm®, PVC, PVDF/Polyprel®
  • Ceramic, Fluorofilm®, PVC, PVDF/Polyprel®
  • Ceramic, Fluorofilm®, PVC, PVDF/Polyprel®
  • Warranty:
  • 2 years
  • 2 years
  • 2 years
  • 2 years
Product Family Options
  • Description
    Maximum Flow Rate
    Power
    Height
    Length
    Width
  • Manual control solenoid diaphragm metering pump, 2.5 gal./h, 115 VAC
    158 ml/min
    115 VAC; 50/60 Hz; 3.5 A
    27.6 cm
    35.2 cm
    14.6 cm
    Each (1UN)
    Each (1UN) - $31,790.00
    $31,790.00
    Each (1UN)
       $31,790.00
    UL Listed
    Specifications
    Cat. No.
    MFLX74513-20
    Accuracy
    ±2%
    CSA regulatory approval
    Yes
    UL(US) regulatory approval
    Yes
    Inlet connection
    3/8" OD tubing
    Max. pressure
    100 psi
    Max. temperature
    60 °C (140 °F)
    Min. flow rate
    0.79 ml/min
    Outlet connection
    3/8" OD tubing
    Repeatability
    Suction lift, dry
    1524 mm of H₂O (60" H₂O)
    Wetted materials
    Ceramic, Fluorofilm®, PVC, PVDF/Polyprel®
    Warranty
    2 years
    UL Listed
  • Manual control solenoid diaphragm metering pump, 8.0 gal./h, 115 VAC
    500 ml/min
    115 VAC; 50/60 Hz; 3.5 A
    27.6 cm
    36.2 cm
    14.6 cm
    Each (1UN)
    Each (1UN) - $44,660.00
    $44,660.00
    Each (1UN)
       $44,660.00
    UL Listed
    Specifications
    Cat. No.
    MFLX74513-40
    Accuracy
    ±2%
    CSA regulatory approval
    Yes
    UL(US) regulatory approval
    Yes
    Inlet connection
    3/8" OD tubing
    Max. pressure
    60 psi
    Max. temperature
    121 °C (250 °F)
    Min. flow rate
    2.52 ml/min
    Outlet connection
    3/8" OD tubing
    Repeatability
    ±2%
    Suction lift, dry
    1524 mm of H₂O (60" H₂O)
    Wetted materials
    Ceramic, Fluorofilm®, PVC, PVDF/Polyprel®
    Warranty
    2 years
    UL Listed
  • Manual control solenoid diaphragm metering pump, 2.5 gal./h, 230 VAC
    158 ml/min
    230 VAC; 50 Hz; 1.75 A
    276 mm
    35.2 cm
    14.6 cm
    UL Listed
    Specifications
    Cat. No.
    MFLX74513-25
    Accuracy
    ±2%
    CSA regulatory approval
    Yes
    UL(US) regulatory approval
    Yes
    Inlet connection
    3/8" OD tubing
    Max. pressure
    100 psi
    Max. temperature
    121 °C (250 °F)
    Min. flow rate
    0.79 ml/min
    Outlet connection
    3/8" OD tubing
    Repeatability
    ±2%
    Suction lift, dry
    1524 mm of H₂O (60" H₂O)
    Wetted materials
    Ceramic, Fluorofilm®, PVC, PVDF/Polyprel®
    Warranty
    2 years
    UL Listed
  • Manual control solenoid diaphragm metering pump, 4.0 gal./h, 115 VAC
    252 ml/min
    115 VAC; 50/60 Hz; 3.5 A
    27.6 cm
    36.2 cm
    14.6 cm
    UL Listed
    Specifications
    Cat. No.
    MFLX74513-30
    Accuracy
    ±2%
    CSA regulatory approval
    Yes
    UL(US) regulatory approval
    Yes
    Inlet connection
    3/8" OD tubing
    Max. pressure
    100 psi
    Max. temperature
    121 °C (250 °F)
    Min. flow rate
    1.26 ml/min
    Outlet connection
    3/8" OD tubing
    Repeatability
    ±2%
    Suction lift, dry
    1524 mm of H₂O (60" H₂O)
    Wetted materials
    Ceramic, Fluorofilm®, PVC, PVDF/Polyprel®
    Warranty
    2 years
    UL Listed
Accessories
  •  
    Description
  • MFLX74513-18
    Service Kit for Solenoid Diaphragm Metering Pumps. 1.0 gal./h
    Each (1UN) - $3,890.00
  •  
    Description
  • MFLX74513-48
    Service Kit for Solenoid Diaphragm Metering Pumps, 8,0 gal./h
    Each (1UN) - $3,890.00
  •  
    Description
  • MFLX74513-38
    Service Kit for Solenoid Diaphragm Metering Pumps, 4,0 gal./h
    Each (1UN) - $3,770.00