PTFE Fitting Manifolds, Female NPT
Chemically-inert PTFE handles some of your most aggressive chemical solutions.
- Constructed of durable PTFE
Come in a variety of configurations—choose the number of ports and connections to suit your needs!
- Environmentally Preferable:
- Catalog No:
- MFLX31521-09
- MFLX31521-07
- MFLX31521-21
- MFLX31521-19
- MFLX31521-27
- MFLX31521-25
- MFLX31521-13
- MFLX31521-11
- MFLX31521-17
- MFLX31521-15
- MFLX31521-01
- Inlet NPT(F):
- 0.375"
- 0.25"
- 0.375"
- 0.25"
- 0.375"
- 0.25"
- 0.25"
- 0.5"
- 0.5"
- 0.375"
- 0.25"
- Presión máx.:
- 120 psi
- 120 psi
- 120 psi
- 120 psi
- 120 psi
- 120 psi
- 120 psi
- 120 psi
- 120 psi
- 120 psi
- 120 psi
- Max. temperature:
- 150 °C (300 °F)
- 150 °C (300 °F)
- 150 °C (300 °F)
- 150 °C (300 °F)
- 150 °C (300 °F)
- 150 °C (300 °F)
- 150 °C (300 °F)
- 150 °C (300 °F)
- 150 °C (300 °F)
- 150 °C (300 °F)
- 150 °C (300 °F)
- Min. temperature:
- –129 °C (–200 °F)
- –129 °C (–200 °F)
- –129 °C (–200 °F)
- –129 °C (–200 °F)
- –129 °C (–200 °F)
- –129 °C (–200 °F)
- –129 °C (–200 °F)
- –129 °C (–200 °F)
- –129 °C (–200 °F)
- –129 °C (–200 °F)
- –129 °C (–200 °F)
- Mounting hole:
- 0.2"
- 0.17"
- 0.2"
- 0.17"
- 0.2"
- 0.17"
- 0.17"
- 0.2"
- 0.2"
- 0.2"
- 0.17"
- No. of outlets:
- 4
- 4
- 8
- 8
- 10
- 10
- 6
- 4
- 6
- 6
- 2
- Outlet NPT(F):
- 0.25"
- 0.125"
- 0.25"
- 0.125"
- 0.25"
- 0.125"
- 0.125"
- 0.375"
- 0.375"
- 0.25"
- 0.125"
- Garantía:
- Out of box
- Out of box
- Out of box
- Out of box
- Out of box
- Out of box
- Out of box
- Out of box
- Out of box
- Out of box
- Out of box