Polypropylene (PP) High-Flow Chemical-Resistance Centrifugal Pumps
Proveedor: BPH PUMP & EQUIPMENT INC
Certificados
Acerca de este artículo
Twin bearing system handles thrust loads for extended service life.
- Quiet, energy efficient, and compact
- Internal cooling circuit reduces bearing temperature
- Enclosed motor for use in harsh environments
- All wetted parts are nonmetallic for excellent chemical resistance
High-flow polypropylene pumps utilize an open impeller design to produce high flows with very little horsepower.
Entrega: Includes a 6-ft (1.8 m) cord with stripped ends.
- Environmentally Preferable:
Especificaciones
- Catalog No:
- MFLX72013-05
- MFLX72013-20
- Duty cycle continuous:
- Yes
- Yes
- Presión máx.:
- 0.4 bar (5.7 psi)
- 0.7 bar (10 psi)
- Max. temperature:
- 80 °C (176 °F)
- 80 °C (176 °F)
- Motor hp:
- 1/38
- 1/125
- Garantía:
- 1 year
- 1 year