Fluoroplastic Enclosed Motor Chemical-Resistance Centrifugal Pumps
Supplier: BPH PUMP & EQUIPMENT INC
Certificates
About this item
Twin bearing system handles thrust loads for extended service life.
- Quiet, energy efficient, and compact
- Internal cooling circuit reduces bearing temperature
- Enclosed motor for use in harsh environments
Fluoroplastic pumps with carbon fiber-reinforced ETFE are designed for highly corrosive or high-purity applications.
Delivery: Includes a 6-ft (1.8 m) cord with stripped ends.
- Environmentally Preferable:
Specifications
- Catalog No:
- MFLX72011-05
- MFLX72011-00
- Duty cycle continuous:
- Yes
- Yes
- Presión máx.:
- 1.2 bar (17 psi)
- 1.2 bar (17 psi)
- Max. temperature:
- 80 °C (176 °F)
- 80 °C (176 °F)
- Motor hp:
- 1/16
- 1/16
- Garantía:
- 1 year
- 1 year