Orders LinkContactUsLinkComponent
Flowline Nonintrusive RF Capacitance Level Switch
Flowline Nonintrusive RF Capacitance Level Switch
Supplier:   FLOWLINE
Flowline Nonintrusive RF Capacitance Level Switch
Supplier:  FLOWLINE
Certificates
Flowline Nonintrusive RF Capacitance Level Switch
Flowline Nonintrusive RF Capacitance Level Switch
1 Options Available Below
Product Details & Documents
Measure level without touching fluid to avoid contamination.
  • Ideal for applications where purity is important
If you are concerned about your liquid's purity, then this nonintrusive level switch is the solution. Sensor mounts on the outside of your nonmetallic tank and never touches your liquid. Use with clean, noncoating, nonscaling, and conductive liquids. NEMA 4X polysulfone sensor enclosure is ideal for industrial environments. Includes 8-ft cable. Weld mounting bracket (sold separately) in place for firm hold.
Specifications for Product Family
Especificaciones
  • Accuracy of depth range:
  • ±1 mm
  • ±1 mm
  • Min. process temperature:
  • -40 °F
  • -40 °F
  • Max. process temperature:
  • 80 °C
  • 80 °C
  • Relay output:
  • (1) SPST relay. 60 VA. selectable NO/NC
  • (1) SPST relay. 60 VA. selectable NO/NC
  • Wetted materials:
  • Polypropylene
  • Polyethylene
  • Warranty:
  • 1 year
  • 1 year
Especificaciones
Product Family Options
Información del producto
  • Power
    Availability
    Price
  • MFLX43299-05
    Non-intrusive RF Capacitance Level Switch, PP Bracket
    Non-intrusive RF Capacitance Level Switch, PP Bracket
    12 - 36 VDC
    Discontinued
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX43299-05
    Accuracy of depth range
    ±1 mm
    Min. process temperature
    -40 °F
    Max. process temperature
    80 °C
    Relay output
    (1) SPST relay. 60 VA. selectable NO/NC
    Wetted materials
    Polypropylene
    Warranty
    1 year
  • MFLX43299-12
    Non-intrusive RF Capacitance Level Switch, PE Bracket
    Non-intrusive RF Capacitance Level Switch, PE Bracket
    12 - 36 VDC
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $9,290.00
    $9,290.00
    Each (1UN)
       $9,290.00
    Especificaciones:
    IP68 washdown
    More
    Specifications
    Cat. No.
    MFLX43299-12
    Accuracy of depth range
    ±1 mm
    Min. process temperature
    -40 °F
    Max. process temperature
    80 °C
    Relay output
    (1) SPST relay. 60 VA. selectable NO/NC
    Wetted materials
    Polyethylene
    Warranty
    1 year
    IP68 washdown