Orders LinkContactUsLinkComponent
Flowline Plastic Mini-Float Level Switches
Supplier:   FLOWLINE
Flowline Plastic Mini-Float Level Switches
Supplier:  FLOWLINE
Certificates
Flowline Plastic Mini-Float Level Switches
2 Options Available Below
Product Details & Documents
Control liquid level even in corrosive environments.
  • Excellent for clean, non-coating or non-scaling liquids
Choose either vertical or horizontal mounting to fit your application. Reliable reed switch output for interface with PLC or relay control devices. Your liquid must have a specific gravity of 0.8 or greater.
 : Includes submersible float with 2-ft (0.6-m) cable.
Specifications for Product Family
Especificaciones
  • Accuracy of depth range:
  • ±5 mm
  • ±5 mm
  • Min. process temperature:
  • -40 °F
  • -40 °F
  • Max. process temperature:
  • 90 °C
  • 90 °C
  • Max. pressure:
  • 25 psi
  • 25 psi
  • Relay output:
  • Reed: 50 VA at 120 VAC/VDC
  • Reed: 50 VA at 120 VAC/VDC
  • Wetted materials:
  • Polypropylene
  • PVDF
  • Warranty:
  • 1 year
  • 1 year
Especificaciones
Product Family Options
Información del producto
  • Power
    Availability
    Price
  • MFLX43299-24
    Float level switches; PP; 1/8" NPT(M)
    Float level switches; PP; 1/8" NPT(M)
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $1,130.00
    $1,130.00
    Each (1UN)
       $1,130.00
    Supplier Number: LV20-1201
    Especificaciones:
    CE Compliant
    More
    Specifications
    Cat. No.
    MFLX43299-24
    Accuracy of depth range
    ±5 mm
    Min. process temperature
    -40 °F
    Max. process temperature
    90 °C
    Max. pressure
    25 psi
    Relay output
    Reed: 50 VA at 120 VAC/VDC
    Wetted materials
    Polypropylene
    Warranty
    1 year
    CE Compliant
  • MFLX43299-26
    Mini-Float PVDF Level Switch, SPST, 1/8"NPT(M)
    Mini-Float PVDF Level Switch, SPST, 1/8"NPT(M)
    120 VAC
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $2,310.00
    $2,310.00
    Each (1UN)
       $2,310.00
    Supplier Number: LV20-5201
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX43299-26
    Accuracy of depth range
    ±5 mm
    Min. process temperature
    -40 °F
    Max. process temperature
    90 °C
    Max. pressure
    25 psi
    Relay output
    Reed: 50 VA at 120 VAC/VDC
    Wetted materials
    PVDF
    Warranty
    1 year