Plug Valves with Compression Fittings, PFA
Proveedor: ANDON SPECIALTIES INC.
Certificados
Acerca de este artículo
Chemically-inert PFA handles most aggressive chemicals.
- Constructed of durable PFA
Plug valves are FDA-approved and feature PFA seals and nuts.
- Environmentally Preferable:
Especificaciones
- Catalog No:
- MFLX98402-05
- MFLX98402-06
- Orifice:
- 1.98 mm (0.0781")
- 3.97 mm (0.1562")
- Tubing ext.Ø:
- 0.125"
- 0.25"
- Wetted materials:
- PFA
- PFA
- Garantía:
- 1 year
- 1 year