Stopcocks with Flow Indicator, PVC
Easily control flow with flow indicator handle.
- Constructed of durable PVC and EPDM
PVC bodies, EPDM seals and seats. Flow indicator on handle. Maximum pressure is 150 psi at the maximum temperature of 120 °F (49 °C); maximum vacuum is 29.9" Hg.
- Connection type:
- NPT(M)
- NPT(M) x Barbed
- Barbed
- NPT(F)
- NPT(M) x NPT(F)
- Inlet port:
- 1/4" NPT(M)
- 1/4" NPT(M)
- 1/4" to 1/2" Barb
- 1/4" NPT(F)
- 1/4" NPT(M)
- Outlet port:
- 1/4" NPT(M)
- 1/4" Barb
- 1/4" to 1/2" Barb
- 1/4" NPT(F)
- 1/4" NPT(F)
- Int. Ø tubing:
- 0.5 - 0.625"
- 0.25 - 0.5"
- Max. pressure:
- 150 psi
- 150 psi
- 150 psi
- 150 psi
- 150 psi
- Max. temperature:
- 48 °C (120 °F)
- 48 °C (120 °F)
- 48 °C (120 °F)
- 48 °C (120 °F)
- 48 °C (120 °F)
- Port size:
- 0.25 NPT(F)
- Orifice:
- 6.35 mm (0.25")
- 6.35 mm (0.25")
- 6.35 to 12.7 mm (0.25 - 0.5")
- 6.35 mm (0.25")
- 6.35 mm (0.25")
- Warranty:
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- Wetted materials:
- PVC
- PVC
- PVC
- PVC
- PVC
Product Family Options
- Description
- PVC Stopcock with Flow Indication, 1/4" NPT(M) x 1/4" NPT(M)Each (1UN)$829.00
Specifications
Cat. No.MFLX06225-52Connection typeNPT(M)Inlet port1/4" NPT(M)Outlet port1/4" NPT(M)Int. Ø tubingMax. pressure150 psiMax. temperature48 °C (120 °F)Port sizeOrifice6.35 mm (0.25")Warranty1 yearWetted materialsPVC - PVC Stopcock with Flow Indication, 1/4" NPT(M) x 1/4" hose barbEach (1UN)$1,010.00
Specifications
Cat. No.MFLX06225-54Connection typeNPT(M) x BarbedInlet port1/4" NPT(M)Outlet port1/4" BarbInt. Ø tubing0.5 - 0.625"Max. pressure150 psiMax. temperature48 °C (120 °F)Port sizeOrifice6.35 mm (0.25")Warranty1 yearWetted materialsPVC - PVC Stopcock w/ Flow Indication, 1/4-1/2" ID x 1/4-1/2" IDEach (1UN)$989.00
Specifications
Cat. No.MFLX06225-56Connection typeBarbedInlet port1/4" to 1/2" BarbOutlet port1/4" to 1/2" BarbInt. Ø tubing0.25 - 0.5"Max. pressure150 psiMax. temperature48 °C (120 °F)Port sizeOrifice6.35 to 12.7 mm (0.25 - 0.5")Warranty1 yearWetted materialsPVC - PVC Stopcock with Flow Indication, 1/4" NPT(F) x 1/4" NPT(F)Each (1UN)$1,010.00
Specifications
Cat. No.MFLX06225-50Connection typeNPT(F)Inlet port1/4" NPT(F)Outlet port1/4" NPT(F)Int. Ø tubingMax. pressure150 psiMax. temperature48 °C (120 °F)Port size0.25 NPT(F)Orifice6.35 mm (0.25")Warranty1 yearWetted materialsPVC - PVC Stopcock with Flow Indication, 1/4" NPT(M) x 1/4" NPT(F)-
Specifications
Cat. No.MFLX06225-58Connection typeNPT(M) x NPT(F)Inlet port1/4" NPT(M)Outlet port1/4" NPT(F)Int. Ø tubingMax. pressure150 psiMax. temperature48 °C (120 °F)Port sizeOrifice6.35 mm (0.25")Warranty1 yearWetted materialsPVC