Nordel Flexible Liner Pumps, Polypropylene
Supplier: VANTON PUMP & EQUIPMENT CORP
Certificates
About this item
Unique sealless design permits gentle pumping action that provides little churning or foaming.
- Transfer liquids containing up to 20% soft solids
- Sealless design with secondary containment system prevents potentially hazardous leakage
- Fixed-speed pumps are self-priming, reversible, and can run dry without damage
Polypropylene pumps have a Nordel® rubber liner; PTFE pumps have a Hypalon® rubber liner. Variable-speed models and DC-powered pumps are available — call our Applications Specialists for more information.
- Environmentally Preferable:
Specifications
- Catalog No:
- MFLX79440-30
- MFLX79440-00
- Duty cycle continuous:
- Yes
- Yes
- Inlet connection:
- 1-1/4" hose barb
- 3/4" hose barb
- Max. temperature:
- 85 °C (185 °F)
- 85 °C (185 °F)
- Motor hp:
- 1/2
- 1/4
- Outlet connection:
- 1" hose barb
- 3/4" hose barb
- Power conduit box connection:
- Yes
- Yes
- Run dry:
- Yes
- Yes
- Self-priming:
- Yes
- Yes
- Suction lift, dry:
- 1800 mm of H₂O (72" H₂O); 629 mm Hg (5.3" Hg)
- 1800 mm of H₂O (72" H₂O); 629 mm Hg (5.3" Hg)
- Wetted materials:
- Polypropylene, Nordel®
- Polypropylene, Nordel®
- Garantía:
- 1 year
- 1 year