Flexible Impeller Self-Priming Non-Metallic Pumps
Proveedor: BPH PUMP & EQUIPMENT INC
Certificados
Acerca de este artículo
Pumps fluids in applications where durability is needed—the 12 VDC models are ideal for remote/portable use.
- Pharmaceutical—processing lab solutions, lotions, preparations, and filtering solutions
- Industrial—circulating and transferring corrosive liquids, transferring solutions with entrained air (such as soaps and detergents), sampling, and pilot plant installations
- Agricultural—transferring liquid fertilizers, herbicides, and pesticides
- Metal plating—filtration of plating solutions, filling, and emptying vats
These flexible impeller self-priming non-metallic pumps are available in low-flow and high-flow options.
Low-flow pumps are ideal for applications where gas-laden fluids need to be transferred. Built-in thermal overload protection prevents motor damage. Model 75421-00 is constructed with a flass-filled polyester body, Buna-N impeller, and stainless steel (SS) shaft.
High-flow pumps are ideal for higher flow applications that require continuous duty. The epoxy body offers great resistance to a wide range of fluids; choose an impeller material compatible with your pumped fluid. Motors have a conduit box for easy wiring.
- Environmentally Preferable:
Especificaciones
- Catalog No:
- MFLX75501-12
- Duty cycle continuous:
- Yes
- Inlet connection:
- 3/4" NPT(M)
- Max. temperature:
- 82 °C (180 °F)
- Motor type:
- TEFC
- Motor hp:
- 3/4
- Outlet connection:
- 3/4" NPT(M)
- Power conduit box connection:
- Yes
- Run dry:
- No
- Suction lift, dry:
- 3100 mm of H₂O (120" H₂O); 534 mm Hg (8.8" Hg)
- Suction lift, wet:
- 6700 mm of H₂O (264" H₂O; 270 mm Hg (19.3" Hg)
- Wetted materials:
- Epoxy, Neoprene
- Garantía:
- 1 year