Hand-Operated Piston Dispensing Drum Pump, PVC
Supplier: ACTION PUMP COMPANY
Certificates
About this item
Economical piston pumps are ideal for a variety of fluids.
- Adjustable to most pails and drums
Models 07079-50 and -55 feature an angled spout to help prevent dripping. Models 07079-60 and -65 are made entirely of PVC, including the piston and seal, making them an ideal option for sanitary applications.
- Environmentally Preferable:
Specifications
- Catalog No:
- MFLX07079-55
- MFLX07079-50
- MFLX07079-60
- Max. temperature:
- 60 °C (140 °F)
- 60 °C (140 °F)
- 77 °C (170 °F)
- Viscosidad máx.:
- 4000 SSU
- 4000 SSU
- 4000 SSU
- Outlet connection:
- Spout
- Spout
- Hose
- Tube ext.Ø, inlet:
- 1.27 cm (0.5")
- 1.27 cm (0.5")
- 1.27 cm (0.5")
- Tube ext.Ø, outlet:
- 1.27 cm (0.5")
- 1.27 cm (0.5")
- 1.27 cm (0.5")
- Tube length, inlet:
- 97 cm (38")
- 56 cm (22")
- 57 cm (22.5")
- Tube length, outlet:
- 56 cm (22")
- Wetted materials:
- PVC
- PVC
- PVC, Polyethylene Tube
- Garantía:
- 6 Months
- 6 Months
- 1 year