Pump Motor Spider Inserts, Buna-N®
Spider Insert, Buna-N, for A/L090/L095 Couplings
Catalog # MFLX07127-69
Supplier: GRAINGER
Supplier #: 1X407
Jump to:
Product Details & Documents
Create your own motor mounting with the hardware below.
- Constructed of durable Buna-N®
Spider smooths power transfer, absorbs vibration, and compensates for minor misalignment.
Especificaciones
- Description:Spider Insert, Buna-N, for A/L090/L095 Couplings
- Cat. No.:MFLX07127-69
- Warranty:None
Accessories
Información del producto
- Catalog #Motor HPAvailabilityPrice
- Coupling hardware for 5/8" diameter bore, 7,2 hp maximumCoupling hardware for 5/8" diameter bore, 7,2 hp maximumMFLX07127-597.2Log in Para disponibilidadEach (1UN) - $517.00