Micropump Compact Low-Flow Gear Pump Heads - C Mount Design
Offering remote C-mount choices for your low-flow applications.
- Use for applications requiring high initial differential pressure.
- Suction-shoe design ensures complete fluid transfer for increased volumetric accuracy
- Pump efficiency increases as discharge pressure increases
The compact pump head requires a drive for proper operation. Order drives separately.
Optional Service kit includes: gears, suction shoe, shaft seals, and springs. Order separately.
Contact our Application Specialists for assistance.
- Environmentally Preferable:
- Material del cuerpo:
- 316 SS
- 316 SS
- 316 SS
- 316 SS
- 316 SS
- Differential pressure:
- 3.4 bar (50 psi)
- 5.2 bar (75 psi)
- 2.8 bar (40 psi)
- 1.4 bar (20 psi)
- 2.8 bar (40 psi)
- Duty cycle continuous:
- Yes
- Yes
- Yes
- Yes
- Yes
- Gear material:
- PTFE
- PPS
- Carbon graphite
- Carbon graphite
- Carbon graphite
- Inlet connection:
- 0.125 NPT(F)
- 0.125 NPT(F)
- 0.125 NPT(F)
- 0.125 NPT(F)
- 0.125 NPT(F)
- Max. temperature:
- 77 °C (170 °F)
- 177 °C (350 °F)
- 177 °C (350 °F)
- 177 °C (350 °F)
- 177 °C (350 °F)
- Viscosidad máx.:
- 100 cP
- 100 cP
- 100 cP
- 100 cP
- 100 cP
- Min. temperature:
- –46 °C (–50 °F)
- –46 °C (–50 °F)
- –46 °C (–50 °F)
- –46 °C (–50 °F)
- –46 °C (–50 °F)
- Outlet connection:
- 0.125 NPT(F)
- 0.125 NPT(F)
- 0.125 NPT(F)
- 0.125 NPT(F)
- 0.125 NPT(F)
- Particulates:
- No
- No
- No
- No
- No
- Relief valve:
- No
- No
- No
- No
- No
- Run dry:
- Under 30 seconds
- Under 30 seconds
- Under 30 seconds
- Under 30 seconds
- Under 30 seconds
- Seal material:
- PTFE
- PTFE
- PTFE
- PTFE
- PTFE
- Suction lift, wet:
- 12" H₂O
- Garantía:
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
Frequently Bought Together