Mini Submersible Magnetic Drive Pumps
Operate fully submerged or in open air.
- Liquid-cooled models keep heat-sensitive fluids from warming up
- Thick (1/8") epoxy coating dissipates heat quickly and provides good chemical resistance
- UL-Listed
These mini submersible magnetic drive pumps are good for low-viscosity liquids including solvents, dyes, inks, and mild corrosives.
: Includes a three-wire cord.
- Environmentally Preferable:
- Duty cycle continuous:
- Yes
- Yes
- Yes
- Yes
- Inlet connection:
- 0.5" hosebarb
- 0.5" hosebarb
- 0.75 NPT(F)
- 0.375 NPT(F)
- Presión máx.:
- 3.4 bar (50 psi)
- 1.7 bar (25 psi)
- 1.7 bar (25 psi)
- 1.7 bar (25 psi)
- Max. temperature:
- 71 °C (160 °F)
- 71 °C (160 °F)
- 71 °C (160 °F)
- 71 °C (160 °F)
- Motor type:
- TENV
- TENV
- TENV
- TENV
- Motor hp:
- 1/200
- 1/200
- 1/20
- 1/35
- Outlet connection:
- 0.5" hosebarb
- 0.5" hosebarb
- Wetted materials:
- PP head, glass-filled nylon impeller, Buna-N O-ring, 316 SS spindle
- PP head, glass-filled nylon impeller, Buna-N O-ring, 316 SS spindle
- PP head, glass-filled nylon impeller, Buna-N O-ring, ceramic spindle
- PP head, glass-filled nylon impeller, Buna-N O-ring, ceramic spindle
- Garantía:
- 1 year
- 1 year
- 1 year
- 1 year
Frequently Bought Together