Orders LinkContactUsLinkComponent
CPC® Quick-Disconnect Fittings, Compression Inserts
CPC® Quick-Disconnect Fittings, Compression Inserts
Certificates
CPC® Quick-Disconnect Fittings, Compression Inserts
17 Options Available Below
Product Details & Documents
No more struggling—just press thumb latch.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Fittings connect in a snap
These CPC® plastic quick-disconnect fittings feature an ergonomic design and thumb latch for an easy grip and simple operation. Simply plug the coupling insert into the coupling body to connect fittings. Fittings will lock into place immediately. The audible click when the coupling engages confirms a solid connection. To disconnect fitting halves, simply press the button on the coupling body. Standard flow quick-disconnect couplings require a body and an insert of the same flow size.
A complete coupler requires a fitting body and an insert (order separately).
Specifications for Product Family
Especificaciones
  • Length:
  • 4.8514 cm
  • 4.8514 cm
  • 3.683 cm
  • 2.921 cm
  • 3.556 cm
  • 3.302 cm
  • 4.445 cm
  • 4.2672 cm
  • 3.175 cm
  • 3.5052 cm
  • 4.318 cm
  • 3.683 cm
  • 2.921 cm
  • 4.2672 cm
  • 4.318 cm
  • 4.8514 cm
  • 4.5212 cm
  • 4.8514 cm
  • 3.683 cm
  • 4.318 cm
  • 3.683 cm
  • 4.445 cm
  • 4.2672 cm
  • 4.2672 cm
  • 3.5052 cm
  • Max. operating pressure:
  • 120 psi
  • 120 psi
  • 120 psi
  • 120 psi
  • 120 psi
  • 120 psi
  • 120 psi
  • 120 psi
  • 120 psi
  • 120 psi
  • 120 psi
  • 120 psi
  • 120 psi
  • 120 psi
  • 120 psi
  • 120 psi
  • 120 psi
  • 120 psi
  • 120 psi
  • 120 psi
  • 120 psi
  • 120 psi
  • 120 psi
  • 120 psi
  • 120 psi
  • 120 psi
  • 120 psi
  • Max. temperature:
  • 82 °C (180 °F)
  • 82 °C (180 °F)
  • 82 °C (180 °F)
  • 82 °C (180 °F)
  • 82 °C (180 °F)
  • 82 °C (180 °F)
  • 82 °C (180 °F)
  • 82 °C (180 °F)
  • 82 °C (180 °F)
  • 82 °C (180 °F)
  • 82 °C (180 °F)
  • 82 °C (180 °F)
  • 82 °C (180 °F)
  • 71 °C (160 °F)
  • 71 °C (160 °F)
  • 71 °C (160 °F)
  • 82 °C (180 °F)
  • 82 °C (180 °F)
  • 82 °C (180 °F)
  • 82 °C (180 °F)
  • 71 °C (160 °F)
  • 82 °C (180 °F)
  • 82 °C (180 °F)
  • 82 °C (180 °F)
  • 82 °C (180 °F)
  • 71 °C (160 °F)
  • 82 °C (180 °F)
  • Max. vacuum:
  • 711 mm Hg (28" Hg)
  • 711 mm Hg (28" Hg)
  • 711 mm Hg (28" Hg)
  • 711 mm Hg (28" Hg)
  • 711 mm Hg (28" Hg)
  • 711 mm Hg (28" Hg)
  • 711 mm Hg (28" Hg)
  • 711 mm Hg (28" Hg)
  • 711 mm Hg (28" Hg)
  • 711 mm Hg (28" Hg)
  • 711 mm Hg (28" Hg)
  • 711 mm Hg (28" Hg)
  • 711 mm Hg (28" Hg)
  • 711 mm Hg (28" Hg)
  • 711 mm Hg (28" Hg)
  • 711 mm Hg (28" Hg)
  • 711 mm Hg (28" Hg)
  • 711 mm Hg (28" Hg)
  • 711 mm Hg (28" Hg)
  • 711 mm Hg (28" Hg)
  • 711 mm Hg (28" Hg)
  • 711 mm Hg (28" Hg)
  • 711 mm Hg (28" Hg)
  • 711 mm Hg (28" Hg)
  • 711 mm Hg (28" Hg)
  • 711 mm Hg (28" Hg)
  • 711 mm Hg (28" Hg)
  • Seal material:
  • Buna N
  • Buna N
  • Buna N
  • Buna N
  • Buna N
  • Buna N
  • Buna N
  • Buna N
  • Buna N
  • Buna N
  • Buna N
  • Buna N
  • Buna N
  • EPDM
  • EPDM
  • EPDM
  • Buna N
  • Buna N
  • Buna N
  • Buna N
  • EPDM
  • Buna N
  • Buna N
  • Buna N
  • Buna N
  • EPDM
  • Buna N
  • Sterilisation:
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Gamma
  • Gamma
  • Gamma
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Gamma
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Gamma
  • Autoclave or ethylene oxide (ETO)
  • Valve spring material:
  • 316 Stainless Steel
  • 316 Stainless Steel
  • 316 Stainless Steel
  • 316 Stainless Steel
  • 316 Stainless Steel
  • 316 Stainless Steel
  • 316 Stainless Steel
  • 316 Stainless Steel
  • 316 Stainless Steel
  • 316 Stainless Steel
  • 316 Stainless Steel
  • 316 Stainless Steel
  • 316 Stainless Steel
  • 316 Stainless Steel
  • 316 Stainless Steel
  • 316 Stainless Steel
  • 316 Stainless Steel
  • 316 Stainless Steel
  • Warranty:
  • 1 year
  • 1 year
  • 1 year
  • 3 Months
  • 3 Months
  • 3 Months
  • 3 Months
  • 1 year
  • 3 Months
  • 3 Months
  • 3 Months
  • 3 Months
  • 1 year
  • 3 Months
  • 3 Months
  • 3 Months
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 3 Months
  • 1 year
  • 3 Months
  • 3 Months
  • 3 Months
  • 3 Months
  • 3 Months
Especificaciones
Product Family Options
Bulk product
Información del producto
  • Type
    Connection Type
    Flow pattern
    Fitting Material
    Quick-disconnect Flow Size
    Availability
    Price
  • MFLX06360-41BLK
    Quick-Disconnect Fitting, Compression Insert, Acetal, Valved, ⁵/₃₂" OD, PMCD20025; 25/PK
    Quick-Disconnect Fitting, Compression Insert, Acetal, Valved, ⁵/₃₂" OD, PMCD20025; 25/PK
    Valved
    Quick-Disconnect to Compression
    Straight
    Acetal
    ¼"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (25UN)
    Each (25UN) - $6,970.00
    $6,970.00
    Each (25UN)
       $6,970.00
    Supplier Number: PMCD20025 (25PK)
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX06360-41BLK
    Length
    Max. operating pressure
    120 psi
    Max. temperature
    82 °C (180 °F)
    Max. vacuum
    711 mm Hg (28" Hg)
    Seal material
    Buna N
    Sterilisation
    Autoclave or ethylene oxide (ETO)
    Valve spring material
    316 Stainless Steel
    Warranty
    1 year
  • MFLX31307-54BLK
    Quick-Disconnect Fitting, Compression Insert, Panel-Mount, Acetal, Valved, ³/₈" OD, PLCD40006; 25/PK
    Quick-Disconnect Fitting, Compression Insert, Panel-Mount, Acetal, Valved, ³/₈" OD, PLCD40006; 25/PK
    Quick-Disconnect x Compression
    Straight
    Acetal
    ¼"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (25UN)
    Each (25UN) - $6,420.00
    $6,420.00
    Each (25UN)
       $6,420.00
    Supplier Number: PLCD40006
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX31307-54BLK
    Length
    4.8514 cm
    Max. operating pressure
    120 psi
    Max. temperature
    82 °C (180 °F)
    Max. vacuum
    711 mm Hg (28" Hg)
    Seal material
    Buna N
    Sterilisation
    Autoclave or ethylene oxide (ETO)
    Valve spring material
    316 Stainless Steel
    Warranty
    1 year
  • MFLX31307-44BLK
    Quick-Disconnect Fitting, Compression Insert, Panel-Mount, Acetal, Straight-Through, ³/₈" OD, PLC40006; 25/PK
    Quick-Disconnect Fitting, Compression Insert, Panel-Mount, Acetal, Straight-Through, ³/₈" OD, PLC40006; 25/PK
    Quick-Disconnect x Compression
    Straight
    Acetal
    ¼"
    Discontinued
    Supplier Number: PLC40006
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX31307-44BLK
    Length
    4.8514 cm
    Max. operating pressure
    120 psi
    Max. temperature
    82 °C (180 °F)
    Max. vacuum
    711 mm Hg (28" Hg)
    Seal material
    Buna N
    Sterilisation
    Autoclave or ethylene oxide (ETO)
    Valve spring material
    Warranty
    1 year
  • MFLX06364-30BLK
    Quick-Disconnect Fitting, Compression Insert, Polypropylene, Valved, ¹/₄" OD, PMCD200412; 25/PK
    Quick-Disconnect Fitting, Compression Insert, Polypropylene, Valved, ¹/₄" OD, PMCD200412; 25/PK
    Valved
    Quick-Disconnect to Compression
    Straight
    PP
    ⅛"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (25UN)
    Each (25UN) - $5,580.00
    $5,580.00
    Each (25UN)
       $5,580.00
    Supplier Number: PMCD200412 (25PK)
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX06364-30BLK
    Length
    3.683 cm
    Max. operating pressure
    120 psi
    Max. temperature
    71 °C (160 °F)
    Max. vacuum
    711 mm Hg (28" Hg)
    Seal material
    EPDM
    Sterilisation
    Gamma
    Valve spring material
    316 Stainless Steel
    Warranty
    3 Months
  • MFLX06361-32BLK
    Quick-Disconnect Fitting, Compression Insert, Panel-Mount, Acetal, Valved; ¹/₄" OD, PMCD4004; 25/PK
    Quick-Disconnect Fitting, Compression Insert, Panel-Mount, Acetal, Valved; ¹/₄" OD, PMCD4004; 25/PK
    Quick-Disconnect x Compression
    Straight
    Acetal
    ⅛"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (25UN)
    Each (25UN) - $6,830.00
    $6,830.00
    Each (25UN)
       $6,830.00
    Supplier Number: PMCD4004 (25PK)
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX06361-32BLK
    Length
    4.318 cm
    Max. operating pressure
    120 psi
    Max. temperature
    82 °C (180 °F)
    Max. vacuum
    711 mm Hg (28" Hg)
    Seal material
    Buna N
    Sterilisation
    Autoclave or ethylene oxide (ETO)
    Valve spring material
    316 Stainless Steel
    Warranty
    1 year
  • MFLX06360-30BLK
    Quick-Disconnect Fitting, Compression Insert, Acetal, Valved, ¹/₈" Flow Size, ¹/₄" OD, PMCD2004; 25/PK
    Quick-Disconnect Fitting, Compression Insert, Acetal, Valved, ¹/₈" Flow Size, ¹/₄" OD, PMCD2004; 25/PK
    Valved
    Quick-Disconnect to Compression
    Straight
    Acetal
    ⅛"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (25UN)
    Each (25UN) - $3,730.00
    $3,730.00
    Each (25UN)
       $3,730.00
    Supplier Number: PMCD2004 (25PK)
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX06360-30BLK
    Length
    3.683 cm
    Max. operating pressure
    120 psi
    Max. temperature
    82 °C (180 °F)
    Max. vacuum
    711 mm Hg (28" Hg)
    Seal material
    Buna N
    Sterilisation
    Autoclave or ethylene oxide (ETO)
    Valve spring material
    316 Stainless Steel
    Warranty
    3 Months
  • MFLX06360-74BLK
    Quick-Disconnect Fitting, Compression Insert, Acetal, Valved, ¹/₄" Flow Size, ¹/₄" OD, PLCD20004; 25/PK
    Quick-Disconnect Fitting, Compression Insert, Acetal, Valved, ¹/₄" Flow Size, ¹/₄" OD, PLCD20004; 25/PK
    Valved
    Quick-Disconnect to Compression
    Straight
    Acetal
    ¼"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (25UN)
    Each (25UN) - $4,550.00
    $4,550.00
    Each (25UN)
       $4,550.00
    Supplier Number: PLCD20004 (25PK)
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX06360-74BLK
    Length
    4.445 cm
    Max. operating pressure
    120 psi
    Max. temperature
    82 °C (180 °F)
    Max. vacuum
    711 mm Hg (28" Hg)
    Seal material
    Buna N
    Sterilisation
    Autoclave or ethylene oxide (ETO)
    Valve spring material
    316 Stainless Steel
    Warranty
    3 Months
  • MFLX06360-75BLK
    Quick-Disconnect Fitting, Compression Insert, Acetal, Valved, ¹/₄" Flow Size, ³/₈" OD, PLCD20006; 25/PK
    Quick-Disconnect Fitting, Compression Insert, Acetal, Valved, ¹/₄" Flow Size, ³/₈" OD, PLCD20006; 25/PK
    Valved
    Quick-Disconnect to Compression
    Straight
    Acetal
    ¼"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (25UN)
    Each (25UN) - $4,550.00
    $4,550.00
    Each (25UN)
       $4,550.00
    Supplier Number: PLCD20006 (25PK)
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX06360-75BLK
    Length
    4.2672 cm
    Max. operating pressure
    120 psi
    Max. temperature
    82 °C (180 °F)
    Max. vacuum
    711 mm Hg (28" Hg)
    Seal material
    Buna N
    Sterilisation
    Autoclave or ethylene oxide (ETO)
    Valve spring material
    316 Stainless Steel
    Warranty
    3 Months
  • MFLX06364-75BLK
    Quick-Disconnect Fitting, Compression Insert, Polypropylene, Valved, ³/₈" OD, PLCD2000612; 25/PK
    Quick-Disconnect Fitting, Compression Insert, Polypropylene, Valved, ³/₈" OD, PLCD2000612; 25/PK
    Valved
    Quick-Disconnect to Compression
    Straight
    PP
    ¼"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (25UN)
    Each (25UN) - $7,100.00
    $7,100.00
    Each (25UN)
       $7,100.00
    Supplier Number: PLCD2000612 (25PK)
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX06364-75BLK
    Length
    4.2672 cm
    Max. operating pressure
    120 psi
    Max. temperature
    71 °C (160 °F)
    Max. vacuum
    711 mm Hg (28" Hg)
    Seal material
    EPDM
    Sterilisation
    Gamma
    Valve spring material
    316 Stainless Steel
    Warranty
    3 Months
  • MFLX06360-77BLK
    Quick-Disconnect Fitting, Compression Insert, Acetal, Straight-Through, ¹/₄" Flow Size, ³/₈" OD, PLC20006; 25/PK
    Quick-Disconnect Fitting, Compression Insert, Acetal, Straight-Through, ¹/₄" Flow Size, ³/₈" OD, PLC20006; 25/PK
    Quick-Disconnect to Compression
    Straight
    Acetal
    ¼"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (25UN)
    Each (25UN) - $2,540.00
    $2,540.00
    Each (25UN)
       $2,540.00
    Supplier Number: PLC20006 (25PK)
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX06360-77BLK
    Length
    3.5052 cm
    Max. operating pressure
    120 psi
    Max. temperature
    82 °C (180 °F)
    Max. vacuum
    711 mm Hg (28" Hg)
    Seal material
    Buna N
    Sterilisation
    Autoclave or ethylene oxide (ETO)
    Valve spring material
    Warranty
    3 Months
Down pack product
Información del producto
  • Type
    Connection Type
    Flow pattern
    Fitting Material
    Quick-disconnect Flow Size
    Availability
    Price
  • MFLX06360-30
    Quick-Disconnect Fitting, Compression Insert, Acetal, Valved, ¹/₈" Flow Size, ¹/₄" OD, PMCD2004; 1/PK
    Quick-Disconnect Fitting, Compression Insert, Acetal, Valved, ¹/₈" Flow Size, ¹/₄" OD, PMCD2004; 1/PK
    Valved
    Quick-Disconnect to Compression
    Straight
    Acetal
    ⅛"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $273.00
    $273.00
    Each (1UN)
       $273.00
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX06360-30
    Length
    3.683 cm
    Max. operating pressure
    120 psi
    Max. temperature
    82 °C (180 °F)
    Max. vacuum
    711 mm Hg (28" Hg)
    Seal material
    Buna N
    Sterilisation
    Autoclave or ethylene oxide (ETO)
    Valve spring material
    316 Stainless Steel
    Warranty
    3 Months
  • MFLX06360-32
    Quick-Disconnect Fitting, Compression Insert, Acetal, Straight-Through, ⅛" Flow Size, ¼" OD; 1/Ea
    Quick-Disconnect Fitting, Compression Insert, Acetal, Straight-Through, ⅛" Flow Size, ¼" OD; 1/Ea
    Open
    Quick-Disconnect to Compression
    Straight
    POM
    ⅛"
    Discontinued
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX06360-32
    Length
    2.921 cm
    Max. operating pressure
    120 psi
    Max. temperature
    82 °C (180 °F)
    Max. vacuum
    711 mm Hg (28" Hg)
    Seal material
    Buna N
    Sterilisation
    Autoclave or ethylene oxide (ETO)
    Valve spring material
    Warranty
    3 Months
  • MFLX06360-35
    Quick-Disconnect Fitting, Compression Insert, Acetal, Valved, ¹/₈" Flow Size, ³/₈" OD, PMCD2006; 1/PK
    Quick-Disconnect Fitting, Compression Insert, Acetal, Valved, ¹/₈" Flow Size, ³/₈" OD, PMCD2006; 1/PK
    Valved
    Quick-Disconnect to Compression
    Straight
    POM
    ⅛"
    Discontinued
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX06360-35
    Length
    3.556 cm
    Max. operating pressure
    120 psi
    Max. temperature
    82 °C (180 °F)
    Max. vacuum
    711 mm Hg (28" Hg)
    Seal material
    Buna N
    Sterilisation
    Autoclave or ethylene oxide (ETO)
    Valve spring material
    316 Stainless Steel
    Warranty
    3 Months
  • MFLX06360-37
    Quick-Disconnect Fitting, Compression Insert, Acetal, Straight-Through, ⅛" Flow Size, ⅜" OD; 1/Ea
    Quick-Disconnect Fitting, Compression Insert, Acetal, Straight-Through, ⅛" Flow Size, ⅜" OD; 1/Ea
    Open
    Quick-Disconnect to Compression
    Straight
    POM
    ⅛"
    Discontinued
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX06360-37
    Length
    3.302 cm
    Max. operating pressure
    120 psi
    Max. temperature
    82 °C (180 °F)
    Max. vacuum
    711 mm Hg (28" Hg)
    Seal material
    Buna N
    Sterilisation
    Autoclave or ethylene oxide (ETO)
    Valve spring material
    Warranty
    3 Months
  • MFLX06360-41
    Quick-Disconnect Fitting, Compression Insert, Acetal, Valved, ⁵/₃₂" OD, PMCD20025; 1/PK
    Quick-Disconnect Fitting, Compression Insert, Acetal, Valved, ⁵/₃₂" OD, PMCD20025; 1/PK
    Valved
    Quick-Disconnect to Compression
    Straight
    Acetal
    ¼"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $541.00
    $541.00
    Each (1UN)
       $541.00
    Especificaciones:
    CE certificate
    More
    Specifications
    Cat. No.
    MFLX06360-41
    Length
    Max. operating pressure
    120 psi
    Max. temperature
    82 °C (180 °F)
    Max. vacuum
    711 mm Hg (28" Hg)
    Seal material
    Buna N
    Sterilisation
    Autoclave or ethylene oxide (ETO)
    Valve spring material
    316 Stainless Steel
    Warranty
    1 year
    CE certificate
  • MFLX06360-74
    Quick-Disconnect Fitting, Compression Insert, Acetal, Valved, ¹/₄" Flow Size, ¹/₄" OD, PLCD20004; 1/PK
    Quick-Disconnect Fitting, Compression Insert, Acetal, Valved, ¹/₄" Flow Size, ¹/₄" OD, PLCD20004; 1/PK
    Valved
    Quick-Disconnect to Compression
    Straight
    Acetal
    ¼"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $303.00
    $303.00
    Each (1UN)
       $303.00
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX06360-74
    Length
    4.445 cm
    Max. operating pressure
    120 psi
    Max. temperature
    82 °C (180 °F)
    Max. vacuum
    711 mm Hg (28" Hg)
    Seal material
    Buna N
    Sterilisation
    Autoclave or ethylene oxide (ETO)
    Valve spring material
    316 Stainless Steel
    Warranty
    3 Months
  • MFLX06360-75
    Quick-Disconnect Fitting, Compression Insert, Acetal, Valved, ¹/₄" Flow Size, ³/₈" OD, PLCD20006; 1/PK
    Quick-Disconnect Fitting, Compression Insert, Acetal, Valved, ¹/₄" Flow Size, ³/₈" OD, PLCD20006; 1/PK
    Valved
    Quick-Disconnect to Compression
    Straight
    Acetal
    ¼"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $303.00
    $303.00
    Each (1UN)
       $303.00
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX06360-75
    Length
    4.2672 cm
    Max. operating pressure
    120 psi
    Max. temperature
    82 °C (180 °F)
    Max. vacuum
    711 mm Hg (28" Hg)
    Seal material
    Buna N
    Sterilisation
    Autoclave or ethylene oxide (ETO)
    Valve spring material
    316 Stainless Steel
    Warranty
    3 Months
  • MFLX06360-76
    Quick-Disconnect Fitting, Compression Insert, Acetal, Straight-Through, ¹/₄" Flow Size, ¹/₄" OD, PLC20004; 1/PK
    Quick-Disconnect Fitting, Compression Insert, Acetal, Straight-Through, ¹/₄" Flow Size, ¹/₄" OD, PLC20004; 1/PK
    Open
    Quick-Disconnect to Compression
    Straight
    POM
    ¼"
    Discontinued
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX06360-76
    Length
    3.175 cm
    Max. operating pressure
    120 psi
    Max. temperature
    82 °C (180 °F)
    Max. vacuum
    711 mm Hg (28" Hg)
    Seal material
    Buna N
    Sterilisation
    Autoclave or ethylene oxide (ETO)
    Valve spring material
    Warranty
    3 Months
  • MFLX06360-77
    Quick-Disconnect Fitting, Compression Insert, Acetal, Straight-Through, ¹/₄" Flow Size, ³/₈" OD, PLC20006; 1/PK
    Quick-Disconnect Fitting, Compression Insert, Acetal, Straight-Through, ¹/₄" Flow Size, ³/₈" OD, PLC20006; 1/PK
    Quick-Disconnect to Compression
    Straight
    Acetal
    ¼"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $197.00
    $197.00
    Each (1UN)
       $197.00
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX06360-77
    Length
    3.5052 cm
    Max. operating pressure
    120 psi
    Max. temperature
    82 °C (180 °F)
    Max. vacuum
    711 mm Hg (28" Hg)
    Seal material
    Buna N
    Sterilisation
    Autoclave or ethylene oxide (ETO)
    Valve spring material
    Warranty
    3 Months
  • MFLX06361-32
    Quick-Disconnect Fitting, Compression Insert, Panel-Mount, Acetal, Valved; ¹/₄" OD, PMCD4004; 1/PK
    Quick-Disconnect Fitting, Compression Insert, Panel-Mount, Acetal, Valved; ¹/₄" OD, PMCD4004; 1/PK
    Quick-Disconnect x Compression
    Straight
    Acetal
    ⅛"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $453.00
    $453.00
    Each (1UN)
       $453.00
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX06361-32
    Length
    4.318 cm
    Max. operating pressure
    120 psi
    Max. temperature
    82 °C (180 °F)
    Max. vacuum
    711 mm Hg (28" Hg)
    Seal material
    Buna N
    Sterilisation
    Autoclave or ethylene oxide (ETO)
    Valve spring material
    316 Stainless Steel
    Warranty
    1 year
  • MFLX06364-30
    Quick-Disconnect Fitting, Compression Insert, Polypropylene, Valved, ¹/₄" OD, PMCD200412; 1/PK
    Quick-Disconnect Fitting, Compression Insert, Polypropylene, Valved, ¹/₄" OD, PMCD200412; 1/PK
    Valved
    Quick-Disconnect to Compression
    Straight
    PP
    ⅛"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $435.00
    $435.00
    Each (1UN)
       $435.00
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX06364-30
    Length
    3.683 cm
    Max. operating pressure
    120 psi
    Max. temperature
    71 °C (160 °F)
    Max. vacuum
    711 mm Hg (28" Hg)
    Seal material
    EPDM
    Sterilisation
    Gamma
    Valve spring material
    316 Stainless Steel
    Warranty
    3 Months
  • MFLX06364-32
    Quick-Disconnect Fitting, Compression Insert, Polypropylene, Straight-Through, ¹/₄" OD, PMC200412; 1/PK
    Quick-Disconnect Fitting, Compression Insert, Polypropylene, Straight-Through, ¹/₄" OD, PMC200412; 1/PK
    Open
    Quick-Disconnect to Compression
    Straight
    PP
    ⅛"
    Discontinued
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX06364-32
    Length
    2.921 cm
    Max. operating pressure
    120 psi
    Max. temperature
    71 °C (160 °F)
    Max. vacuum
    711 mm Hg (28" Hg)
    Seal material
    EPDM
    Sterilisation
    Gamma
    Valve spring material
    Warranty
    3 Months
  • MFLX06364-75
    Quick-Disconnect Fitting, Compression Insert, Polypropylene, Valved, ³/₈" OD, PLCD2000612; 1/PK
    Quick-Disconnect Fitting, Compression Insert, Polypropylene, Valved, ³/₈" OD, PLCD2000612; 1/PK
    Valved
    Quick-Disconnect to Compression
    Straight
    PP
    ¼"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $484.00
    $484.00
    Each (1UN)
       $484.00
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX06364-75
    Length
    4.2672 cm
    Max. operating pressure
    120 psi
    Max. temperature
    71 °C (160 °F)
    Max. vacuum
    711 mm Hg (28" Hg)
    Seal material
    EPDM
    Sterilisation
    Gamma
    Valve spring material
    316 Stainless Steel
    Warranty
    3 Months
  • MFLX31307-08
    Quick-Disconnect Fitting, Compression Insert, Panel-Mount, Acetal, Straight-Through, ⅛" Flow Size, ¼" OD; 25/Pk
    Quick-Disconnect Fitting, Compression Insert, Panel-Mount, Acetal, Straight-Through, ⅛" Flow Size, ¼" OD; 25/Pk
    Open
    Quick-Disconnect to Compression
    Straight
    POM
    ⅛"
    Discontinued
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX31307-08
    Length
    4.318 cm
    Max. operating pressure
    120 psi
    Max. temperature
    82 °C (180 °F)
    Max. vacuum
    711 mm Hg (28" Hg)
    Seal material
    Buna N
    Sterilisation
    Autoclave or ethylene oxide (ETO)
    Valve spring material
    Warranty
    1 year
  • MFLX31307-44
    Quick-Disconnect Fitting, Compression Insert, Panel-Mount, Acetal, Straight-Through, ⅜" OD; 25/Pk
    Quick-Disconnect Fitting, Compression Insert, Panel-Mount, Acetal, Straight-Through, ⅜" OD; 25/Pk
    Open
    Quick-Disconnect to Compression
    Straight
    POM
    ¼"
    Discontinued
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX31307-44
    Length
    4.8514 cm
    Max. operating pressure
    120 psi
    Max. temperature
    82 °C (180 °F)
    Max. vacuum
    711 mm Hg (28" Hg)
    Seal material
    Buna N
    Sterilisation
    Autoclave or ethylene oxide (ETO)
    Valve spring material
    Warranty
    1 year
  • MFLX31307-53
    Quick-Disconnect Fitting, Compression Insert, Panel-Mount, Acetal, Valved, ¼" OD; 25/Pk
    Quick-Disconnect Fitting, Compression Insert, Panel-Mount, Acetal, Valved, ¼" OD; 25/Pk
    Quick-Disconnect x Compression
    Valved
    Acetal
    ¼"
    Discontinued
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX31307-53
    Length
    4.5212 cm
    Max. operating pressure
    120 psi
    Max. temperature
    82 °C (180 °F)
    Max. vacuum
    711 mm Hg (28" Hg)
    Seal material
    Buna N
    Sterilisation
    Autoclave or ethylene oxide (ETO)
    Valve spring material
    316 Stainless Steel
    Warranty
    1 year
  • MFLX31307-54
    Quick-Disconnect Fitting, Compression Insert, Panel-Mount, Acetal, Valved, ⅜" OD; 25/Pk
    Quick-Disconnect Fitting, Compression Insert, Panel-Mount, Acetal, Valved, ⅜" OD; 25/Pk
    Quick-Disconnect x Compression
    Valved
    Acetal
    ¼"
    Discontinued
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX31307-54
    Length
    4.8514 cm
    Max. operating pressure
    120 psi
    Max. temperature
    82 °C (180 °F)
    Max. vacuum
    711 mm Hg (28" Hg)
    Seal material
    Buna N
    Sterilisation
    Autoclave or ethylene oxide (ETO)
    Valve spring material
    316 Stainless Steel
    Warranty
    1 year