Male Compression Run Tee
Acerca de este artículo
Get a secure seal at higher pressures.
- Constructed of chemically inert PFA
Compression fittings are an excellent choice to connect rigid tubing systems and withstand higher pressures than barbed fittings. Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal.
Choose PFA material for enhanced chemical resistance. Black polypropylene is an economical option that can be autoclaved.
6 Options Available Below
Product Details & Documents
Get a secure seal at higher pressures.
- Constructed of chemically inert PFA
Compression fittings are an excellent choice to connect rigid tubing systems and withstand higher pressures than barbed fittings. Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal.
Choose PFA material for enhanced chemical resistance. Black polypropylene is an economical option that can be autoclaved.
Specifications for Product Family
Especificaciones
- Orifice dimensions:
- 0.09375"
- 0.1875"
- 0.09375"
- 0.1875"
- 0.25"
- 0.28125"
- Sterilisation:
- Autoclave or ethylene oxide (ETO)
- Autoclave or ethylene oxide (ETO)
- Autoclave or ethylene oxide (ETO)
- Autoclave or ethylene oxide (ETO)
- Autoclave or ethylene oxide (ETO)
- Autoclave or ethylene oxide (ETO)
- Warranty:
- 18 months
- 18 months
- 18 months
- 18 months
- 18 months
- 18 months
Especificaciones
Product Family Options
Información del producto
- Connection TypeFlow patternFitting MaterialAvailabilityPrice
- MFLX31321-73Compression Run Tee, PFA, ⅛" OD X ¼" NPT(M)Compression Run Tee, PFA, ⅛" OD X ¼" NPT(M)Compression to ThreadTPFALog in Para disponibilidadEach (1UN)$2,161.00Especificaciones:
More
Specifications
Cat. No.MFLX31321-73Orifice dimensions0.09375"SterilisationAutoclave or ethylene oxide (ETO)Warranty18 months - MFLX31321-75Compression Run Tee, PFA, ¼" OD X ¼" NPT(M)Compression Run Tee, PFA, ¼" OD X ¼" NPT(M)Compression to ThreadTPFALog in Para disponibilidadEach (1UN)$1,425.00Especificaciones:
More
Specifications
Cat. No.MFLX31321-75Orifice dimensions0.1875"SterilisationAutoclave or ethylene oxide (ETO)Warranty18 months - MFLX31321-72Compression Run Tee, PFA, ⅛" OD×⅛" NPT(M)Compression Run Tee, PFA, ⅛" OD×⅛" NPT(M)Compression to ThreadTPFALog in Para disponibilidadEach (1UN)$1,316.00Especificaciones:
More
Specifications
Cat. No.MFLX31321-72Orifice dimensions0.09375"SterilisationAutoclave or ethylene oxide (ETO)Warranty18 months - MFLX31321-74Compression Run Tee, PFA, ¼" OD X ⅛" NPT(M)Compression Run Tee, PFA, ¼" OD X ⅛" NPT(M)Compression to ThreadTPFALog in Para disponibilidadEach (1UN)$1,425.00Especificaciones:
More
Specifications
Cat. No.MFLX31321-74Orifice dimensions0.1875"SterilisationAutoclave or ethylene oxide (ETO)Warranty18 months - MFLX31321-76Compression Run Tee, PFA, ⅜" OD×¼" NPT(M)Compression Run Tee, PFA, ⅜" OD×¼" NPT(M)Compression to ThreadTPFALog in Para disponibilidadEach (1UN)$1,535.00Especificaciones:
More
Specifications
Cat. No.MFLX31321-76Orifice dimensions0.25"SterilisationAutoclave or ethylene oxide (ETO)Warranty18 months - MFLX31321-77Compression Run Tee, PFA, ½" OD X ¼" NPT(M)Compression Run Tee, PFA, ½" OD X ¼" NPT(M)Compression to ThreadTPFALog in Para disponibilidadEach (1UN)$2,065.00Especificaciones:
More
Specifications
Cat. No.MFLX31321-77Orifice dimensions0.28125"SterilisationAutoclave or ethylene oxide (ETO)Warranty18 months