Orders LinkContactUsLinkComponent
Male Compression Run Tee
Male Compression Run Tee
Male Compression Run Tee
Certificates
Male Compression Run Tee
6 Options Available Below
Product Details & Documents
Get a secure seal at higher pressures.
  • Constructed of chemically inert PFA
Compression fittings are an excellent choice to connect rigid tubing systems and withstand higher pressures than barbed fittings. Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal.
Choose PFA material for enhanced chemical resistance. Black polypropylene is an economical option that can be autoclaved.
Specifications for Product Family
Especificaciones
  • Orifice dimensions:
  • 0.09375"
  • 0.1875"
  • 0.09375"
  • 0.1875"
  • 0.25"
  • 0.28125"
  • Sterilisation:
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Warranty:
  • 18 months
  • 18 months
  • 18 months
  • 18 months
  • 18 months
  • 18 months
Especificaciones
Product Family Options
Información del producto
  • Connection Type
    Flow pattern
    Fitting Material
    Availability
    Price
  • MFLX31321-73
    Compression Run Tee, PFA, ⅛" OD X ¼" NPT(M)
    Compression Run Tee, PFA, ⅛" OD X ¼" NPT(M)
    Compression to Thread
    T
    PFA
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $2,161.00
    $2,161.00
    Each (1UN)
       $2,161.00
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX31321-73
    Orifice dimensions
    0.09375"
    Sterilisation
    Autoclave or ethylene oxide (ETO)
    Warranty
    18 months
  • MFLX31321-75
    Compression Run Tee, PFA, ¼" OD X ¼" NPT(M)
    Compression Run Tee, PFA, ¼" OD X ¼" NPT(M)
    Compression to Thread
    T
    PFA
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $1,425.00
    $1,425.00
    Each (1UN)
       $1,425.00
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX31321-75
    Orifice dimensions
    0.1875"
    Sterilisation
    Autoclave or ethylene oxide (ETO)
    Warranty
    18 months
  • MFLX31321-72
    Compression Run Tee, PFA, ⅛" OD×⅛" NPT(M)
    Compression Run Tee, PFA, ⅛" OD×⅛" NPT(M)
    Compression to Thread
    T
    PFA
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $1,316.00
    $1,316.00
    Each (1UN)
       $1,316.00
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX31321-72
    Orifice dimensions
    0.09375"
    Sterilisation
    Autoclave or ethylene oxide (ETO)
    Warranty
    18 months
  • MFLX31321-74
    Compression Run Tee, PFA, ¼" OD X ⅛" NPT(M)
    Compression Run Tee, PFA, ¼" OD X ⅛" NPT(M)
    Compression to Thread
    T
    PFA
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $1,425.00
    $1,425.00
    Each (1UN)
       $1,425.00
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX31321-74
    Orifice dimensions
    0.1875"
    Sterilisation
    Autoclave or ethylene oxide (ETO)
    Warranty
    18 months
  • MFLX31321-76
    Compression Run Tee, PFA, ⅜" OD×¼" NPT(M)
    Compression Run Tee, PFA, ⅜" OD×¼" NPT(M)
    Compression to Thread
    T
    PFA
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $1,535.00
    $1,535.00
    Each (1UN)
       $1,535.00
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX31321-76
    Orifice dimensions
    0.25"
    Sterilisation
    Autoclave or ethylene oxide (ETO)
    Warranty
    18 months
  • MFLX31321-77
    Compression Run Tee, PFA, ½" OD X ¼" NPT(M)
    Compression Run Tee, PFA, ½" OD X ¼" NPT(M)
    Compression to Thread
    T
    PFA
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $2,065.00
    $2,065.00
    Each (1UN)
       $2,065.00
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX31321-77
    Orifice dimensions
    0.28125"
    Sterilisation
    Autoclave or ethylene oxide (ETO)
    Warranty
    18 months