Metric Size Straight Barbed Reducing Fittings
Ideal for low-pressure applications.
- Quick and easy assembly without tools
- Chemical resistant
Hose barb fittings make tubing connections quick and easy, requiring no tools for installation. Just slip your flexible tubing or hose onto the barb to create a secure seal.
- Environmentally Preferable:
- Catalog No:
- MFLX06458-60
- Orifice dimensions:
- 7 mm (0.28125")
- Sterilisation:
- Autoclave or ethylene oxide (ETO)
- Garantía:
- None
Frequently Bought Together