VWR® Hotplates
VWR® hotplate, 7×7", 115 V, 60 HZ, 750 W, Ceramic white, Ambient 5...400 °C, 17.8×17.8 cm (7×7"), 18W×30D×10.4H cm (7.1×11.8×4.1")
Catalog # 76549-914
Supplier: VWR International
Jump to:
Product Details & Documents
VWR® hotplates with temperature control up to 400 °C.
- White ceramic surface for excellent chemical resistance
- Digital speed control up to 400 °C
- Available with external temperature probe (displays actual sample temperature)
- 'HOT' LED safety indicator alerts user when plate is unsafe to touch
Corresponding magnetic hotplate stirrers (heating and stirring 76549-912, 76549-918, 76549-920 and 76549-926) or magnetic stirrers (stirring only 76549-916 and 76549-924) with ceramic plates are also available. Full specification details are given under separate entries, please view full details by searching for the relevant product code.
: CE marked.
Especificaciones
- Temperature Range:Ambient 5...400 °C
- Description:VWR® hotplate, 7×7"
- Electrical:115 V, 60 HZ, 750 W
- Overall Dimensions:18W×30D×10.4H cm (7.1×11.8×4.1")
- Top Plate Dimensions:17.8×17.8 cm (7×7")
- Top Plate Material:Ceramic white