Masterflex® Flow Indicators for Liquids, Avantor®
Easily view flow through clear plastic lens.
- Indicators feature a bright orange rotor that spins to provide visual verification
Flow indicators feature a paddle-wheel that spins at a rate proportional to the flow. Lens can be removed with a 1/4 turn for easy cleaning. Use with fluids that have a viscosity less than 34 centipoise (150 µm filter is r ecommended for protection).
Indicators/Sensors feature a Hall-effect sensor that produces a 4.5 to 24 VDC pulse output. Maximum current source output is 70 mA, and frequency output range is approximately 25 Hz at low flow to 225 Hz at high flow.
Indicators/Switches offer extra level of reliability and protection with a built-in switch. Each Single Pole Double Throw (SPDT) switch is rated for 1 amp, 24 VDC resistive. Repeatability is ±2% ; accuracy is ±5%. An LED lights when the set point is reached for easy field calibration.
Easily view flow through clear plastic lens.
- Indicators feature a bright orange rotor that spins to provide visual verification
Flow indicators feature a paddle-wheel that spins at a rate proportional to the flow. Lens can be removed with a 1/4 turn for easy cleaning. Use with fluids that have a viscosity less than 34 centipoise (150 µm filter is r ecommended for protection).
Indicators/Sensors feature a Hall-effect sensor that produces a 4.5 to 24 VDC pulse output. Maximum current source output is 70 mA, and frequency output range is approximately 25 Hz at low flow to 225 Hz at high flow.
Indicators/Switches offer extra level of reliability and protection with a built-in switch. Each Single Pole Double Throw (SPDT) switch is rated for 1 amp, 24 VDC resistive. Repeatability is ±2% ; accuracy is ±5%. An LED lights when the set point is reached for easy field calibration.
- Catalog No:
- MFLX32779-06
- MFLX32779-04
- MFLX32779-09
- MFLX32779-08
- MFLX32779-02
- MFLX32779-10
- MFLX32779-00
- MFLX32880-32
- Model:
- 32779-06
- 32779-04
- 32779-09
- 32779-08
- 32779-02
- 32779-10
- 32779-00
- 32880-32
- Min. operating temperature:
- –29 °C (–20 °F)
- –29 °C (–20 °F)
- –29 °C (–20 °F)
- –29 °C (–20 °F)
- –29 °C (–20 °F)
- Max. operating temperature:
- 100 °C (212 °F)
- 100 °C (212 °F)
- 100 °C (212 °F)
- 100 °C (212 °F)
- 82 °C (180 °F)
- 82 °C (180 °F)
- 82 °C (180 °F)
- 82 °C (180 °F)
- Wetted materials:
- Rotor pin: Ceramic; Lens: Polysulfone
- Rotor pin: Ceramic; Lens: Polysulfone
- Rotor pin: Ceramic; Lens: Polysulfone
- Rotor pin: Ceramic; Lens: Polysulfone
- Rotor pin: Ceramic; Lens: Polysulfone
- Rotor pin: Ceramic; Lens: Polysulfone
- O-Ring material:
- Viton®
- Viton®
- Viton®
- Viton®
- Buna-N
- Buna-N
- Display type:
- High Visibility Orange. Molded Nylon Rotor
- High Visibility Orange. Molded Nylon Rotor
- High Visibility Orange. Molded Nylon Rotor
- High Visibility Orange. Molded Nylon Rotor
- High Visibility Orange. Molded Nylon Rotor
- Warranty:
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
Product Family Options
- Housing MaterialPipe SizeDescriptionMaximum Flow Rate
- Brass½" NPT(F)Flow Indicator; Brass, 1/2" NPT(F), Orange Rotor20 gal./minEach (1UN)$6,420.001 year warranty
Specifications
Cat. No.MFLX32779-06Model32779-06Min. operating temperature–29 °C (–20 °F)Max. operating temperature100 °C (212 °F)Wetted materialsRotor pin: Ceramic; Lens: PolysulfoneO-Ring materialViton®Display typeHigh Visibility Orange. Molded Nylon RotorWarranty1 year1 year warranty - Brass¼" NPT(F)Flow Indicator; Brass, 1/4" NPT(F), Orange Rotor5 gal./minEach (1UN)$6,420.001 year warranty
Specifications
Cat. No.MFLX32779-04Model32779-04Min. operating temperature–29 °C (–20 °F)Max. operating temperature100 °C (212 °F)Wetted materialsRotor pin: Ceramic; Lens: PolysulfoneO-Ring materialViton®Display typeHigh Visibility Orange. Molded Nylon RotorWarranty1 year1 year warranty - Brass1" NPT(F)Flow Indicator; Brass, 1" NPT(F)30 gal./minEach (1UN)$6,440.001 year warranty
Specifications
Cat. No.MFLX32779-09Model32779-09Min. operating temperatureMax. operating temperature100 °C (212 °F)Wetted materialsRotor pin: Ceramic; Lens: PolysulfoneO-Ring materialViton®Display typeWarranty1 year1 year warranty - Brass¾" NPT(F)Flow Indicator; Brass, 3/4" NPT(F), Orange Rotor30 gal./minEach (1UN)$6,440.001 year warranty
Specifications
Cat. No.MFLX32779-08Model32779-08Min. operating temperature–29 °C (–20 °F)Max. operating temperature100 °C (212 °F)Wetted materialsRotor pin: Ceramic; Lens: PolysulfoneO-Ring materialViton®Display typeHigh Visibility Orange. Molded Nylon RotorWarranty1 year1 year warranty - PP½" NPT(F)Flow Indicator; Polypropylene, 1/2" NPT(F), Orange Rotor20 gal./minEach (1UN)$3,140.001 year warranty
Specifications
Cat. No.MFLX32779-02Model32779-02Min. operating temperature–29 °C (–20 °F)Max. operating temperature82 °C (180 °F)Wetted materialsRotor pin: Ceramic; Lens: PolysulfoneO-Ring materialBuna-NDisplay typeHigh Visibility Orange. Molded Nylon RotorWarranty1 year1 year warranty - ¼" NPT(F)Flow Indicator; Polypropylene, 1/4" NPT(F)5 gal./minEach (1UN)$5,190.00CE CompliantUL recognized
Specifications
Cat. No.MFLX32779-10Model32779-10Min. operating temperatureMax. operating temperature82 °C (180 °F)Wetted materialsO-Ring materialDisplay typeWarranty1 yearCE CompliantUL recognized - PP¼" NPT(F)Flow Indicator; Polypropylene, 1/4" NPT(F), Orange Rotor5 gal./minEach (1UN)$3,140.001 year warrantyCE Compliant
Specifications
Cat. No.MFLX32779-00Model32779-00Min. operating temperature–29 °C (–20 °F)Max. operating temperature82 °C (180 °F)Wetted materialsRotor pin: Ceramic; Lens: PolysulfoneO-Ring materialBuna-NDisplay typeHigh Visibility Orange. Molded Nylon RotorWarranty1 year1 year warrantyCE Compliant - ½" NPT(F)Flow Sensor/Switch; Polypropylene, 1/2" NPT(F), 24 VDC20 gal./minEach (1UN)$11,370.00
Specifications
Cat. No.MFLX32880-32Model32880-32Min. operating temperatureMax. operating temperature82 °C (180 °F)Wetted materialsO-Ring materialDisplay typeWarranty1 year