Orders LinkContactUsLinkComponent
Masterflex® Liquid Flow Switches for High Inline Pressures, Avantor®
Masterflex® Liquid Flow Switches for High Inline Pressures, Avantor®
Supplier:   VWR International
Masterflex® Liquid Flow Switches for High Inline Pressures, Avantor®
Supplier:  VWR International
Certificates
Masterflex® Liquid Flow Switches for High Inline Pressures, Avantor®
Masterflex® Liquid Flow Switches for High Inline Pressures, Avantor®
3 Options Available Below
Product Details & Documents
Ideal for high pressure applications.
  • Withstands up to 1500 psi
  • Effortlessly install with 3/8" NPT(M) connections
The one-piece magnetic PPS composite piston makes these normally open switches ideal for high pressure applications. The single pole single throw (SPST) switches withstand up to 1500 psi at 70 °F (21 °C)! Use a 100 µm filter to protect your switch. Switches include 24 to 26" lead wires.
Specifications for Product Family
Especificaciones
  • Model:
  • 32774-32
  • 32774-40
  • 32774-48
  • Max. pressure:
  • 1500 psi
  • 1500 psi
  • 1500 psi
  • Max. operating temperature:
  • 135 °C (275 °F)
  • 135 °C (275 °F)
  • 135 °C (275 °F)
  • Wetted materials:
  • Brass, PPS composite, 316 SS, and fluorocarbon
  • Stainless Steel, PPS composite, 316 SS, and fluorocarbon
  • Stainless Steel, PPS composite, 316 SS, and fluorocarbon
  • Process connection:
  • 0.375 NPT(M)
  • 0.375 NPT(M)
  • 0.375 NPT(M)
  • Height:
  • 40751"
  • 40751"
  • 40751"
  • Warranty:
  • 1 year
  • 1 year
  • 1 year
Especificaciones
Product Family Options
Información del producto
  • Maximum Flow Rate
    Housing Material
    Availability
    Price
  • MFLX32774-32
    Brass liquid flow switch for high inline pressures; 0.5 gal./min
    Brass liquid flow switch for high inline pressures; 0.5 gal./min
    0.5 gal./min
    Brass
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $6,480.00
    $6,480.00
    Each (1UN)
       $6,480.00
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX32774-32
    Model
    32774-32
    Max. pressure
    1500 psi
    Max. operating temperature
    135 °C (275 °F)
    Wetted materials
    Brass, PPS composite, 316 SS, and fluorocarbon
    Process connection
    0.375 NPT(M)
    Height
    40751"
    Warranty
    1 year
  • MFLX32774-40
    Stainless steel liquid flow switch for high inline pressures; 0.25 gal./min
    Stainless steel liquid flow switch for high inline pressures; 0.25 gal./min
    0.25 gal./min
    Stainless steel
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $14,550.00
    $14,550.00
    Each (1UN)
       $14,550.00
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX32774-40
    Model
    32774-40
    Max. pressure
    1500 psi
    Max. operating temperature
    135 °C (275 °F)
    Wetted materials
    Stainless Steel, PPS composite, 316 SS, and fluorocarbon
    Process connection
    0.375 NPT(M)
    Height
    40751"
    Warranty
    1 year
  • MFLX32774-48
    Stainless steel liquid flow switch for high inline pressures; 2.0 gal./min
    Stainless steel liquid flow switch for high inline pressures; 2.0 gal./min
    2 gal./min
    Stainless steel
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $15,100.00
    $15,100.00
    Each (1UN)
       $15,100.00
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX32774-48
    Model
    32774-48
    Max. pressure
    1500 psi
    Max. operating temperature
    135 °C (275 °F)
    Wetted materials
    Stainless Steel, PPS composite, 316 SS, and fluorocarbon
    Process connection
    0.375 NPT(M)
    Height
    40751"
    Warranty
    1 year