Masterflex® Normally Open Flow Switch for Low Pressure-Drop Liquids, Avantor®
Proveedor: VWR International
Certificados
Acerca de este artículo
Minimizes flow constriction.
- Provides low pressure drop
These SPST switches feature a unique, dual-diameter, internal bore and piston configuration to minimize flow constriction and provide low pressure drop. Use a 150 um filter to protect your switch.
Entrega: Includes 1/4" male quick connect terminals.
- Environmentally Preferable:
Especificaciones
- Presión máx.:
- 200 psi
- 200 psi
- 200 psi
- 200 psi
- Temperatura máximo de operación:
- 100 °C (212 °F)
- 100 °C (212 °F)
- 100 °C (212 °F)
- 100 °C (212 °F)
- Wetted materials:
- PP, Ryton® PPS, Viton®, and 316 SS
- PP, Ryton® PPS, Viton®, and 316 SS
- PP, Ryton® PPS, Viton®, and 316 SS
- PP, Ryton® PPS, Viton®, and 316 SS
- Conexión:
- 1/4" male quick connect terminals
- 1/4" male quick connect terminals
- 1/4" male quick connect terminals
- 1/4" male quick connect terminals
- Alimentación:
- 20 VA VAC
- 20 VA VAC
- 20 VA VAC
- 20 VA VAC
- Garantía:
- 1 year
- 1 year
- 1 year
- 1 year