Masterflex® L/S® High-Performance Pump Head Mounting Hardware, Avantor®
Proveedor: Avantor Fluid Handling
Certificados
Acerca de este artículo
Secure your Masterflex® High-Performance pump head!
- Constructed of durable stainless steel
Keep your pump head operating at its best with these replacement parts.
Entrega: Includes four 10-24 stainless steel mounting screws.
- Environmentally Preferable:
Frequently Bought Together