Orders LinkContactUsLinkComponent
Masterflex® L/S® Pump Head Mounting Hardware, Avantor®
Masterflex® L/S® Pump Head Mounting Hardware, Avantor®
Masterflex® L/S® Pump Head Mounting Hardware, Avantor®
Certificates
Masterflex® L/S® Pump Head Mounting Hardware, Avantor®
Masterflex® L/S® Pump Head Mounting Hardware, Avantor®
11 Options Available Below
Product Details & Documents
Easy stack multiple pump heads on your Masterflex® L/S® drive.
  • Constructed of durable stainless steel
  • See drive specifications for recommended number of heads
Stack multiple pump heads or replacement single pump head mounting hardware. Choose mounting hardware based on the Masterflex® L/S® pump head that you have. Compatible with the Masterflex L/S series pumps.
Product Family Options
Product Information
  • Material
    Compatible with
    Pump Series
    Availability
    Price
  • MFLX07013-04
    L/S® Pump head mounting hardware, Standard and Easy-Load®, one Pump Head, Stainless Steel; 2/Set, 2 years warranty
    L/S® Pump head mounting hardware, Standard and Easy-Load®, one Pump Head, Stainless Steel; 2/Set, 2 years warranty
    Stainless steel
    Masterflex L/S® Easy-Load® Pump head
    L/S
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $1,200.00
    $1,200.00
    Each (1UN)
       $1,200.00
    Supplier Number: 7013-04
  • MFLX07013-05
    L/S® Pump head mounting hardware, Standard and Easy-Load®, two pump heads, stainless steel; 2/set, 2 years warranty
    L/S® Pump head mounting hardware, Standard and Easy-Load®, two pump heads, stainless steel; 2/set, 2 years warranty
    Stainless steel
    Masterflex L/S® Easy-Load® Pump head
    L/S
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $1,290.00
    $1,290.00
    Each (1UN)
       $1,290.00
    Supplier Number: 7013-05
  • MFLX07013-08
    L/S® Pump head mounting hardware, Standard and Easy-Load®, three pump heads, stainless steel; 2/set, 2 years warranty
    L/S® Pump head mounting hardware, Standard and Easy-Load®, three pump heads, stainless steel; 2/set, 2 years warranty
    Stainless steel
    Masterflex L/S® Easy-Load® Pump head
    L/S
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $1,500.00
    $1,500.00
    Each (1UN)
       $1,500.00
    Supplier Number: 7013-08
  • MFLX07013-09
    L/S® Pump head mounting hardware, Standard and Easy-Load®, four pump heads, stainless steel; 2/set, 2 years warranty
    L/S® Pump head mounting hardware, Standard and Easy-Load®, four pump heads, stainless steel; 2/set, 2 years warranty
    Stainless steel
    Masterflex L/S® Easy-Load® Pump head
    L/S
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $1,660.00
    $1,660.00
    Each (1UN)
       $1,660.00
    Supplier Number: 7013-09
  • MFLX07514-04
    L/S® Easy-Load® Pump Head Mounting Hardware, Mounts One, Stainless Steel. 2 years warranty
    L/S® Easy-Load® Pump Head Mounting Hardware, Mounts One, Stainless Steel. 2 years warranty
    Stainless steel
    Masterflex L/S® Easy-Load® Pump head
    L/S
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $1,190.00
    $1,190.00
    Each (1UN)
       $1,190.00
    Supplier Number: 7514-04
  • MFLX07534-01
    L/S® Multichannel Pump Head Mounting Hardware, One Pump Head, Stainless Steel; 2/Set. 2 years warranty
    L/S® Multichannel Pump Head Mounting Hardware, One Pump Head, Stainless Steel; 2/Set. 2 years warranty
    Stainless steel
    Masterflex L/S® Multichannel pump head
    L/S
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $1,340.00
    $1,340.00
    Each (1UN)
       $1,340.00
    Supplier Number: 7534-01
  • MFLX77200-01
    L/S® Easy-Load® II Pump Head Mounting Hardware, One Pump Head, Stainless Steel; 2/Set. 2 years warranty
    L/S® Easy-Load® II Pump Head Mounting Hardware, One Pump Head, Stainless Steel; 2/Set. 2 years warranty
    Stainless steel
    Masterflex L/S® Easy-Load® II Pump head
    L/S
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $1,160.00
    $1,160.00
    Each (1UN)
       $1,160.00
    Supplier Number: 77200-01
  • MFLX77200-02
    L/S® Easy-Load® II Pump Head Mounting Hardware, Two Pump Heads, Stainless Steel; 2/Set. 2 years warranty
    L/S® Easy-Load® II Pump Head Mounting Hardware, Two Pump Heads, Stainless Steel; 2/Set. 2 years warranty
    Stainless steel
    Masterflex L/S® Easy-Load® II Pump head
    L/S
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $1,330.00
    $1,330.00
    Each (1UN)
       $1,330.00
    Supplier Number: 77200-02
  • MFLX77200-03
    L/S® Easy-Load® II Pump Head Mounting Hardware, Three Pump Heads, Stainless Steel; 2/Set. 2 years warranty
    L/S® Easy-Load® II Pump Head Mounting Hardware, Three Pump Heads, Stainless Steel; 2/Set. 2 years warranty
    Stainless steel
    Masterflex L/S® Easy-Load® II Pump head
    L/S
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $1,650.00
    $1,650.00
    Each (1UN)
       $1,650.00
    Supplier Number: 77200-03
  • MFLX77200-04
    L/S® Easy-Load® II Pump Head Mounting Hardware, Four Pump Heads, Stainless Steel. 2 years warranty
    L/S® Easy-Load® II Pump Head Mounting Hardware, Four Pump Heads, Stainless Steel. 2 years warranty
    Stainless steel
    Masterflex L/S® Easy-Load® II Pump head
    L/S
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $1,910.00
    $1,910.00
    Each (1UN)
       $1,910.00
    Supplier Number: 77200-04
  • MFLX77800-00
    L/S® Easy-Load® 3 Pump Head Mounting Hardware, Mounts One Pump Head
    L/S® Easy-Load® 3 Pump Head Mounting Hardware, Mounts One Pump Head
    Stainless steel
    Masterflex L/S® Easy-Load® 3 Pump head
    L/S
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $1,510.00
    $1,510.00
    Each (1UN)
       $1,510.00
    Supplier Number: 77800-00