Masterflex® Reducer Fittings, Male NPT Threaded, Straight, Avantor®
Quick and easy assembly.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimize downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Threaded fittings provide a more permanent fluid path. Use these fittings to reduce or enlarge connection sizes. Choose the size and material to meet your application needs.
- Environmentally Preferable:
- Catalog No:
- MFLX30623-38
- MFLX30623-32
- MFLX30623-46
- MFLX30623-42
- MFLX30623-48
- MFLX40627-35
- MFLX40627-43
- MFLX40623-47
- MFLX40623-39
- MFLX40623-55
- MFLX30623-50
- MFLX40623-43
- MFLX30623-40
- MFLX40623-51
- MFLX30623-54
- MFLX30623-52
- MFLX40623-35
- MFLX30627-54
- MFLX31701-89
- MFLX31701-88
- MFLX40615-31
- MFLX30623-36
- MFLX30623-34
- MFLX40627-55
- MFLX40615-32
- MFLX40615-34
- MFLX40615-33
- MFLX40615-35
- Modelo:
- 30623-38
- 30623-32
- 30623-46
- 30623-42
- 30623-48
- 40627-35
- 40627-43
- 40623-47
- 40623-39
- 40623-55
- 30623-50
- 40623-43
- 30623-40
- 40623-51
- 30623-54
- 30623-52
- 40623-35
- 30627-54
- 31701-89
- 31701-88
- 30623-36
- 30623-34
- 40627-55
- Sterilisation:
- Ethylene oxide (ETO) only
- Ethylene oxide (ETO) only
- Ethylene oxide (ETO) only
- Ethylene oxide (ETO) only
- Ethylene oxide (ETO) only
- Autoclave or ethylene oxide (ETO)
- Autoclave or ethylene oxide (ETO)
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) only
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) only
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) only
- Ethylene oxide (ETO) only
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) only
- Ethylene oxide (ETO) only
- Ethylene oxide (ETO) or gamma
- Autoclave or ethylene oxide (ETO)
- Ethylene oxide (ETO) only
- Ethylene oxide (ETO) only
- Autoclave or ethylene oxide (ETO)
- Autoclave or ethylene oxide (ETO)
- Autoclave or ethylene oxide (ETO)
- Autoclave or ethylene oxide (ETO)
- Autoclave or ethylene oxide (ETO)
- Garantía:
- 1 year
- 1 year
Frequently Bought Together