Masterflex® C/L® Power Supply for VDC Powered Variable-Speed Pump; 100 to 240 VAC
Catálogo # MFLX77120-11
Supplier: Avantor Fluid Handling
CE Compliant
Specifications
- Descripción:Accessory Replacement power supply, 115/230 VAC
- Garantía:1 year
- Referencia:MFLX77120-11
Specifications
About this item
Safely adapt your VDC powered C/L® pump.
- Suitable for use with C/L® pumps
Easily convert your 12 VDC C/L® Variable-Speed pumps to 100 to 240 VAC power. This power supply is suitable for use with 12 VDC models 77122-06, -16, and 26 single-channel pumps and models 77120-52 and -62 dual-channel pumps.
- Descripción:Accessory Replacement power supply, 115/230 VAC
- Environmentally Preferable:
- Garantía:1 year
- Referencia:MFLX77120-11