Masterflex® Compact Gas Mass Flow Sensors/Transmitters, Avantor®
Acerca de este artículo
Accurate and corrosion resistant.
- Nonclogging straight sensor tube provides high accuracy
- All models include a calibration certificate to NIST-traceable standards
Flow transmitters combine a restrictor flow element with an advanced straight sensor tube to provide high accuracy and repeatability. Gas flow rates are unaffected by temperature and pressure variants within limitations.
Select from aluminum or 316 stainless steel (SS) models. Wetted materials are anodized aluminum, brass, and Viton® for aluminum models; SS and Viton for SS models.
These precalibrated flow transmitters operate at inlet pressures between 5 and 60 psi and at gas temperatures between 59 and 77 °F (15 to 25 °C) while maintaining the stated percentage of full scale accuracy and linearity. When operating beyond 5 to 60 psi, add 0.01%/psi full scale; if operating beyond 59 to 77 °F (15 to 25 °C), add ±0.15%/ °C full scale.
12 Options Available Below
Product Details & Documents
Accurate and corrosion resistant.
- Nonclogging straight sensor tube provides high accuracy
- All models include a calibration certificate to NIST-traceable standards
Flow transmitters combine a restrictor flow element with an advanced straight sensor tube to provide high accuracy and repeatability. Gas flow rates are unaffected by temperature and pressure variants within limitations.
Select from aluminum or 316 stainless steel (SS) models. Wetted materials are anodized aluminum, brass, and Viton® for aluminum models; SS and Viton for SS models.
These precalibrated flow transmitters operate at inlet pressures between 5 and 60 psi and at gas temperatures between 59 and 77 °F (15 to 25 °C) while maintaining the stated percentage of full scale accuracy and linearity. When operating beyond 5 to 60 psi, add 0.01%/psi full scale; if operating beyond 59 to 77 °F (15 to 25 °C), add ±0.15%/ °C full scale.
Specifications for Product Family
Especificaciones
- Catalog No:
- MFLX32658-00
- MFLX32658-02
- MFLX32658-03
- MFLX32658-04
- MFLX32658-05
- MFLX32658-06
- MFLX32658-07
- MFLX32658-08
- MFLX32658-09
- MFLX32658-14
- MFLX32658-23
- MFLX32658-29
- Model:
- 32658-00
- 32658-02
- 32658-03
- 32658-04
- 32658-05
- 32658-06
- 32658-07
- 32658-08
- 32658-09
- 32658-14
- 32658-23
- 32658-29
- Accuracy:
- ±1.5% full-scale
- ±1.5% full-scale
- ±1.5% full-scale
- ±1.5% full-scale
- ±1.5% full-scale
- ±1.5% full-scale
- ±1.5% full-scale
- ±1.5% full-scale
- ±1.5% full-scale
- ±1.5% full-scale
- ±1.5% full-scale
- ±1.5% full-scale
- Certification:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Leak integrity:
- 1 X 10-7 cm³/s He (max)
- 1 X 10-7 cm³/s He (max)
- 1 X 10-7 cm³/s He (max)
- 1 X 10-7 cm³/s He (max)
- 1 X 10-7 cm³/s He (max)
- 1 X 10-7 cm³/s He (max)
- 1 X 10-7 cm³/s He (max)
- 1 X 10-7 cm³/s He (max)
- 1 X 10-7 cm³/s He (max)
- 1 X 10-7 cm³/s He (max)
- 1 X 10-7 cm³/s He (max)
- 1 X 10-7 cm³/s He (max)
- Max. air flow rate:
- 1 ml/min
- 50 ml/min
- 100 ml/min
- 200 ml/min
- 500 ml/min
- 100 ml/min
- Min. operating temperature:
- 0 °C (32 °F)
- 0 °C (32 °F)
- 0 °C (32 °F)
- 0 °C (32 °F)
- 0 °C (32 °F)
- 0 °C (32 °F)
- 0 °C (32 °F)
- 0 °C (32 °F)
- 0 °C (32 °F)
- 0 °C (32 °F)
- 0 °C (32 °F)
- 0 °C (32 °F)
- Max. operating temperature:
- 50 °C (122 °F)
- 50 °C (122 °F)
- 50 °C (122 °F)
- 50 °C (122 °F)
- 50 °C (122 °F)
- 50 °C (122 °F)
- 50 °C (122 °F)
- 50 °C (122 °F)
- 50 °C (122 °F)
- 50 °C (122 °F)
- 50 °C (122 °F)
- 50 °C (122 °F)
- Max. particulate size:
- 5 µm
- 5 µm
- 5 µm
- 5 µm
- 5 µm
- 5 µm
- 5 µm
- 5 µm
- 5 µm
- 5 µm
- 5 µm
- 5 µm
- Max. pressure:
- 1000 psi
- 1000 psi
- 1000 psi
- 1000 psi
- 1000 psi
- 1000 psi
- 1000 psi
- 1000 psi
- 1000 psi
- 1000 psi
- 1000 psi
- 1000 psi
- O-Ring material:
- Viton
- Viton
- Viton
- Viton
- Viton
- Viton
- Viton
- Viton
- Viton
- Viton
- Viton
- Viton
- Repeatability:
- ±0.5% full-scale
- ±0.5% full-scale
- ±0.5% full-scale
- ±0.5% full-scale
- ±0.5% full-scale
- ±0.5% full-scale
- ±0.5% full-scale
- ±0.5% full-scale
- ±0.5% full-scale
- ±0.5% full-scale
- ±0.5% full-scale
- ±0.5% full-scale
- Response time:
- 2000 ms
- 2000 ms
- 2000 ms
- 2000 ms
- 2000 ms
- 2000 ms
- 2000 ms
- 2000 ms
- 2000 ms
- 2000 ms
- 2000 ms
- 2000 ms
- Wetted materials:
- Anodized aluminum, brass, and Viton®
- Anodized aluminum, brass, and Viton®
- Anodized aluminum, brass, and Viton®
- Anodized aluminum, brass, and Viton®
- Anodized aluminum, brass, and Viton®
- Anodized aluminum, brass, and Viton®
- Anodized aluminum, brass, and Viton®
- Anodized aluminum, brass, and Viton®
- Anodized aluminum, brass, and Viton®
- Anodized aluminum, brass, and Viton®
- SS and Viton®
- SS and Viton®
- Warranty:
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
Especificaciones
Product Family Options
Información del producto
- Process ConnectionMin. flow rateMaximum Flow RateOutputMedia TypeHousing MaterialGas TypeFitting MaterialAvailabilityPrice
- MFLX32658-00Aluminum gas mass flow transmitter, 10 cm³/minAluminum gas mass flow transmitter, 10 cm³/min1/4" Compression0 ml/min0.01 L/minLinear 0 to 5 VDC (1000 Ω minimum load), 4 to 20 mA (50 to 250 Ω loop resistance)AirAnodized aluminiumAirBrassLog in Para disponibilidadEach (1UN)$12,990.00Supplier Number: GFMS-010207Especificaciones:CE Compliant
More
Specifications
Cat. No.MFLX32658-00Model32658-00Accuracy±1.5% full-scaleCertificationYesLeak integrity1 X 10-7 cm³/s He (max)Max. air flow rate1 ml/minMin. operating temperature0 °C (32 °F)Max. operating temperature50 °C (122 °F)Max. particulate size5 µmMax. pressure1000 psiO-Ring materialVitonRepeatability±0.5% full-scaleResponse time2000 msWetted materialsAnodized aluminum, brass, and Viton®Warranty1 yearCE Compliant - MFLX32658-02Aluminum gas mass flow transmitter, 50 cm³/minAluminum gas mass flow transmitter, 50 cm³/min1/4" Compression0 ml/min0.05 L/minLinear 0 to 5 VDC (1000 Ω minimum load), 4 to 20 mA (50 to 250 Ω loop resistance)AirAnodized aluminiumAirBrassLog in Para disponibilidadEach (1UN)$12,990.00Supplier Number: GFMS-010209Especificaciones:CE Compliant
More
Specifications
Cat. No.MFLX32658-02Model32658-02Accuracy±1.5% full-scaleCertificationYesLeak integrity1 X 10-7 cm³/s He (max)Max. air flow rate50 ml/minMin. operating temperature0 °C (32 °F)Max. operating temperature50 °C (122 °F)Max. particulate size5 µmMax. pressure1000 psiO-Ring materialVitonRepeatability±0.5% full-scaleResponse time2000 msWetted materialsAnodized aluminum, brass, and Viton®Warranty1 yearCE Compliant - MFLX32658-03Aluminum gas mass flow transmitter, 100 cm³/minAluminum gas mass flow transmitter, 100 cm³/min1/4" Compression0 ml/min0.1 L/minLinear 0 to 5 VDC (1000 Ω minimum load), 4 to 20 mA (50 to 250 Ω loop resistance)AirAnodized aluminiumAirBrassLog in Para disponibilidadEach (1UN)$12,990.00Supplier Number: GFMS-010210Especificaciones:CE Compliant
More
Specifications
Cat. No.MFLX32658-03Model32658-03Accuracy±1.5% full-scaleCertificationYesLeak integrity1 X 10-7 cm³/s He (max)Max. air flow rate100 ml/minMin. operating temperature0 °C (32 °F)Max. operating temperature50 °C (122 °F)Max. particulate size5 µmMax. pressure1000 psiO-Ring materialVitonRepeatability±0.5% full-scaleResponse time2000 msWetted materialsAnodized aluminum, brass, and Viton®Warranty1 yearCE Compliant - MFLX32658-04Aluminum gas mass flow transmitter, 200 cm³/minAluminum gas mass flow transmitter, 200 cm³/min1/4" Compression0 ml/min0.2 L/minLinear 0 to 5 VDC (1000 Ω minimum load), 4 to 20 mA (50 to 250 Ω loop resistance)AirAnodized aluminiumAirBrassLog in Para disponibilidadEach (1UN)$12,990.00Supplier Number: GFMS-010211Especificaciones:CE Compliant
More
Specifications
Cat. No.MFLX32658-04Model32658-04Accuracy±1.5% full-scaleCertificationYesLeak integrity1 X 10-7 cm³/s He (max)Max. air flow rate200 ml/minMin. operating temperature0 °C (32 °F)Max. operating temperature50 °C (122 °F)Max. particulate size5 µmMax. pressure1000 psiO-Ring materialVitonRepeatability±0.5% full-scaleResponse time2000 msWetted materialsAnodized aluminum, brass, and Viton®Warranty1 yearCE Compliant - MFLX32658-05Aluminum gas mass flow transmitter, 500 cm³/minAluminum gas mass flow transmitter, 500 cm³/min1/4" Compression0 ml/min0.5 L/minLinear 0 to 5 VDC (1000 Ω minimum load), 4 to 20 mA (50 to 250 Ω loop resistance)AirAnodized aluminiumAirBrassLog in Para disponibilidadEach (1UN)$12,990.00Supplier Number: GFMS-010212Especificaciones:CE Compliant
More
Specifications
Cat. No.MFLX32658-05Model32658-05Accuracy±1.5% full-scaleCertificationYesLeak integrity1 X 10-7 cm³/s He (max)Max. air flow rate500 ml/minMin. operating temperature0 °C (32 °F)Max. operating temperature50 °C (122 °F)Max. particulate size5 µmMax. pressure1000 psiO-Ring materialVitonRepeatability±0.5% full-scaleResponse time2000 msWetted materialsAnodized aluminum, brass, and Viton®Warranty1 yearCE Compliant - MFLX32658-06Aluminum gas mass flow transmitter, 1 sL/minAluminum gas mass flow transmitter, 1 sL/min1/4" Compression0 ml/min1 L/minLinear 0 to 5 VDC (1000 Ω minimum load), 4 to 20 mA (50 to 250 Ω loop resistance)AirAnodized aluminiumAirBrassLog in Para disponibilidadEach (1UN)$12,990.00Supplier Number: GFMS-010213Especificaciones:CE Compliant
More
Specifications
Cat. No.MFLX32658-06Model32658-06Accuracy±1.5% full-scaleCertificationYesLeak integrity1 X 10-7 cm³/s He (max)Max. air flow rateMin. operating temperature0 °C (32 °F)Max. operating temperature50 °C (122 °F)Max. particulate size5 µmMax. pressure1000 psiO-Ring materialVitonRepeatability±0.5% full-scaleResponse time2000 msWetted materialsAnodized aluminum, brass, and Viton®Warranty1 yearCE Compliant - MFLX32658-07Aluminum gas mass flow transmitter, 2 sL/minAluminum gas mass flow transmitter, 2 sL/min1/4" Compression0 ml/min2 L/minLinear 0 to 5 VDC (1000 Ω minimum load), 4 to 20 mA (50 to 250 Ω loop resistance)AirAnodized aluminiumAirBrassLog in Para disponibilidadEach (1UN)$12,990.00Supplier Number: GFMS-010214Especificaciones:CE Compliant
More
Specifications
Cat. No.MFLX32658-07Model32658-07Accuracy±1.5% full-scaleCertificationYesLeak integrity1 X 10-7 cm³/s He (max)Max. air flow rateMin. operating temperature0 °C (32 °F)Max. operating temperature50 °C (122 °F)Max. particulate size5 µmMax. pressure1000 psiO-Ring materialVitonRepeatability±0.5% full-scaleResponse time2000 msWetted materialsAnodized aluminum, brass, and Viton®Warranty1 yearCE Compliant - MFLX32658-08Aluminum gas mass flow transmitter, 5 sL/minAluminum gas mass flow transmitter, 5 sL/min1/4" Compression0 ml/min5 L/minLinear 0 to 5 VDC (1000 Ω minimum load), 4 to 20 mA (50 to 250 Ω loop resistance)AirAnodized aluminiumAirBrassLog in Para disponibilidadEach (1UN)$12,990.00Supplier Number: GFMS-010215Especificaciones:CE Compliant
More
Specifications
Cat. No.MFLX32658-08Model32658-08Accuracy±1.5% full-scaleCertificationYesLeak integrity1 X 10-7 cm³/s He (max)Max. air flow rateMin. operating temperature0 °C (32 °F)Max. operating temperature50 °C (122 °F)Max. particulate size5 µmMax. pressure1000 psiO-Ring materialVitonRepeatability±0.5% full-scaleResponse time2000 msWetted materialsAnodized aluminum, brass, and Viton®Warranty1 yearCE Compliant - MFLX32658-09Aluminum gas mass flow transmitter, 10 sL/minAluminum gas mass flow transmitter, 10 sL/min1/4" Compression0 ml/min10 L/minLinear 0 to 5 VDC (1000 Ω minimum load), 4 to 20 mA (50 to 250 Ω loop resistance)AirAnodized aluminiumAirBrassLog in Para disponibilidadEach (1UN)$12,990.00Supplier Number: GFMS-010216Especificaciones:CE Compliant
More
Specifications
Cat. No.MFLX32658-09Model32658-09Accuracy±1.5% full-scaleCertificationYesLeak integrity1 X 10-7 cm³/s He (max)Max. air flow rateMin. operating temperature0 °C (32 °F)Max. operating temperature50 °C (122 °F)Max. particulate size5 µmMax. pressure1000 psiO-Ring materialVitonRepeatability±0.5% full-scaleResponse time2000 msWetted materialsAnodized aluminum, brass, and Viton®Warranty1 yearCE Compliant - MFLX32658-14Aluminum gas mass flow transmitter, 50 sL/minAluminum gas mass flow transmitter, 50 sL/min1/4" Compression0 ml/min50 L/minLinear 0 to 5 VDC (1000 Ω minimum load), 4 to 20 mA (50 to 250 Ω loop resistance)AirAnodized aluminiumAirBrassLog in Para disponibilidadEach (1UN)$16,890.00Supplier Number: GFMS-010221Especificaciones:CE Compliant
More
Specifications
Cat. No.MFLX32658-14Model32658-14Accuracy±1.5% full-scaleCertificationYesLeak integrity1 X 10-7 cm³/s He (max)Max. air flow rateMin. operating temperature0 °C (32 °F)Max. operating temperature50 °C (122 °F)Max. particulate size5 µmMax. pressure1000 psiO-Ring materialVitonRepeatability±0.5% full-scaleResponse time2000 msWetted materialsAnodized aluminum, brass, and Viton®Warranty1 yearCE Compliant - MFLX32658-23316 stainless steel gas mass flow transmitter, 100 cm³/min316 stainless steel gas mass flow transmitter, 100 cm³/min1/4" Compression0 ml/min0.1 L/minLinear 0 to 5 VDC (1000 Ω minimum load), 4 to 20 mA (50 to 250 Ω loop resistance)AirStainless steelAirStainless steelLog in Para disponibilidadEach (1UN)$21,450.00Supplier Number: GFMS-010228Especificaciones:CE Compliant
More
Specifications
Cat. No.MFLX32658-23Model32658-23Accuracy±1.5% full-scaleCertificationYesLeak integrity1 X 10-7 cm³/s He (max)Max. air flow rate100 ml/minMin. operating temperature0 °C (32 °F)Max. operating temperature50 °C (122 °F)Max. particulate size5 µmMax. pressure1000 psiO-Ring materialVitonRepeatability±0.5% full-scaleResponse time2000 msWetted materialsSS and Viton®Warranty1 yearCE Compliant - MFLX32658-29316 stainless steel gas mass flow transmitter, 10 sL/min316 stainless steel gas mass flow transmitter, 10 sL/min1/4" Compression0 ml/min10 L/minLinear 0 to 5 VDC (1000 Ω minimum load), 4 to 20 mA (50 to 250 Ω loop resistance)AirStainless steelAirStainless steelLog in Para disponibilidadEach (1UN)$21,450.00Supplier Number: GFMS-010234Especificaciones:CE Compliant
More
Specifications
Cat. No.MFLX32658-29Model32658-29Accuracy±1.5% full-scaleCertificationYesLeak integrity1 X 10-7 cm³/s He (max)Max. air flow rateMin. operating temperature0 °C (32 °F)Max. operating temperature50 °C (122 °F)Max. particulate size5 µmMax. pressure1000 psiO-Ring materialVitonRepeatability±0.5% full-scaleResponse time2000 msWetted materialsSS and Viton®Warranty1 yearCE Compliant
Accessories
Información del producto
- Catalog #ElectricalAvailabilityPrice
- Mass flowmeter power supply/adapterMass flowmeter power supply/adapterMFLX03277-05220 VACLog in Para disponibilidadEach (1UN) - $1,500.00
Información del producto
- Catalog #AvailabilityPrice
- Output cable for 4 to 20 mA signal
Información del producto
- Catalog #AvailabilityPrice
- Output cable for 0 to 5 VDC signal
Información del producto
- Catalog #ElectricalAvailabilityPrice
- Mass flowmeter power supply/adapterMass flowmeter power supply/adapterMFLX03277-00110 VACLog in Para disponibilidadEach (1UN) - $1,070.00