Masterflex® Variable-Area Flowmeter Acrylic Tripod Bases, Avantor®
Easily mount flowmeters on tripod for easy viewing.
- Constructed of durable acrylic
These tripod bases are ideal for benchtop applications. A tripod base is required to use the multitube frame with the common inlet flow pattern; all other multitube frames are designed for panel and tripod mounting. Tripod bases are made of acrylic and include a spirit leveler.
- Environmentally Preferable:
- Catalog No:
- MFLX03218-58
- MFLX03226-40
- MFLX03226-50
- Modelo:
- 03218-58
- 03226-40
- 03226-50
- Ancho:
- 1755"
- 1755"
- 1755"
- Garantía:
- 1 year
- 1 year
- 1 year
Frequently Bought Together