Orders LinkContactUsLinkComponent
Masterflex® Variable-Area Flowmeter Valve Cartridges, Avantor®
Masterflex® Variable-Area Flowmeter Valve Cartridges, Avantor®
Supplier:   VWR International
Masterflex® Variable-Area Flowmeter Valve Cartridges, Avantor®
Supplier:  VWR International
Certificates
Masterflex® Variable-Area Flowmeter Valve Cartridges, Avantor®
Masterflex® Variable-Area Flowmeter Valve Cartridges, Avantor®
16 Options Available Below
Product Details & Documents
Control your flow with the addition of a valve.
  • Aluminum is economical and good for general use with noncorrosive gases and liquids
  • 316 Stainless steel (SS) withstands higher temperatures and pressures, and features excellent chemical compatibility
Select a valve-cartridge that matches your frame. Insert these needle valves into the flowmeter frames to control flow rate. Choose a 16-turn, high-resolution metering valve-cartridge for greater precision of flow rate. If your flowtube is in units other than ml/min, convert them to ml/min and choose a valve within that flow range from the table below. See website and virtual catalog page for more details.
Specifications for Product Family
Specifications
  • Model:
  • 03217-82
  • 03217-84
  • 03217-86
  • 03217-88
  • 03217-92
  • 03217-96
  • 03218-61
  • 03218-62
  • 03218-63
  • 03218-64
  • 03218-67
  • 03218-68
  • 03218-69
  • 03218-70
  • 03218-72
  • 03218-74
  • Min. air flow rate:
  • 5000 ml/min
  • 20000 ml/min
  • 60001 ml/min
  • 20000 ml/min
  • 5000 ml/min
  • 60001 ml/min
  • 201 ml/min
  • 1001 ml/min
  • 2501 ml/min
  • 6201 ml/min
  • 201 ml/min
  • 1001 ml/min
  • 2501 ml/min
  • 6201 ml/min
  • 0 ml/min
  • 0 ml/min
  • Max. air flow rate:
  • 20000 ml/min
  • 60000 ml/min
  • 60001 ml/min
  • 60000 ml/min
  • 20000 ml/min
  • 600001 ml/min
  • 1000 ml/min
  • 2500 ml/min
  • 6200 ml/min
  • 6201 ml/min
  • 1000 ml/min
  • 2500 ml/min
  • 6200 ml/min
  • 6201 ml/min
  • 200 ml/min
  • 200 ml/min
  • Min. water flow rate:
  • 0 ml/min
  • 0 ml/min
  • 0 ml/min
  • 0 ml/min
  • 0 ml/min
  • 0 ml/min
  • 3.1 ml/min
  • 30 ml/min
  • 71 ml/min
  • 650 ml/min
  • 3.1 ml/min
  • 30 ml/min
  • 71 ml/min
  • 650 ml/min
  • 0 ml/min
  • 0 ml/min
  • Max. water flow rate:
  • 350 ml/min
  • 1200 ml/min
  • 3500 ml/min
  • 1200 ml/min
  • 350 ml/min
  • 3500 ml/min
  • 30 ml/min
  • 70 ml/min
  • 200 ml/min
  • 650 ml/min
  • 30 ml/min
  • 70 ml/min
  • 200 ml/min
  • 650 ml/min
  • 6 ml/min
  • 6 ml/min
  • O-Ring material:
  • Viton®
  • Viton®
  • Viton®
  • Buna-N
  • Buna-N
  • Buna-N
  • Buna-N
  • Buna-N
  • Buna-N
  • Buna-N
  • Viton®
  • Viton®
  • Viton®
  • Viton®
  • Buna-N
  • Viton®
  • Width:
  • 1754"
  • 1754"
  • 1754"
  • 1754"
  • 1754"
  • 1754"
  • 1754"
  • 1754"
  • 1754"
  • 1754"
  • 1754"
  • 1754"
  • 1754"
  • 1754"
  • 1754"
  • 1754"
  • Warranty:
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
Specifications
Product Family Options
Product Information
  • Maximum Flow Rate
    Housing Material
    Availability
    Price
  • MFLX03217-82
    Variable-Area Flowmeter Valve Cartridge, 10-Turn, for Stainless Steel Frame Systems, Low Flow
    Variable-Area Flowmeter Valve Cartridge, 10-Turn, for Stainless Steel Frame Systems, Low Flow
    20000 ml/min
    316 stainless steel
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $1,430.00
    $1,430.00
    Each (1UN)
       $1,430.00
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX03217-82
    Model
    03217-82
    Min. air flow rate
    5000 ml/min
    Max. air flow rate
    20000 ml/min
    Min. water flow rate
    0 ml/min
    Max. water flow rate
    350 ml/min
    O-Ring material
    Viton®
    Width
    1754"
    Warranty
    1 year
  • MFLX03217-84
    Variable-Area Flowmeter Valve Cartridge, 10-Turn, for Stainless Steel Frame Systems, Medium Flow
    Variable-Area Flowmeter Valve Cartridge, 10-Turn, for Stainless Steel Frame Systems, Medium Flow
    60000 ml/min
    316 stainless steel
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $1,430.00
    $1,430.00
    Each (1UN)
       $1,430.00
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX03217-84
    Model
    03217-84
    Min. air flow rate
    20000 ml/min
    Max. air flow rate
    60000 ml/min
    Min. water flow rate
    0 ml/min
    Max. water flow rate
    1200 ml/min
    O-Ring material
    Viton®
    Width
    1754"
    Warranty
    1 year
  • MFLX03217-86
    Variable-Area Flowmeter Valve Cartridge, 10-Turn, for Stainless Steel Frame Systems, High Flow
    Variable-Area Flowmeter Valve Cartridge, 10-Turn, for Stainless Steel Frame Systems, High Flow
    60001 ml/min
    316 stainless steel
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $1,430.00
    $1,430.00
    Each (1UN)
       $1,430.00
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX03217-86
    Model
    03217-86
    Min. air flow rate
    60001 ml/min
    Max. air flow rate
    60001 ml/min
    Min. water flow rate
    0 ml/min
    Max. water flow rate
    3500 ml/min
    O-Ring material
    Viton®
    Width
    1754"
    Warranty
    1 year
  • MFLX03217-88
    Variable-Area Flowmeter Valve Cartridge, 10-Turn, for Aluminum Frame Systems, Medium Flow
    Variable-Area Flowmeter Valve Cartridge, 10-Turn, for Aluminum Frame Systems, Medium Flow
    60000 ml/min
    Aluminium
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $1,080.00
    $1,080.00
    Each (1UN)
       $1,080.00
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX03217-88
    Model
    03217-88
    Min. air flow rate
    20000 ml/min
    Max. air flow rate
    60000 ml/min
    Min. water flow rate
    0 ml/min
    Max. water flow rate
    1200 ml/min
    O-Ring material
    Buna-N
    Width
    1754"
    Warranty
    1 year
  • MFLX03217-92
    Variable-Area Flowmeter Valve Cartridge, 10-Turn, for Aluminum Frame Systems, Low Flow
    Variable-Area Flowmeter Valve Cartridge, 10-Turn, for Aluminum Frame Systems, Low Flow
    20000 ml/min
    Aluminium
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $1,080.00
    $1,080.00
    Each (1UN)
       $1,080.00
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX03217-92
    Model
    03217-92
    Min. air flow rate
    5000 ml/min
    Max. air flow rate
    20000 ml/min
    Min. water flow rate
    0 ml/min
    Max. water flow rate
    350 ml/min
    O-Ring material
    Buna-N
    Width
    1754"
    Warranty
    1 year
  • MFLX03217-96
    Variable-Area Flowmeter Valve Cartridge, 10-Turn, for Aluminum Frame Systems, High Flow
    Variable-Area Flowmeter Valve Cartridge, 10-Turn, for Aluminum Frame Systems, High Flow
    600001 ml/min
    Aluminium
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $1,080.00
    $1,080.00
    Each (1UN)
       $1,080.00
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX03217-96
    Model
    03217-96
    Min. air flow rate
    60001 ml/min
    Max. air flow rate
    600001 ml/min
    Min. water flow rate
    0 ml/min
    Max. water flow rate
    3500 ml/min
    O-Ring material
    Buna-N
    Width
    1754"
    Warranty
    1 year
  • MFLX03218-61
    Variable-Area Flowmeter Valve Cartridge, 16-Turn, for Aluminum Frame Systems, Range 3
    Variable-Area Flowmeter Valve Cartridge, 16-Turn, for Aluminum Frame Systems, Range 3
    1000 ml/min
    Aluminium
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $1,840.00
    $1,840.00
    Each (1UN)
       $1,840.00
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX03218-61
    Model
    03218-61
    Min. air flow rate
    201 ml/min
    Max. air flow rate
    1000 ml/min
    Min. water flow rate
    3.1 ml/min
    Max. water flow rate
    30 ml/min
    O-Ring material
    Buna-N
    Width
    1754"
    Warranty
    1 year
  • MFLX03218-62
    Variable-Area Flowmeter Valve Cartridge, 16-Turn, for Aluminum Frame Systems, Range 4
    Variable-Area Flowmeter Valve Cartridge, 16-Turn, for Aluminum Frame Systems, Range 4
    2500 ml/min
    Aluminium
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $1,840.00
    $1,840.00
    Each (1UN)
       $1,840.00
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX03218-62
    Model
    03218-62
    Min. air flow rate
    1001 ml/min
    Max. air flow rate
    2500 ml/min
    Min. water flow rate
    30 ml/min
    Max. water flow rate
    70 ml/min
    O-Ring material
    Buna-N
    Width
    1754"
    Warranty
    1 year
  • MFLX03218-63
    Variable-Area Flowmeter Valve Cartridge, 16-Turn, for Aluminum Frame Systems, Range 5
    Variable-Area Flowmeter Valve Cartridge, 16-Turn, for Aluminum Frame Systems, Range 5
    6200 ml/min
    Aluminium
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $1,840.00
    $1,840.00
    Each (1UN)
       $1,840.00
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX03218-63
    Model
    03218-63
    Min. air flow rate
    2501 ml/min
    Max. air flow rate
    6200 ml/min
    Min. water flow rate
    71 ml/min
    Max. water flow rate
    200 ml/min
    O-Ring material
    Buna-N
    Width
    1754"
    Warranty
    1 year
  • MFLX03218-64
    Variable-Area Flowmeter Valve Cartridge, 16-Turn, for Aluminum Frame Systems, Range 6
    Variable-Area Flowmeter Valve Cartridge, 16-Turn, for Aluminum Frame Systems, Range 6
    6201 ml/min
    Aluminium
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $1,840.00
    $1,840.00
    Each (1UN)
       $1,840.00
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX03218-64
    Model
    03218-64
    Min. air flow rate
    6201 ml/min
    Max. air flow rate
    6201 ml/min
    Min. water flow rate
    650 ml/min
    Max. water flow rate
    650 ml/min
    O-Ring material
    Buna-N
    Width
    1754"
    Warranty
    1 year
  • MFLX03218-67
    Variable-Area Flowmeter Valve Cartridge, 16-Turn, for Stainless Steel Frame Systems, Range 3
    Variable-Area Flowmeter Valve Cartridge, 16-Turn, for Stainless Steel Frame Systems, Range 3
    1000 ml/min
    316 stainless steel
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $3,420.00
    $3,420.00
    Each (1UN)
       $3,420.00
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX03218-67
    Model
    03218-67
    Min. air flow rate
    201 ml/min
    Max. air flow rate
    1000 ml/min
    Min. water flow rate
    3.1 ml/min
    Max. water flow rate
    30 ml/min
    O-Ring material
    Viton®
    Width
    1754"
    Warranty
    1 year
  • MFLX03218-68
    Variable-Area Flowmeter Valve Cartridge, 16-Turn, for Stainless Steel Frame Systems, Range 4
    Variable-Area Flowmeter Valve Cartridge, 16-Turn, for Stainless Steel Frame Systems, Range 4
    2500 ml/min
    316 stainless steel
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $3,420.00
    $3,420.00
    Each (1UN)
       $3,420.00
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX03218-68
    Model
    03218-68
    Min. air flow rate
    1001 ml/min
    Max. air flow rate
    2500 ml/min
    Min. water flow rate
    30 ml/min
    Max. water flow rate
    70 ml/min
    O-Ring material
    Viton®
    Width
    1754"
    Warranty
    1 year
  • MFLX03218-69
    Variable-Area Flowmeter Valve Cartridge, 16-Turn, for Stainless Steel Frame Systems, Range 5
    Variable-Area Flowmeter Valve Cartridge, 16-Turn, for Stainless Steel Frame Systems, Range 5
    6200 ml/min
    316 stainless steel
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $3,420.00
    $3,420.00
    Each (1UN)
       $3,420.00
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX03218-69
    Model
    03218-69
    Min. air flow rate
    2501 ml/min
    Max. air flow rate
    6200 ml/min
    Min. water flow rate
    71 ml/min
    Max. water flow rate
    200 ml/min
    O-Ring material
    Viton®
    Width
    1754"
    Warranty
    1 year
  • MFLX03218-70
    Variable-Area Flowmeter Valve Cartridge, 16-Turn, for Stainless Steel Frame Systems, Range 6
    Variable-Area Flowmeter Valve Cartridge, 16-Turn, for Stainless Steel Frame Systems, Range 6
    6201 ml/min
    316 stainless steel
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $3,420.00
    $3,420.00
    Each (1UN)
       $3,420.00
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX03218-70
    Model
    03218-70
    Min. air flow rate
    6201 ml/min
    Max. air flow rate
    6201 ml/min
    Min. water flow rate
    650 ml/min
    Max. water flow rate
    650 ml/min
    O-Ring material
    Viton®
    Width
    1754"
    Warranty
    1 year
  • MFLX03218-72
    Variable-Area Flowmeter Valve Cartridge, 16-Turn, for Aluminum Frame Systems, Range 1
    Variable-Area Flowmeter Valve Cartridge, 16-Turn, for Aluminum Frame Systems, Range 1
    200 ml/min
    Aluminium
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $1,840.00
    $1,840.00
    Each (1UN)
       $1,840.00
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX03218-72
    Model
    03218-72
    Min. air flow rate
    0 ml/min
    Max. air flow rate
    200 ml/min
    Min. water flow rate
    0 ml/min
    Max. water flow rate
    6 ml/min
    O-Ring material
    Buna-N
    Width
    1754"
    Warranty
    1 year
  • MFLX03218-74
    Variable-Area Flowmeter Valve Cartridge, 16-Turn, for Stainless Steel Frame Systems, Range 1
    Variable-Area Flowmeter Valve Cartridge, 16-Turn, for Stainless Steel Frame Systems, Range 1
    200 ml/min
    316 stainless steel
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $3,420.00
    $3,420.00
    Each (1UN)
       $3,420.00
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX03218-74
    Model
    03218-74
    Min. air flow rate
    0 ml/min
    Max. air flow rate
    200 ml/min
    Min. water flow rate
    0 ml/min
    Max. water flow rate
    6 ml/min
    O-Ring material
    Viton®
    Width
    1754"
    Warranty