Orders LinkContactUsLinkComponent
Masterflex® L/S® Precision Modular Drives with Benchtop Controller, Avantor®
Masterflex® L/S® Precision Modular Drives with Benchtop Controller, Avantor®
Certificates
Masterflex® L/S® Precision Modular Drives with Benchtop Controller, Avantor®
Masterflex® L/S® Precision Modular Drives with Benchtop Controller, Avantor®
2 Options Available Below
Product Details & Documents
Place where convenient—ideal for limited spaces, hoods, or isolation chambers.
  • Flow rate 0.06 to 3400 ml/min with L/S tubing—flow rate depends on drive rpm and tubing size
  • Three digit LED display shows motor rpm — confirm speed setting at a glance
  • Reversible motor—purge tubing before or after pumping; pump fluid in either direction
  • LED indicators confirm motor direction
  • Splash-resistant ABS plastic controller housing is IP33
  • rated
  • Remote control capabilities via a DB9 female connector on back of controller
L/S® modular drives feature reversible pumping and allow you the flexibility to separate the motor from the controller. These accurate modular drives have powerful motors, speed display, and ±0.25% speed control for precise flow regulation—ideal for metering applications.
 : Includes 6-ft (1.8-m) line cord with IEC 320/CEE22 socket. These models are shipped with country-specific plug/cord set; please specify ultimate destination when ordering.
Specifications for Product Family
Specifications
  • Model:
  • 07557-00
  • 07557-10
  • Cloud connected:
  • No
  • No
  • Controller height:
  • 140 mm (5.5")
  • 140 mm (5.5")
  • Controller IP rating:
  • IP 33
  • IP 33
  • Controller length:
  • 263 mm (10.3437")
  • 263 mm (10.3437")
  • Controller width:
  • 211 mm (8.3281")
  • 211 mm (8.3281")
  • Control type:
  • Digital Variable Speed
  • Digital Variable Speed
  • Digital dispensing:
  • No
  • No
  • Drive IP rating:
  • IP 33
  • IP 33
  • Min. RPM:
  • 6 rpm
  • 1 rpm
  • Motor:
  • 75 W
  • 75 W
  • Motor height:
  • 115 mm (4.5")
  • 115 mm (4.5")
  • Motor hp:
  • 1/10
  • 1/10
  • Motor IP rating:
  • IP 33
  • IP 33
  • Motor length:
  • 273 mm (10.75")
  • 273 mm (10.75")
  • Motor width:
  • 97 mm (3.7968")
  • 97 mm (3.7968")
  • Open-head sensor:
  • No
  • No
  • Pump series:
  • L/S
  • L/S
  • Remote I/O:
  • 4 to 20 mA. 0 to 10 V. start/stop and direction via contact closure
  • 4 to 20 mA. 0 to 10 V. start/stop and direction via contact closure
  • Reversible motor:
  • Yes
  • Yes
  • Speed:
  • Variable
  • Variable
  • Speed control:
  • 0.25%
  • 0.25%
  • Type of pump heads accepted:
  • Standard, Easy-Load, Easy-Load II, Easy-Load 3, High Performance, Cartridge, Multichannel, PTFE Tubing, PTFE Diaphragm, Cytoflow
  • Standard, Easy-Load, Easy-Load II, Easy-Load 3, High Performance, Cartridge, Multichannel, PTFE Tubing, PTFE Diaphragm, Cytoflow
  • Washdown capable:
  • No
  • No
  • Current:
  • 2.2 A
  • 2.2 A
  • Frequency:
  • 50/60 Hz
  • 50/60 Hz
  • Voltage:
  • 90 - 260 VAC
  • 90 - 260 VAC
  • Warranty:
  • 2 years
  • 2 years
Specifications
Product Family Options
US plug models
Product Information
  • Plug Type
    Maximum Flow Rate
    No. of Pump Heads Accepted
    Housing Material
    Max. RPM
    Power
    Availability
    Price
  • MFLX07557-00
    L/S Precision Modular Drive w/ Remote I/O; 600 rpm
    L/S Precision Modular Drive w/ Remote I/O; 600 rpm
    US
    3400 ml/min
    2
    ABS plastic and aluminium controller; painted steel drive
    600 rpm
    90 - 260 VAC; 50/60 Hz; 2.2 A
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $45,440.00
    $45,440.00
    Each (1UN)
       $45,440.00
    Supplier Number: 7557-00
    Specifications:
    2 year warranty
    CE Compliant
    More
    Specifications
    Cat. No.
    MFLX07557-00
    Model
    07557-00
    Cloud connected
    No
    Controller height
    140 mm (5.5")
    Controller IP rating
    IP 33
    Controller length
    263 mm (10.3437")
    Controller width
    211 mm (8.3281")
    Control type
    Digital Variable Speed
    Digital dispensing
    No
    Drive IP rating
    IP 33
    Min. RPM
    6 rpm
    Motor
    75 W
    Motor height
    115 mm (4.5")
    Motor hp
    1/10
    Motor IP rating
    IP 33
    Motor length
    273 mm (10.75")
    Motor width
    97 mm (3.7968")
    Open-head sensor
    No
    Pump series
    L/S
    Remote I/O
    4 to 20 mA. 0 to 10 V. start/stop and direction via contact closure
    Reversible motor
    Yes
    Speed
    Variable
    Speed control
    0.25%
    Type of pump heads accepted
    Standard, Easy-Load, Easy-Load II, Easy-Load 3, High Performance, Cartridge, Multichannel, PTFE Tubing, PTFE Diaphragm, Cytoflow
    Washdown capable
    No
    Current
    2.2 A
    Frequency
    50/60 Hz
    Voltage
    90 - 260 VAC
    Warranty
    2 years
    2 year warranty
    CE Compliant
  • MFLX07557-10
    L/S Precision Modular Drive w/ Remote I/O; 100 rpm
    L/S Precision Modular Drive w/ Remote I/O; 100 rpm
    US
    580 ml/min
    4
    ABS plastic and aluminium controller; painted steel drive
    100 rpm
    90 - 260 VAC; 50/60 Hz; 2.2 A
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $45,440.00
    $45,440.00
    Each (1UN)
       $45,440.00
    Supplier Number: 7557-10
    Specifications:
    2 year warranty
    CE Compliant
    More
    Specifications
    Cat. No.
    MFLX07557-10
    Model
    07557-10
    Cloud connected
    No
    Controller height
    140 mm (5.5")
    Controller IP rating
    IP 33
    Controller length
    263 mm (10.3437")
    Controller width
    211 mm (8.3281")
    Control type
    Digital Variable Speed
    Digital dispensing
    No
    Drive IP rating
    IP 33
    Min. RPM
    1 rpm
    Motor
    75 W
    Motor height
    115 mm (4.5")
    Motor hp
    1/10
    Motor IP rating
    IP 33
    Motor length
    273 mm (10.75")
    Motor width
    97 mm (3.7968")
    Open-head sensor
    No
    Pump series
    L/S
    Remote I/O
    4 to 20 mA. 0 to 10 V. start/stop and direction via contact closure
    Reversible motor
    Yes
    Speed
    Variable
    Speed control
    0.25%
    Type of pump heads accepted
    Standard, Easy-Load, Easy-Load II, Easy-Load 3, High Performance, Cartridge, Multichannel, PTFE Tubing, PTFE Diaphragm, Cytoflow
    Washdown capable
    No
    Current
    2.2 A
    Frequency
    50/60 Hz
    Voltage
    90 - 260 VAC
    Warranty
    2 years
    2 year warranty
    CE Compliant
Accessories
Product Information
  • Catalog #
    Length
    Availability
    Price
  • Extension cable for L/S and I/P precision modular drives
    Extension cable for L/S and I/P precision modular drives
    MFLX07557-51
    9 ft.
    Log in for availability
    Each (9Pie) - $11,510.00
Product Information
  • Catalog #
    Length
    Availability
    Price
  • Cable assembly for remote control, DB9(M) connector
    Cable assembly for remote control, DB9(M) connector
    MFLX07595-47
    7.9 m (25 ft.)
    Log in for availability
    Each (25Pie) - $6,380.00