Masterflex® L/S® Fixed-Speed VDC Compact Drives, Avantor®
Acerca de este artículo
Take it where you need it—ideal for field use.
- Flow rate 6 to 2700 ml/min with L/S tubing—flow rate depends on drive rpm, pump head and tubing size
- Rugged, continuous-duty motor is ideal for applications that require full-time duty cycle
- Vary input voltage to change drive speed and reverse input voltage polarity to reverse direction—Control drive completely using power source
- Constructed with chemical-resistant epoxy-coated steel motor housing for IP52 rating
These fixed-speed compact drives operate on 12 VDC input power. Wire power from your 12 or 24 VDC power source with three-wire, 16-gauge, 12-inch (30 cm) long stripped-end leads for power connection. Drives accept 8 different pump head types: Standard, Easy-Load®, Easy-Load® II, Easy-Load® 3, High-Performance, Cartridge, PTFE Tubing, or PTFE-Diaphragm†.
†Drives that accept only one pump head cannot be used with the High-Performance or PTFE-Tubing pump heads due to insufficient torque.
4 Options Available Below
Product Details & Documents
Take it where you need it—ideal for field use.
- Flow rate 6 to 2700 ml/min with L/S tubing—flow rate depends on drive rpm, pump head and tubing size
- Rugged, continuous-duty motor is ideal for applications that require full-time duty cycle
- Vary input voltage to change drive speed and reverse input voltage polarity to reverse direction—Control drive completely using power source
- Constructed with chemical-resistant epoxy-coated steel motor housing for IP52 rating
These fixed-speed compact drives operate on 12 VDC input power. Wire power from your 12 or 24 VDC power source with three-wire, 16-gauge, 12-inch (30 cm) long stripped-end leads for power connection. Drives accept 8 different pump head types: Standard, Easy-Load®, Easy-Load® II, Easy-Load® 3, High-Performance, Cartridge, PTFE Tubing, or PTFE-Diaphragm†.
†Drives that accept only one pump head cannot be used with the High-Performance or PTFE-Tubing pump heads due to insufficient torque.
Documents
Specifications for Product Family
Especificaciones
- Model:
- 07533-50
- 07533-60
- 07533-70
- 07533-80
- Control type:
- Fixed speed
- Fixed speed
- Fixed speed
- Fixed speed
- Drive height:
- 117 mm (4.625")
- 117 mm (4.625")
- 117 mm (4.625")
- 117 mm (4.625")
- Drive IP rating:
- IP 52
- IP 52
- IP 52
- IP 52
- Drive length:
- 216 mm (8.5625")
- 216 mm (8.5625")
- 216 mm (8.5625")
- 216 mm (8.5625")
- Drive width:
- 97 mm (3.8125")
- 97 mm (3.8125")
- 97 mm (3.8125")
- 97 mm (3.8125")
- Motor:
- 37 W
- 37 W
- 37 W
- 37 W
- Motor hp:
- 1/20
- 1/20
- 1/20
- 1/20
- Open-head sensor:
- No
- No
- No
- No
- Pump series:
- L/S
- L/S
- L/S
- L/S
- Speed:
- Fixed
- Fixed
- Fixed
- Fixed
- Speed control:
- ±5%
- ±5%
- ±5%
- ±5%
- Type of pump heads accepted:
- Standard, Easy-Load, Easy-Load II, Easy-Load 3, High Performance, PTFE-Tubing, PTFE Diaphragm
- Standard, Easy-Load, Easy-Load II, Easy-Load 3
- Standard, Easy-Load, Easy-Load II, Easy-Load 3, High Performance, PTFE-Tubing, PTFE Diaphragm
- Standard, Easy-Load, Easy-Load II, Easy-Load 3
- Warranty:
- 1 year
- 1 year
- 1 year
- 1 year
Especificaciones
Product Family Options
Información del producto
- Maximum Flow RateNo. of Pump Heads AcceptedHousing MaterialMax. RPMPowerAvailabilityPrice
- MFLX07533-50L/S Compact 12-VDC drive; 100 rpm.L/S Compact 12-VDC drive; 100 rpm.580 ml/min2Painted steel100 rpm12 VDC; 3.2 ALog in Para disponibilidadEach (1UN)$21,060.00Supplier Number: 7533-50Especificaciones:1 year warrantyISO9001
More
Specifications
Cat. No.MFLX07533-50Model07533-50Control typeFixed speedDrive height117 mm (4.625")Drive IP ratingIP 52Drive length216 mm (8.5625")Drive width97 mm (3.8125")Motor37 WMotor hp1/20Open-head sensorNoPump seriesL/SSpeedFixedSpeed control±5%Type of pump heads acceptedStandard, Easy-Load, Easy-Load II, Easy-Load 3, High Performance, PTFE-Tubing, PTFE DiaphragmWarranty1 year1 year warrantyISO9001 - MFLX07533-60L/S Compact 12-VDC drive; 540 rpm.L/S Compact 12-VDC drive; 540 rpm.2600 ml/min1Painted steel540 rpm12 VDC; 5.7 ALog in Para disponibilidadEach (1UN)$19,890.00Supplier Number: 7533-60Especificaciones:1 year warrantyISO9001
More
Specifications
Cat. No.MFLX07533-60Model07533-60Control typeFixed speedDrive height117 mm (4.625")Drive IP ratingIP 52Drive length216 mm (8.5625")Drive width97 mm (3.8125")Motor37 WMotor hp1/20Open-head sensorNoPump seriesL/SSpeedFixedSpeed control±5%Type of pump heads acceptedStandard, Easy-Load, Easy-Load II, Easy-Load 3Warranty1 year1 year warrantyISO9001 - MFLX07533-70L/S Compact 24-VDC drive; 100 rpm.L/S Compact 24-VDC drive; 100 rpm.580 ml/min2Painted steel100 rpm24 VDC; 1.5 ALog in Para disponibilidadEach (1UN)$21,260.00Supplier Number: 7533-70Especificaciones:1 year warrantyISO9001
More
Specifications
Cat. No.MFLX07533-70Model07533-70Control typeFixed speedDrive height117 mm (4.625")Drive IP ratingIP 52Drive length216 mm (8.5625")Drive width97 mm (3.8125")Motor37 WMotor hp1/20Open-head sensorNoPump seriesL/SSpeedFixedSpeed control±5%Type of pump heads acceptedStandard, Easy-Load, Easy-Load II, Easy-Load 3, High Performance, PTFE-Tubing, PTFE DiaphragmWarranty1 year1 year warrantyISO9001 - MFLX07533-80L/S Compact 24-VDC drive; 570 rpm.L/S Compact 24-VDC drive; 570 rpm.2700 ml/min1Painted steel570 rpm24 VDC; 2.6 ALog in Para disponibilidadEach (1UN)$19,890.00Supplier Number: 7533-80Especificaciones:1 year warrantyISO9001
More
Specifications
Cat. No.MFLX07533-80Model07533-80Control typeFixed speedDrive height117 mm (4.625")Drive IP ratingIP 52Drive length216 mm (8.5625")Drive width97 mm (3.8125")Motor37 WMotor hp1/20Open-head sensorNoPump seriesL/SSpeedFixedSpeed control±5%Type of pump heads acceptedStandard, Easy-Load, Easy-Load II, Easy-Load 3Warranty1 year1 year warrantyISO9001