Masterflex® L/S® Multichannel Cartridge Pump Head with Reduced Pulsation for Microbore 2-Stop Tubing, 4-Channel, 8-Roller
Catálogo # MFLX07519-20
Proveedor: Avantor Fluid Handling
2 year warranty
Especificaciones
- Tasa de flujo máxima:340 ml/min
- No. of channels:4
- Pump series:L/S
- Descripción:L/S® Multichannel Cartridge Pump Head with Reduced Pulsation for Microbore 2-Stop Tubing, 4-Channel, 8-Roller
- Altura:102 mm (4")
- Longitud:100 mm (3.9375")
- Gear ratio:1:1
- Max. flow rate per channel:85 ml/min
- Max. no. of cartridges (small):4
- Min. flow rate per channel:0.0006 ml/min
- Occlusion:Adjustable. Offset. Tangential
- Open-head sensor:No
- Optimum RPM range:0.02 - 250 rpm
- Tubing retention:Stopped Tubing or Manual
- Tubing sizes accepted:Two-stop microbore
- Volume per revolution with L/S 14 tubing:0.21 ml
- Volume per revolution with L/S 16 tubing:0.8 ml
- Volume per revolution with L/S 17 tubing:2.8 ml
- Volume per revolution with L/S 25 tubing:1.7 ml
- Garantía:2 years
- Referencia:MFLX07519-20
- Rollers:8
- Rotor Material:Stainless steel
- Anchura:111 mm (4.375")
- Housing Material:PSF
Especificaciones
Acerca de este artículo
Synchronous, low-pulsation flow from multiple channels.
- Cartridges snap in and out for fast tubing changes—no need to remove pump head from the drive
- Improved retainer design holds tubing securely in place
- Flow rate 0.00006 to 85 ml/min with microbore 2-stop tubing—flow rate depends on drive rpm, cartridge, and tubing size
- Accepts up to 4 large cartridges
- Require drives with a minimum of 1⁄10 hp and a maximum of 250 rpm
- Constructed with 8 Rulon® rollers, stainless steel rotor, and PSF ends
Each pump head system is composed of a heavy-duty pump head, cartridges, and tubing—each optimal preset tubing retention (order cartridges separately). They feature stainless steel rotors, Rulon® rollers, and PSF ends. Occlusion is offset, tangential, and adjustable. For small cartridges, optimum retention is preset—simply load tubing and start pumping. Recommended for use with small cartridge 07519-85 and microbore 2-stop tubing sizes.
- Tasa de flujo máxima:340 ml/min
- No. of channels:4
- Pump series:L/S
- Descripción:L/S® Multichannel Cartridge Pump Head with Reduced Pulsation for Microbore 2-Stop Tubing, 4-Channel, 8-Roller
- Altura:102 mm (4")
- Longitud:100 mm (3.9375")
- Environmentally Preferable:
- Gear ratio:1:1
- Max. flow rate per channel:85 ml/min
- Max. no. of cartridges (small):4
- Min. flow rate per channel:0.0006 ml/min
- Occlusion:Adjustable. Offset. Tangential
- Open-head sensor:No
- Optimum RPM range:0.02 - 250 rpm
- Tubing retention:Stopped Tubing or Manual
- Tubing sizes accepted:Two-stop microbore
- Volume per revolution with L/S 14 tubing:0.21 ml
- Volume per revolution with L/S 16 tubing:0.8 ml
- Volume per revolution with L/S 17 tubing:2.8 ml
- Volume per revolution with L/S 25 tubing:1.7 ml
- Garantía:2 years
- Referencia:MFLX07519-20
- Rollers:8
- Rotor Material:Stainless steel
- Anchura:111 mm (4.375")
- Housing Material:PSF