Orders LinkContactUsLinkComponent
Masterflex® L/S® Digital Modular Dispensing Pump Systems, Avantor®
Masterflex® L/S® Digital Modular Dispensing Pump Systems, Avantor®
Masterflex® L/S® Digital Modular Dispensing Pump Systems, Avantor®
Certificates
Masterflex® L/S® Digital Modular Dispensing Pump Systems, Avantor®
Masterflex® L/S® Digital Modular Dispensing Pump Systems, Avantor®
2 Options Available Below
Product Details & Documents
Precision digital drives offer three dispensing modes and precision low-maintenance motors— modular format lets you place motor and controller where convenient.
  • Digital interface with graphical LCD shows pump performance and operating mode— continuous run, timed dispense, copy dispense, or volume dispense
  • Remote control available using analog communication—voltage, current and contact closure
  • Select your preferred language from the user-friendly menu for programming and operation
  • Dispense by Volume in ml (0.001 to 9,999 mL), L, or gal; Copy—1 to 9,999 dispense cycles; or Time—time interval of on/off cycles (1 sec to 99 hr, 59 min, 59 sec per cycle)
  • Antidrip function ensures dispensing accuracy
  • Brushless, maintenance-free motor offers ±0.1% speed control accuracy with a 6000 to 1 turndown ratio
  • Reversible motor—purge tubing before or after pumping or pump fluid in either direction
  • Powerful motors let you stack pump heads for increased flow rates and for multichannel applications
These precision drives feature a programmable dispense interval that lets you set the delay between dispense cycles for convenient, automated dispensing by time or by volume. Precise PWM speed control (±0.1%) maximizes accuracy and efficiency while the brushless motor gives you quiet, maintenance-free reliability. Programmed calibration ensures dispense and display accuracy—drive stores one calibration value per tubing size. Program the drive via the simple membrane keypad; keypad lock/unlock feature protects settings.
Specifications for Product Family
Especificaciones
  • Model:
  • 77923-80
  • 77923-90
  • No. of channels:
  • 1
  • 1
  • No. of rollers:
  • 4
  • 4
  • Control type:
  • Digital
  • Digital
  • Min. flow rate per channel:
  • 0.28 ml/min
  • 0.28 ml/min
  • Max. flow rate per channel:
  • 1700 ml/min
  • 1700 ml/min
  • Min. flow rate:
  • 0.28 ml/min
  • 0.28 ml/min
  • Drive IP rating:
  • IP 33
  • IP 66
  • Accuracy:
  • ±0.1% (0.1 rpm at 600 rpm)
  • ±0.1% (0.1 rpm at 600 rpm)
  • Anti-drip:
  • Yes
  • Yes
  • Cloud connected:
  • No
  • No
  • Digital dispensing:
  • Yes
  • Yes
  • Drive:
  • 77301-40 (L/S® Digital Modular Dispensing Drive with Remote I/O)
  • 77301-50 (L/S® Digital Modular Washdown Dispensing Drive with Remote I/O)
  • Duty cycle:
  • Continuous
  • Continuous
  • Min. RPM:
  • 0.1 rpm
  • 0.1 rpm
  • Open-head sensor:
  • No
  • No
  • Pump series:
  • L/S
  • L/S
  • Remote I/O:
  • 0 to 20 mA. 4 to 20 mA. 0 to 10 V. start/stop. direction. and prime via contact closure
  • 0 to 20 mA. 4 to 20 mA. 0 to 10 V. start/stop. direction. and prime via contact closure
  • Reversible motor:
  • Yes
  • Yes
  • Tubing:
  • 06434-24, 10-ft (C-FLEX® Ultra, L/S® 24)
  • 06434-24, 10-ft (C-FLEX® Ultra, L/S® 24)
  • Tubing sizes accepted:
  • L/S® 15, 24, 35, 36
  • L/S® 15, 24, 35, 36
  • Washdown capable:
  • No
  • Yes
  • Current:
  • 2.2 A @ 115 V; 1.1 A @ 230 V
  • 2.2 A @ 115 V; 1.1 A @ 230 V
  • Frequency:
  • 50/60 Hz
  • 50/60 Hz
  • Voltage:
  • 90 - 260 VAC
  • 90 - 260 VAC
  • Warranty:
  • 2 years
  • 2 years
Especificaciones
Product Family Options
US plug models
Información del producto
  • Plug Type
    Maximum Flow Rate
    Height
    Length
    Width
    Pump Head
    Power
    Max. RPM
    Housing Material
    Availability
    Price
  • MFLX77923-80
    L/S® Digital Modular Dispensing Drive with Remote I/O and Easy-Load® II Pump Head; 90 to 260 VAC
    L/S® Digital Modular Dispensing Drive with Remote I/O and Easy-Load® II Pump Head; 90 to 260 VAC
    US
    1700 ml/min
    117.6 mm (4.63")
    332.2 mm (13.08")
    96.8 mm (3.81")
    77200-62 (L/S® Easy-Load® II pump head)
    90 - 260 VAC; 50/60 Hz; 2.2 A @ 115 V; 1.1 A @ 230 V
    600 rpm
    Painted steel
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $108,620.00
    $108,620.00
    Each (1UN)
       $108,620.00
    Supplier Number: 77923-80
    Especificaciones:
    2 year warranty
    CE Compliant
    More
    Specifications
    Cat. No.
    MFLX77923-80
    Model
    77923-80
    No. of channels
    1
    No. of rollers
    4
    Control type
    Digital
    Min. flow rate per channel
    0.28 ml/min
    Max. flow rate per channel
    1700 ml/min
    Min. flow rate
    0.28 ml/min
    Drive IP rating
    IP 33
    Accuracy
    ±0.1% (0.1 rpm at 600 rpm)
    Anti-drip
    Yes
    Cloud connected
    No
    Digital dispensing
    Yes
    Drive
    77301-40 (L/S® Digital Modular Dispensing Drive with Remote I/O)
    Duty cycle
    Continuous
    Min. RPM
    0.1 rpm
    Open-head sensor
    No
    Pump series
    L/S
    Remote I/O
    0 to 20 mA. 4 to 20 mA. 0 to 10 V. start/stop. direction. and prime via contact closure
    Reversible motor
    Yes
    Tubing
    06434-24, 10-ft (C-FLEX® Ultra, L/S® 24)
    Tubing sizes accepted
    L/S® 15, 24, 35, 36
    Washdown capable
    No
    Current
    2.2 A @ 115 V; 1.1 A @ 230 V
    Frequency
    50/60 Hz
    Voltage
    90 - 260 VAC
    Warranty
    2 years
    2 year warranty
    CE Compliant
  • MFLX77923-90
    L/S® Digital Modular Washdown Dispensing Drive with Remote I/O and Easy-Load® II Pump Head; 90 to 260 VAC
    L/S® Digital Modular Washdown Dispensing Drive with Remote I/O and Easy-Load® II Pump Head; 90 to 260 VAC
    US
    1700 ml/min
    152.4 mm (6")
    460.8 mm (18.14")
    177.8 mm (7")
    77200-62 (L/S® Easy-Load® II pump head)
    90 - 260 VAC; 50/60 Hz; 2.2 A @ 115 V; 1.1 A @ 230 V
    600 rpm
    Painted steel and aluminium
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $134,940.00
    $134,940.00
    Each (1UN)
       $134,940.00
    Supplier Number: 77923-90
    Especificaciones:
    2 year warranty
    CE Compliant
    IP66 washdown
    More
    Specifications
    Cat. No.
    MFLX77923-90
    Model
    77923-90
    No. of channels
    1
    No. of rollers
    4
    Control type
    Digital
    Min. flow rate per channel
    0.28 ml/min
    Max. flow rate per channel
    1700 ml/min
    Min. flow rate
    0.28 ml/min
    Drive IP rating
    IP 66
    Accuracy
    ±0.1% (0.1 rpm at 600 rpm)
    Anti-drip
    Yes
    Cloud connected
    No
    Digital dispensing
    Yes
    Drive
    77301-50 (L/S® Digital Modular Washdown Dispensing Drive with Remote I/O)
    Duty cycle
    Continuous
    Min. RPM
    0.1 rpm
    Open-head sensor
    No
    Pump series
    L/S
    Remote I/O
    0 to 20 mA. 4 to 20 mA. 0 to 10 V. start/stop. direction. and prime via contact closure
    Reversible motor
    Yes
    Tubing
    06434-24, 10-ft (C-FLEX® Ultra, L/S® 24)
    Tubing sizes accepted
    L/S® 15, 24, 35, 36
    Washdown capable
    Yes
    Current
    2.2 A @ 115 V; 1.1 A @ 230 V
    Frequency
    50/60 Hz
    Voltage
    90 - 260 VAC
    Warranty
    2 years
    2 year warranty
    CE Compliant
    IP66 washdown