Masterflex® Ismatec® Fixed-Speed Rack-Mount Minicartridge Peristaltic Pumps, Avantor®
Supplier: Avantor Fluid Handling
Certificates
About this item
Compact size mounting rod—use these pumps almost anywhere.
- A constant-speed, synchronous motor ensures a steady, repeatable flow rate
- Change tubing sizes to obtain different flow rates
Minicartridge design offers the easiest and most convenient tubing changes of all—even replace tubing while your pump is running! Order extra optional minicartridges with adjustable occlusion to manually vary pressure on each channel. Have extra 3-stop tubing on hand when fast tubing changes are crucial. (Scroll down to see all accessory choices.)
Delivery: Includes a 5-1/2"L x 1/2" dia (14 x 0.5 cm) mounting rod, a 6-ft (1.8-m) power cord with plug.
- Environmentally Preferable:
Specifications
- Catalog No:
- MFLX78012-30
- MFLX78012-20
- MFLX78012-50
- MFLX78012-40
- MFLX78012-65
- MFLX78012-55
- MFLX78012-45
- MFLX78012-35
- MFLX78012-25
- MFLX78012-15
- MFLX78012-05
- MFLX78012-97
- MFLX78012-95
- MFLX78012-85
- MFLX78012-60
- MFLX78012-90
- MFLX78012-80
- MFLX78012-92
- MFLX78012-00
- Modelo:
- 78012-30
- 78012-20
- 78012-50
- 78012-40
- 78012-65
- 78012-55
- 78012-45
- 78012-35
- 78012-25
- 78012-15
- 78012-05
- 78012-97
- 78012-95
- 78012-85
- 78012-60
- 78012-90
- 78012-80
- 78012-92
- 78012-00
- No. of channels:
- 4
- 2
- 4
- 4
- 4
- 4
- 4
- 4
- 2
- 2
- 2
- 2
- 2
- 2
- 4
- 2
- 2
- 2
- 2
- Número de rodillos:
- 8
- 8
- 6
- 8
- 6
- 6
- 8
- 8
- 8
- 8
- 8
- 6
- 6
- 6
- 6
- 6
- 6
- 6
- 8
- Tipo de control:
- Fixed-Speed
- Fixed-Speed
- Fixed-Speed
- Fixed-Speed
- Fixed-Speed
- Fixed-Speed
- Fixed-Speed
- Fixed-Speed
- Fixed-Speed
- Fixed-Speed
- Fixed-Speed
- Fixed-Speed
- Fixed-Speed
- Fixed-Speed
- Fixed-Speed
- Fixed-Speed
- Fixed-Speed
- Fixed-Speed
- Fixed-Speed
- Min. flow rate per channel:
- 6.2 ml/min
- 19 ml/min
- 7.4 ml/min
- 12 ml/min
- 15 ml/min
- 7.4 ml/min
- 12 ml/min
- 6.2 ml/min
- 19 ml/min
- 14 ml/min
- 6.2 ml/min
- 7.4 ml/min
- 7.4 ml/min
- 22 ml/min
- 15 ml/min
- 7.4 ml/min
- 22 ml/min
- 7.4 ml/min
- 6.2 ml/min
- Max. flow rate per channel:
- 6.2 ml/min
- 19 ml/min
- 7.4 ml/min
- 12 ml/min
- 15 ml/min
- 7.4 ml/min
- 12 ml/min
- 6.2 ml/min
- 19 ml/min
- 14 ml/min
- 6.2 ml/min
- 7.4 ml/min
- 7.4 ml/min
- 22 ml/min
- 15 ml/min
- 7.4 ml/min
- 22 ml/min
- 7.4 ml/min
- 6.2 ml/min
- Drive IP rating:
- IP 30
- IP 30
- IP 30
- IP 30
- IP 30
- IP 30
- IP 30
- IP 30
- IP 30
- IP 30
- IP 30
- IP 30
- IP 30
- IP 30
- IP 30
- IP 30
- IP 30
- IP 30
- IP 30
- Impulso:
- MS-CA
- MS-CA
- MS-CA
- MS-CA
- MS-CA
- MS-CA
- MS-CA
- MS-CA
- MS-CA
- MS-CA
- MS-CA
- MS-CA
- MS-CA
- MS-CA
- MS-CA
- MS-CA
- MS-CA
- MS-CA
- MS-CA
- Duty cycle:
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Continuous
- Open-head sensor:
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- Series:
- MS-CA
- MS-CA
- MS-CA
- MS-CA
- MS-CA
- MS-CA
- MS-CA
- MS-CA
- MS-CA
- MS-CA
- MS-CA
- MS-CA
- MS-CA
- MS-CA
- MS-CA
- MS-CA
- MS-CA
- MS-CA
- MS-CA
- Tubing sizes accepted:
- Three-stop 0.13 mm ID to 3.17 mm ID
- Three-stop 0.13 mm ID to 3.17 mm ID
- Three-stop 0.13 mm ID to 3.17 mm ID
- Three-stop 0.13 mm ID to 3.17 mm ID
- Three-stop 0.13 mm ID to 3.17 mm ID
- Three-stop 0.13 mm ID to 3.17 mm ID
- Three-stop 0.13 mm ID to 3.17 mm ID
- Three-stop 0.13 mm ID to 3.17 mm ID
- Three-stop 0.13 mm ID to 3.17 mm ID
- Three-stop 0.13 mm ID to 3.17 mm ID
- Three-stop 0.13 mm ID to 3.17 mm ID
- Three-stop 0.13 mm ID to 3.17 mm ID
- Three-stop 0.13 mm ID to 3.17 mm ID
- Three-stop 0.13 mm ID to 3.17 mm ID
- Three-stop 0.13 mm ID to 3.17 mm ID
- Three-stop 0.13 mm ID to 3.17 mm ID
- Three-stop 0.13 mm ID to 3.17 mm ID
- Three-stop 0.13 mm ID to 3.17 mm ID
- Three-stop 0.13 mm ID to 3.17 mm ID
- Washdown capable:
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- Presión máx.:
- 22 psi
- 22 psi
- 22 psi
- 22 psi
- 22 psi
- 22 psi
- 22 psi
- 22 psi
- 22 psi
- 22 psi
- 22 psi
- 22 psi
- 22 psi
- 22 psi
- 22 psi
- 22 psi
- 22 psi
- 22 psi
- 22 psi
- Garantía:
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years