Orders LinkContactUsLinkComponent
Masterflex® Metering Pump Accessories, Avantor®
Masterflex® Metering Pump Accessories, Avantor®
Masterflex® Metering Pump Accessories, Avantor®
Masterflex® Metering Pump Accessories, Avantor®
Masterflex® Metering Pump Accessories, Avantor®
Make the most of your Masterflex® metering pumps with these accessories.
  • Optimize performance of your Masterflex pumps
Choose from the following service kits and foot switches to meet your metering pump needs.
  • Warranty:
  • 1 year
  • 1 year
  • 1 year
  • 1 year
Product Family Options
  • Description
  • Foot valve for models with 3/8" tubing OD
    Each (1UN)
    Each (1UN) - $851.00
    $851.00
    Each (1UN)
       $851.00
    Specifications
    Cat. No.
    MFLX07135-49
    Warranty
    1 year
  • Foot Switch for Modular Touch-Screen Pumps
    Each (1UN)
    Each (1UN) - $3,850.00
    $3,850.00
    Each (1UN)
       $3,850.00
    Specifications
    Cat. No.
    MFLX74906-92
    Warranty
    1 year
  • Service kit for models with 1/4" tubing OD
    Specifications
    Cat. No.
    MFLX07135-38
    Warranty
    1 year
  • Service kit for models with 3/8" tubing OD, PTFE diaphragm
    Specifications
    Cat. No.
    MFLX07135-48
    Warranty