Masterflex® PTFE Ball Valves, Female NPT, Avantor®
Proveedor: MIDWEST VALVE & FITTING INC
Certificados
Acerca de este artículo
PTFE wetted parts allow for maximum chemical resistance.
- Bidirectional 1/4-turn from OFF to ON
- PTFE and polypropylene (PP) body construction
- Perfect for use in harsh environments
These Masterflex® PTFE ball valves are ideal for use in semiconductor or pharmaceutical manufacturing and chemical processing.
- Environmentally Preferable:
Especificaciones
- Catalog No:
- MFLX98514-05
- MFLX98514-09
- MFLX98514-01
- Modelo:
- 98514-05
- 98514-09
- 98514-01
- Presión máx.:
- 4.1 bar (60 psi)
- 4.1 bar (60 psi)
- 5.5 bar (80 psi)
- Max. temperature:
- 70 °C (158 °F)
- 70 °C (158 °F)
- 70 °C (158 °F)
- Wetted materials:
- PTFE
- PTFE
- PTFE
- Garantía:
- 1 year
- 1 year
- 1 year