Masterflex® Solenoid-Operated Pinch Valve For Masterflex® Pump Tubing, Avantor®
Proveedor: CLARK SOLUTIONS
Certificados
Acerca de este artículo
Specifically designed for Masterflex® pump tubing.
- Use with tubing that has durometer rating of 50 or less such as C-Flex® or silicone—7.25 psi max
- Excellent for clean processes—fluid stays contained within tubing
- Use for on/off flow control applications
Easily control the flow through your Masterflex tubing.
- Environmentally Preferable:
Especificaciones
- Catalog No:
- MFLX98306-24
- MFLX98306-28
- MFLX98306-30
- MFLX98306-20
- MFLX98306-18
- MFLX98306-16
- MFLX98306-08
- MFLX98306-04
- MFLX98306-02
- MFLX98306-12
- MFLX98306-14
- MFLX98306-26
- MFLX98306-00
- MFLX98306-10
- MFLX98306-22
- Alimentación:
- 12 VDC. 10 W
- 12 VDC. 10 W
- 24 VDC. 10 W
- 12 VDC. 10 W
- 24 VDC. 10 W
- 12 VDC. 10 W
- 12 VDC. 10 W
- 12 VDC. 10 W
- 24 VDC. 10 W
- 12 VDC. 10 W
- 24 VDC. 10 W
- 24 VDC. 10 W
- 12 VDC. 10 W
- 24 VDC. 10 W
- 24 VDC. 10 W
- Tiempo de respuesta:
- 30 - 50 ms
- 30 - 50 ms
- 30 - 50 ms
- 30 - 50 ms
- 30 - 50 ms
- 30 - 50 ms
- 30 - 50 ms
- 30 - 50 ms
- 30 - 50 ms
- 30 - 50 ms
- 30 - 50 ms
- 30 - 50 ms
- 30 - 50 ms
- 30 - 50 ms
- 30 - 50 ms
- Garantía:
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year