Check Valves with Pipe Thread Connections
Supplier: KODIAK CONTROLS
Acerca de este artículo
Create a secure pipe threaded connection.
- Choose from PVDF, PVC, or polypropylene (PP) bodies
All valves have Viton® diaphragms.
2 Options Available Below
Product Details & Documents
Create a secure pipe threaded connection.
- Choose from PVDF, PVC, or polypropylene (PP) bodies
All valves have Viton® diaphragms.
Specifications for Product Family
Especificaciones
- Model:
- 01350-20
- 01350-24
- 01350-26
- 01350-30
- 01350-32
- 01350-36
- Max. pressure:
- 140 psi
- 150 psi
- 125 psi
- 100 psi
- 100 psi
- 150 psi
- Max. temperature:
- 60 °C (140 °F)
- 60 °C (140 °F)
- 81 °C (180 °F)
- 81 °C (180 °F)
- 60 °C (140 °F)
- 60 °C (140 °F)
- Wetted materials:
- PVC, Viton
- PVC, Viton
- Polypropylene, Viton
- Polypropylene, Viton
- PTFE, Viton
- PVDF, Viton
Especificaciones
Product Family Options
Información del producto
- Breaking PressureCvConnection TypeInlet PortOutlet PortAvailabilityPrice
- MFLX01350-20PVC Check Valve, 1/4" NPT(F) connectionsPVC Check Valve, 1/4" NPT(F) connections0.5 psi0.5NPT(F)¼" NPT(F)¼" NPT(F)Discontinued-Especificaciones:
More
Specifications
Cat. No.MFLX01350-20Model01350-20Max. pressure140 psiMax. temperature60 °C (140 °F)Wetted materialsPVC, Viton - MFLX01350-24PVC Check Valve, 1/2" NPT(F) connectionsPVC Check Valve, 1/2" NPT(F) connections1.5 psi8NPT(F)½" NPT(F)½" NPT(F)Discontinued-Especificaciones:
More
Specifications
Cat. No.MFLX01350-24Model01350-24Max. pressure150 psiMax. temperature60 °C (140 °F)Wetted materialsPVC, Viton - MFLX01350-26PP Check Valve, 1/4" NPT (F) connectionsPP Check Valve, 1/4" NPT (F) connections0.5 psi0.5NPT(F)¼" NPT(F)¼" NPT(F)Discontinued-Especificaciones:
More
Specifications
Cat. No.MFLX01350-26Model01350-26Max. pressure125 psiMax. temperature81 °C (180 °F)Wetted materialsPolypropylene, Viton - MFLX01350-30PP Check Valve, 1/2" NPT (F) connectionsPP Check Valve, 1/2" NPT (F) connections1.5 psi8NPT(F)½" NPT(F)½" NPT(F)Log in Para disponibilidadEach (1UN)$4,680.00Especificaciones:
More
Specifications
Cat. No.MFLX01350-30Model01350-30Max. pressure100 psiMax. temperature81 °C (180 °F)Wetted materialsPolypropylene, Viton - MFLX01350-32PTFE Check Valve, 1/4" NPT(F) connectionsPTFE Check Valve, 1/4" NPT(F) connections0.5 psi0.5NPT(F)¼" NPT(F)¼" NPT(F)Discontinued-Especificaciones:
More
Specifications
Cat. No.MFLX01350-32Model01350-32Max. pressure100 psiMax. temperature60 °C (140 °F)Wetted materialsPTFE, Viton - MFLX01350-36PVDF Check Valve, 1/2" NPT(F) connectionsPVDF Check Valve, 1/2" NPT(F) connections1.5 psi8NPT(F)½" NPT(F)½" NPT(F)Log in Para disponibilidadEach (1UN)$8,760.00Especificaciones:
More
Specifications
Cat. No.MFLX01350-36Model01350-36Max. pressure150 psiMax. temperature60 °C (140 °F)Wetted materialsPVDF, Viton