Orders LinkContactUsLinkComponent
Masterflex® Sanitary Gaskets, Polymers, Avantor®
Masterflex® Sanitary Gaskets, Polymers, Avantor®
Supplier:  VWR International
Certificates
Masterflex® Sanitary Gaskets, Polymers, Avantor®
32 Options Available Below
Product Details & Documents
Ensure a tight seal.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Single-use custom configuration solutions to minimize downtime
These gaskets are for Tri-Clamp® style fittings and come in a wide variety of materials to match specific process needs. All gaskets meet USDA, 3-A and CGMP criteria. FDA Compliant. All silicone, EPDM, PTFE, and Viton® gaskets are animal-derived ingredient free and meet USP Class VI and cytotoxicity criteria.
Platinum-cured silicone is ideal for most pharmaceutical, food, and beverage processing applications. Odorless and tasteless to ensure media purity. May undergo SIP. Usable from –58 to 446 °F (–50 to 230 °C).
Viton offers excellent chemical resistance, especially to many acids and alkalis.
Specifications for Product Family
Especificaciones
  • Connection type:
  • Sanitary
  • Sanitary
  • Sanitary
  • Sanitary
  • Sanitary
  • Sanitary
  • Sanitary
  • Sanitary
  • Sanitary
  • Sanitary
  • Sanitary
  • Sanitary
  • Sanitary
  • Sanitary
  • Sanitary
  • Sanitary
  • Sanitary
  • Sanitary
  • Sanitary
  • Sanitary
  • Sanitary
  • Sanitary
  • Sanitary
  • Sanitary
  • Sanitary
  • Sanitary
  • Sanitary
  • Sanitary
  • Sanitary
  • Sanitary
  • Sanitary
  • Sanitary
  • Sanitary
  • Sanitary
  • Sanitary
  • Warranty:
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
Especificaciones
Product Family Options
Información del producto
  • Sanitary clamp size (Port 1)
    Fitting Material
    Sanitary Gasket Size
    Availability
    Price
  • MFLX30548-40
    Sanitary Gasket, Buna-N, 3/4" Tri-Clamp®; 10/PK
    Sanitary Gasket, Buna-N, 3/4" Tri-Clamp®; 10/PK
    ¾"
    Buna-N
    ¾"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (10UN)
    Each (10UN) - $486.00
    $486.00
    Each (10UN)
       $486.00
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX30548-40
    Connection type
    Sanitary
    Warranty
    1 year
  • MFLX30548-42
    Sanitary Gasket, Buna-N, 1" Tri-Clamp®; 10/PK
    Sanitary Gasket, Buna-N, 1" Tri-Clamp®; 10/PK
    1½"
    Buna-N
    1"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (10UN)
    Each (10UN) - $240.00
    $240.00
    Each (10UN)
       $240.00
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX30548-42
    Connection type
    Sanitary
    Warranty
    1 year
  • MFLX30548-44
    Sanitary Gasket, Buna-N, 1-1/2" Tri-Clamp®; 10/PK
    Sanitary Gasket, Buna-N, 1-1/2" Tri-Clamp®; 10/PK
    1½"
    Buna-N
    1½"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (10UN)
    Each (10UN) - $231.00
    $231.00
    Each (10UN)
       $231.00
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX30548-44
    Connection type
    Sanitary
    Warranty
    1 year
  • MFLX30548-46
    Sanitary Gasket, Buna-N, 2" Tri-Clamp®; 10/PK
    Sanitary Gasket, Buna-N, 2" Tri-Clamp®; 10/PK
    2"
    Buna-N
    2"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (10UN)
    Each (10UN) - $213.00
    $213.00
    Each (10UN)
       $213.00
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX30548-46
    Connection type
    Sanitary
    Warranty
    1 year
  • MFLX30548-48
    Sanitary Gasket, Buna-N, 2-1/2" Tri-Clamp®; 10/PK
    Sanitary Gasket, Buna-N, 2-1/2" Tri-Clamp®; 10/PK
    2"
    Buna-N
    2½"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (10UN)
    Each (10UN) - $256.00
    $256.00
    Each (10UN)
       $256.00
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX30548-48
    Connection type
    Sanitary
    Warranty
    1 year
  • MFLX29590-80
    Buna-N Sanitary Gasket, Black, 4" Tri-Clamp; 1/Pk
    Buna-N Sanitary Gasket, Black, 4" Tri-Clamp; 1/Pk
    4"
    Buna-N
    4"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $89.70
    $89.70
    Log in Para disponibilidad
    Paquete de 1
    Paquete de 1 - $93.40
    $93.40
    Each (1UN)
       $89.70
    Paquete de 1
       $93.40
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX29590-80
    Connection type
    Sanitary
    Warranty
    1 year
  • MFLX30548-60
    Sanitary Gasket, EPDM, 3/4" Tri-Clamp®; 10/PK
    Sanitary Gasket, EPDM, 3/4" Tri-Clamp®; 10/PK
    ¾"
    EPDM
    ¾"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (10UN)
    Each (10UN) - $500.00
    $500.00
    Each (10UN)
       $500.00
    Especificaciones:
    CE certificate
    More
    Specifications
    Cat. No.
    MFLX30548-60
    Connection type
    Sanitary
    Warranty
    1 year
    CE certificate
  • MFLX30548-62
    Sanitary Gasket, EPDM, 1" Tri-Clamp®; 10/PK
    Sanitary Gasket, EPDM, 1" Tri-Clamp®; 10/PK
    1½"
    EPDM
    1"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (10UN)
    Each (10UN) - $515.00
    $515.00
    Log in Para disponibilidad
    Paquete de 10
    Paquete de 10 - $547.00
    $547.00
    Each (10UN)
       $515.00
    Paquete de 10
       $547.00
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX30548-62
    Connection type
    Sanitary
    Warranty
    1 year
  • MFLX30548-64
    Sanitary Gasket, EPDM, 1-1/2" Tri-Clamp®; 10/PK
    Sanitary Gasket, EPDM, 1-1/2" Tri-Clamp®; 10/PK
    1½"
    EPDM
    1½"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (10UN)
    Each (10UN) - $488.00
    $488.00
    Each (10UN)
       $488.00
    Especificaciones:
    CE certificate
    More
    Specifications
    Cat. No.
    MFLX30548-64
    Connection type
    Sanitary
    Warranty
    1 year
    CE certificate
  • MFLX30548-66
    Sanitary Gasket, EPDM, 2" Tri-Clamp®; 10/PK
    Sanitary Gasket, EPDM, 2" Tri-Clamp®; 10/PK
    2"
    EPDM
    2"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (10UN)
    Each (10UN) - $525.00
    $525.00
    Log in Para disponibilidad
    Paquete de 10
    Paquete de 10 - $556.00
    $556.00
    Each (10UN)
       $525.00
    Paquete de 10
       $556.00
    Especificaciones:
    CE certificate
    More
    Specifications
    Cat. No.
    MFLX30548-66
    Connection type
    Sanitary
    Warranty
    1 year
    CE certificate
  • MFLX30548-68
    Sanitary Gasket, EPDM, 2-1/2" Tri-Clamp®; 10/PK
    Sanitary Gasket, EPDM, 2-1/2" Tri-Clamp®; 10/PK
    2"
    EPDM
    2½"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (10UN)
    Each (10UN) - $558.00
    $558.00
    Each (10UN)
       $558.00
    Especificaciones:
    CE certificate
    More
    Specifications
    Cat. No.
    MFLX30548-68
    Connection type
    Sanitary
    Warranty
    1 year
    CE certificate
  • MFLX29590-82
    EPDM Sanitary Gasket, 4" Tri-Clamp; 1/Pk
    EPDM Sanitary Gasket, 4" Tri-Clamp; 1/Pk
    4"
    EPDM
    4"
    Discontinued
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX29590-82
    Connection type
    Sanitary
    Warranty
    1 year
  • MFLX29590-83
    FKM/Viton Sanitary Gasket, 4" Tri-Clamp; 1/Pk
    FKM/Viton Sanitary Gasket, 4" Tri-Clamp; 1/Pk
    4"
    FKM/Viton
    4"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $375.00
    $375.00
    Log in Para disponibilidad
    Paquete de 1
    Paquete de 1 - $397.00
    $397.00
    Each (1UN)
       $375.00
    Paquete de 1
       $397.00
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX29590-83
    Connection type
    Sanitary
    Warranty
    1 year
  • MFLX00158-NC
    Sanitary Gaskets, Platinum Cured-Silicone, 3/4"; 10/PK
    Sanitary Gaskets, Platinum Cured-Silicone, 3/4"; 10/PK
    ¾"
    Platinum-Cured Silicone
    ½"
    Discontinued
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX00158-NC
    Connection type
    Sanitary
    Warranty
    1 year
  • MFLX00162-NJ
    Platinum-Cured Silicone Gasket, White, 2"; 10/PK
    Platinum-Cured Silicone Gasket, White, 2"; 10/PK
    2"
    Platinum-Cured Silicone
    2"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (10UN)
    Each (10UN) - $480.00
    $480.00
    Each (10UN)
       $480.00
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX00162-NJ
    Connection type
    Sanitary
    Warranty
    1 year
  • MFLX00364-YQ
    Sanitary Gasket, Platinum-Cured Silicone, 4"; 10/PK
    Sanitary Gasket, Platinum-Cured Silicone, 4"; 10/PK
    4"
    Platinum-Cured Silicone
    4"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (10UN)
    Each (10UN) - $1,350.00
    $1,350.00
    Each (10UN)
       $1,350.00
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX00364-YQ
    Connection type
    Sanitary
    Warranty
    1 year
  • MFLX00404-MO
    Sanitary Gasket, Platinum-Cured Silicone, 6"; 10/PK
    Sanitary Gasket, Platinum-Cured Silicone, 6"; 10/PK
    6"
    Platinum-Cured Silicone
    6"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (10UN)
    Each (10UN) - $8,000.00
    $8,000.00
    Each (10UN)
       $8,000.00
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX00404-MO
    Connection type
    Sanitary
    Warranty
    1 year
  • MFLX30548-00
    Sanitary Gasket, Silicone, 3/4" Tri-Clamp®; 10/PK
    Sanitary Gasket, Silicone, 3/4" Tri-Clamp®; 10/PK
    ¾"
    Platinum-Cured Silicone
    ¾"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (10UN)
    Each (10UN) - $558.00
    $558.00
    Log in Para disponibilidad
    Paquete de 10
    Paquete de 10 - $513.00
    $513.00
    Each (10UN)
       $558.00
    Paquete de 10
       $513.00
    Especificaciones:
    CE certificate
    More
    Specifications
    Cat. No.
    MFLX30548-00
    Connection type
    Sanitary
    Warranty
    1 year
    CE certificate
  • MFLX30548-02
    Sanitary Gasket, Silicone, 1"; 10/PK
    Sanitary Gasket, Silicone, 1"; 10/PK
    1½"
    Platinum-Cured Silicone
    1"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (10UN)
    Each (10UN) - $774.00
    $774.00
    Log in Para disponibilidad
    Paquete de 10
    Paquete de 10 - $706.00
    $706.00
    Each (10UN)
       $774.00
    Paquete de 10
       $706.00
    Especificaciones:
    CE certificate
    More
    Specifications
    Cat. No.
    MFLX30548-02
    Connection type
    Sanitary
    Warranty
    1 year
    CE certificate
  • MFLX30548-04
    Sanitary Gasket, Silicone, 1-1/2"; 10/PK
    Sanitary Gasket, Silicone, 1-1/2"; 10/PK
    1½"
    Platinum-Cured Silicone
    1½"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (10UN)
    Each (10UN) - $565.00
    $565.00
    Log in Para disponibilidad
    Paquete de 10
    Paquete de 10 - $513.00
    $513.00
    Each (10UN)
       $565.00
    Paquete de 10
       $513.00
    Especificaciones:
    CE certificate
    More
    Specifications
    Cat. No.
    MFLX30548-04
    Connection type
    Sanitary
    Warranty
    1 year
    CE certificate
  • MFLX30548-06
    Sanitary Gasket, Silicone, 2"; 10/PK
    Sanitary Gasket, Silicone, 2"; 10/PK
    2"
    Platinum-Cured Silicone
    2"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (10UN)
    Each (10UN) - $774.00
    $774.00
    Log in Para disponibilidad
    Paquete de 10
    Paquete de 10 - $706.00
    $706.00
    Each (10UN)
       $774.00
    Paquete de 10
       $706.00
    Especificaciones:
    CE certificate
    More
    Specifications
    Cat. No.
    MFLX30548-06
    Connection type
    Sanitary
    Warranty
    1 year
    CE certificate
  • MFLX30548-08
    Sanitary Gasket, Silicone, 2-1/2"; 10/PK
    Sanitary Gasket, Silicone, 2-1/2"; 10/PK
    2"
    Platinum-Cured Silicone
    2½"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (10UN)
    Each (10UN) - $780.00
    $780.00
    Log in Para disponibilidad
    Paquete de 10
    Paquete de 10 - $827.00
    $827.00
    Each (10UN)
       $780.00
    Paquete de 10
       $827.00
    Especificaciones:
    CE certificate
    More
    Specifications
    Cat. No.
    MFLX30548-08
    Connection type
    Sanitary
    Warranty
    1 year
    CE certificate
  • MFLX41001-77
    Sanitary Gasket, Platinum-Cured Silicone, 3"; 10/PK
    Sanitary Gasket, Platinum-Cured Silicone, 3"; 10/PK
    3"
    Platinum-Cured Silicone
    3"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (10UN)
    Each (10UN) - $899.00
    $899.00
    Log in Para disponibilidad
    Paquete de 10
    Paquete de 10 - $1,090.00
    $1,090.00
    Each (10UN)
       $899.00
    Paquete de 10
       $1,090.00
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX41001-77
    Connection type
    Sanitary
    Warranty
    1 year
  • MFLX30548-80
    Sanitary Gasket, PTFE, 3/4" Tri-Clamp®; 10/PK
    Sanitary Gasket, PTFE, 3/4" Tri-Clamp®; 10/PK
    ¾"
    PTFE
    ¾"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (10UN)
    Each (10UN) - $613.00
    $613.00
    Log in Para disponibilidad
    Paquete de 10
    Paquete de 10 - $652.00
    $652.00
    Each (10UN)
       $613.00
    Paquete de 10
       $652.00
    Especificaciones:
    CE certificate
    More
    Specifications
    Cat. No.
    MFLX30548-80
    Connection type
    Sanitary
    Warranty
    1 year
    CE certificate
  • MFLX30548-82
    Sanitary Gasket, PTFE, 1" Tri-Clamp®; 10/PK
    Sanitary Gasket, PTFE, 1" Tri-Clamp®; 10/PK
    1½"
    PTFE
    1"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (10UN)
    Each (10UN) - $808.00
    $808.00
    Log in Para disponibilidad
    Paquete de 10
    Paquete de 10 - $854.00
    $854.00
    Each (10UN)
       $808.00
    Paquete de 10
       $854.00
    Especificaciones:
    CE certificate
    More
    Specifications
    Cat. No.
    MFLX30548-82
    Connection type
    Sanitary
    Warranty
    1 year
    CE certificate
  • MFLX30548-84
    Sanitary Gasket, PTFE, 1-1/2" Tri-Clamp®; 10/PK
    Sanitary Gasket, PTFE, 1-1/2" Tri-Clamp®; 10/PK
    1½"
    PTFE
    1½"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (10UN)
    Each (10UN) - $747.00
    $747.00
    Log in Para disponibilidad
    Paquete de 10
    Paquete de 10 - $795.00
    $795.00
    Each (10UN)
       $747.00
    Paquete de 10
       $795.00
    Especificaciones:
    CE certificate
    More
    Specifications
    Cat. No.
    MFLX30548-84
    Connection type
    Sanitary
    Warranty
    1 year
    CE certificate
  • MFLX30548-86
    Sanitary Gasket, PTFE, 2" Tri-Clamp®; 10/PK
    Sanitary Gasket, PTFE, 2" Tri-Clamp®; 10/PK
    2"
    PTFE
    2"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (10UN)
    Each (10UN) - $780.00
    $780.00
    Log in Para disponibilidad
    Paquete de 10
    Paquete de 10 - $1,110.00
    $1,110.00
    Each (10UN)
       $780.00
    Paquete de 10
       $1,110.00
    Especificaciones:
    CE certificate
    More
    Specifications
    Cat. No.
    MFLX30548-86
    Connection type
    Sanitary
    Warranty
    1 year
    CE certificate
  • MFLX30548-88
    Sanitary Gasket, PTFE, 2-1/2" Tri-Clamp®; 10/PK
    Sanitary Gasket, PTFE, 2-1/2" Tri-Clamp®; 10/PK
    2"
    PTFE
    2½"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (10UN)
    Each (10UN) - $857.00
    $857.00
    Log in Para disponibilidad
    Paquete de 10
    Paquete de 10 - $907.00
    $907.00
    Each (10UN)
       $857.00
    Paquete de 10
       $907.00
    Especificaciones:
    CE certificate
    More
    Specifications
    Cat. No.
    MFLX30548-88
    Connection type
    Sanitary
    Warranty
    1 year
    CE certificate
  • MFLX16011-66
    Silicone Sanitary Gasket, 0.5" Tri-Clamp; 10/Pk
    Silicone Sanitary Gasket, 0.5" Tri-Clamp; 10/Pk
    ¾"
    Silicone
    0.5"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (10UN)
    Each (10UN) - $537.00
    $537.00
    Log in Para disponibilidad
    Paquete de 10
    Paquete de 10 - $565.00
    $565.00
    Each (10UN)
       $537.00
    Paquete de 10
       $565.00
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX16011-66
    Connection type
    Sanitary
    Warranty
    1 year
  • MFLX29590-81
    Silicone Sanitary Gasket, 4" Tri-Clamp; 1/Pk
    Silicone Sanitary Gasket, 4" Tri-Clamp; 1/Pk
    4"
    Silicone
    4"
    Discontinued
    Especificaciones:
    More
    Specifications
    Cat. No.
    MFLX29590-81
    Connection type
    Sanitary
    Warranty
    1 year
  • MFLX30548-20
    Sanitary Gasket, Viton®, 3/4" Tri-Clamp®; 10/PK
    Sanitary Gasket, Viton®, 3/4" Tri-Clamp®; 10/PK
    ¾"
    Viton®
    ¾"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (10UN)
    Each (10UN) - $1,107.00
    $1,107.00
    Log in Para disponibilidad
    Paquete de 10
    Paquete de 10 - $1,010.00
    $1,010.00
    Each (10UN)
       $1,107.00
    Paquete de 10
       $1,010.00
    Especificaciones:
    CE certificate
    More
    Specifications
    Cat. No.
    MFLX30548-20
    Connection type
    Sanitary
    Warranty
    1 year
    CE certificate
  • MFLX30548-22
    Sanitary Gasket, Viton®, 1" Tri-Clamp®; 10/PK
    Sanitary Gasket, Viton®, 1" Tri-Clamp®; 10/PK
    1½"
    Viton®
    1"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (10UN)
    Each (10UN) - $1,110.00
    $1,110.00
    Each (10UN)
       $1,110.00
    Especificaciones:
    CE certificate
    More
    Specifications
    Cat. No.
    MFLX30548-22
    Connection type
    Sanitary
    Warranty
    1 year
    CE certificate
  • MFLX30548-24
    Sanitary Gasket, Viton®, 1-1/2" Tri-Clamp®; 10/PK
    Sanitary Gasket, Viton®, 1-1/2" Tri-Clamp®; 10/PK
    1½"
    Viton®
    1½"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (10UN)
    Each (10UN) - $1,091.00
    $1,091.00
    Log in Para disponibilidad
    Paquete de 10
    Paquete de 10 - $995.00
    $995.00
    Each (10UN)
       $1,091.00
    Paquete de 10
       $995.00
    Especificaciones:
    CE certificate
    More
    Specifications
    Cat. No.
    MFLX30548-24
    Connection type
    Sanitary
    Warranty
    1 year
    CE certificate
  • MFLX30548-26
    Sanitary Gasket, Viton®, 2" Tri-Clamp®; 10/PK
    Sanitary Gasket, Viton®, 2" Tri-Clamp®; 10/PK
    2"
    Viton®
    2"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (10UN)
    Each (10UN) - $1,030.00
    $1,030.00
    Log in Para disponibilidad
    Paquete de 10
    Paquete de 10 - $1,100.00
    $1,100.00
    Each (10UN)
       $1,030.00
    Paquete de 10
       $1,100.00
    Especificaciones:
    CE certificate
    More
    Specifications
    Cat. No.
    MFLX30548-26
    Connection type
    Sanitary
    Warranty
    1 year
    CE certificate
  • MFLX30548-28
    Sanitary Gasket, Viton®, 2-1/2" Tri-Clamp®; 10/PK
    Sanitary Gasket, Viton®, 2-1/2" Tri-Clamp®; 10/PK
    2"
    Viton®
    2½"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (10UN)
    Each (10UN) - $1,200.00
    $1,200.00
    Log in Para disponibilidad
    Paquete de 10
    Paquete de 10 - $1,270.00
    $1,270.00
    Each (10UN)
       $1,200.00
    Paquete de 10
       $1,270.00
    Especificaciones:
    CE certificate
    More
    Specifications
    Cat. No.
    MFLX30548-28
    Connection type
    Sanitary
    Warranty
    1 year
    CE certificate