Masterflex® Interchangeable Power Cord Assemblies - IEC to Unique Country Plug, Avantor®
Supplier: VWR International
Certificates
About this item
Optimize performance of your Masterflex® pump.
- Choose from a wide range of power cords
Choose an interchangeable cord/plug assembly for 115 or 230 VAC operation. Each 6-ft (1.8 m) cord and moded plug assembly features an IEC 320 appliance connector and rated for 10 A at 250 VAC.
- Environmentally Preferable:
Specifications
- Catalog No:
- MFLX50001-70
- MFLX50001-72
- MFLX50001-68
- MFLX50001-69
- MFLX50001-60
- MFLX50001-64
- MFLX50001-78
- MFLX50001-76
- MFLX50001-74
- MFLX50001-62
- Modelo:
- 50001-70
- 50001-72
- 50001-68
- 50001-69
- 50001-60
- 50001-64
- 50001-78
- 50001-76
- 50001-74
- 50001-62
- Garantía:
- 1 year
- 1 year
- 1 year
- 1 year