Masterflex® Connection cable, "D" connector to stripped ends, 3-ft
MFLX98651-50
- Descripción:Connection cable, "D" connector to stripped ends
- Longitud:3 ft
- Modelo:98651-50
- Garantía:1 year
- Referencia:MFLX98651-50
Easy connect to your solenoid valve.
- Optimize performance of your Masterflex® Solenoid Valves
"D" connector connection cable with stripped wire ends for Aalborg stepping motor proportioning valves. This cable is 3-ft (0.9m) long.
- Descripción:Connection cable, "D" connector to stripped ends
- Longitud:3 ft
- Environmentally Preferable:
- Modelo:98651-50
- Garantía:1 year
- Referencia:MFLX98651-50