Orders LinkContactUsLinkComponent
Masterflex® B/T® Replacement Fuses, Avantor®
Masterflex® B/T® Replacement Fuses, Avantor®
Masterflex® B/T® Replacement Fuses, Avantor®
Certificates
Masterflex® B/T® Replacement Fuses, Avantor®
Masterflex® B/T® Replacement Fuses, Avantor®
12 Options Available Below
Product Details & Documents
Make the most of your drive with these replacement parts.
  • Optimize performance of your Masterflex pumps
Suitable for use with Masterflex® B/T® drive 77111-30.
Product Family Options
Información del producto
  • Compatible with
    Availability
    Price
  • MFLX77500-02
    Replacement Fuse, 5 A/250 V, 3 AG, Slo-Blo
    Replacement Fuse, 5 A/250 V, 3 AG, Slo-Blo
    Masterflex I/P Modular drives
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (2UN)
    Each (2UN) - $515.00
    $515.00
    Each (2UN)
       $515.00
    Supplier Number: 77500-02
  • MFLX77500-06
    Replacement Fuse, 3 A/250 V, 3 AG, standard
    Replacement Fuse, 3 A/250 V, 3 AG, standard
    Masterflex L/S and I/P drives
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (2UN)
    Each (2UN) - $1,190.00
    $1,190.00
    Each (2UN)
       $1,190.00
    Supplier Number: 77500-06
  • MFLX77500-08
    Replacement Fuse, 2,5 A/250 V, 5 X 20 mm, Slo-Blo
    Replacement Fuse, 2,5 A/250 V, 5 X 20 mm, Slo-Blo
    Masterflex I/P drives
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (2UN)
    Each (2UN) - $492.00
    $492.00
    Each (2UN)
       $492.00
    Supplier Number: 77500-08
  • MFLX77500-11
    Replacement Fuse, 1,6 A/250 V, 5 X 20 mm, Slo-Blo
    Replacement Fuse, 1,6 A/250 V, 5 X 20 mm, Slo-Blo
    Masterflex L/S drives
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (2UN)
    Each (2UN) - $517.00
    $517.00
    Each (2UN)
       $517.00
    Supplier Number: 77500-11
  • MFLX77500-24
    Replacement Fuse, 6,3 A/250 V, 5 X 20 mm, Slo-Blo
    Replacement Fuse, 6,3 A/250 V, 5 X 20 mm, Slo-Blo
    Masterflex I/P drives
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (2UN)
    Each (2UN) - $755.00
    $755.00
    Log in Para disponibilidad
    Paquete de 2
    Paquete de 2 - $754.00
    $754.00
    Each (2UN)
       $755.00
    Paquete de 2
       $754.00
    Supplier Number: 77500-24
  • MFLX77500-25
    Replacement Fuse, 3,15 A/250 V, 5 X 20 mm, Slo-Blo
    Replacement Fuse, 3,15 A/250 V, 5 X 20 mm, Slo-Blo
    Masterflex L/S and I/P drives
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (2UN)
    Each (2UN) - $779.00
    $779.00
    Log in Para disponibilidad
    Paquete de 2
    Paquete de 2 - $779.00
    $779.00
    Each (2UN)
       $779.00
    Paquete de 2
       $779.00
    Supplier Number: 77500-25
  • MFLX77500-28
    Replacement Fuse, 8 A/250 V, 3 AG, Slo-Blo
    Replacement Fuse, 8 A/250 V, 3 AG, Slo-Blo
    Masterflex B/T drives
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $439.00
    $439.00
    Each (1UN)
       $439.00
    Supplier Number: 77500-28
  • MFLX77500-30
    Replacement Fuse, 15 A/250 V, 5 X 20 mm, Slo-Blo
    Replacement Fuse, 15 A/250 V, 5 X 20 mm, Slo-Blo
    Masterflex B/T drives
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (2UN)
    Each (2UN) - $1,120.00
    $1,120.00
    Log in Para disponibilidad
    Paquete de 2
    Paquete de 2 - $1,100.00
    $1,100.00
    Each (2UN)
       $1,120.00
    Paquete de 2
       $1,100.00
    Supplier Number: 77500-30
  • MFLX77500-58
    Replacement Fuse, 1 A/250 V, 5 X 20 mm, Slo-Blo
    Replacement Fuse, 1 A/250 V, 5 X 20 mm, Slo-Blo
    Masterflex L/S drives
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (2UN)
    Each (2UN) - $509.00
    $509.00
    Each (2UN)
       $509.00
    Supplier Number: 77500-58
  • MFLX77500-12
    Replacement Fuse, 1,5 A/250 V, 3 AG, standard
    Replacement Fuse, 1,5 A/250 V, 3 AG, standard
    Masterflex L/S drives
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (2UN)
    Each (2UN) - $517.00
    $517.00
    Each (2UN)
       $517.00
    Supplier Number: 77500-12
  • MFLX77500-26
    Replacement Fuse, 4 A/250 V, 5 X 20 mm, Slo-Blo
    Replacement Fuse, 4 A/250 V, 5 X 20 mm, Slo-Blo
    Masterflex B/T drives
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (2UN)
    Each (2UN) - $404.00
    $404.00
    Each (2UN)
       $404.00
    Supplier Number: 77500-26
  • MFLX77500-27
    Replacement Fuse, 8 A/250 V, 5 X 20 mm, Slo-Blo
    Replacement Fuse, 8 A/250 V, 5 X 20 mm, Slo-Blo
    Masterflex B/T drives
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (2UN)
    Each (2UN) - $831.00
    $831.00
    Each (2UN)
       $831.00
    Supplier Number: 77500-27