Masterflex® Adapter Fittings, Hose Barb to Male NPT Threaded, Tee, Polypropylene, Avantor®
Supplier: VWR International
Certificates
About this item
Quick and easy assembly.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimize downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
These tee adapters help you install your flexible tubing lines into more rigid, threaded lines. Choose the size and material to suit your application needs.
- Environmentally Preferable:
Specifications
- Catalog No:
- MFLX40640-73
- MFLX40640-75
- MFLX40640-81
- MFLX40640-78
- MFLX40610-79
- MFLX40610-86
- MFLX40610-85
- MFLX40624-09
- MFLX40610-88
- MFLX40610-87
- MFLX40610-82
- MFLX40610-83
- MFLX40610-74
- MFLX40610-77
- MFLX40610-78
- MFLX40610-76
- MFLX40610-73
- MFLX40610-72
- Modelo:
- 40640-73
- 40640-75
- 40640-81
- 40640-78
- 40610-79
- 40610-86
- 40610-85
- 40624-09
- 40610-88
- 40610-87
- 40610-82
- 40610-83
- 40610-74
- 40610-77
- 40610-78
- 40610-76
- 40610-73
- 40610-72
- Orifice dimensions:
- 0.25"
- 0.125"
- 0.25"
- 0.25"
- 0.34375"
- 0.34375"
- 0.25"
- 0.25"
- 0.78125"
- 0.5625"
- 0.34375"
- 0.5625"
- 0.125"
- 0.125"
- 0.25"
- 0.34375"
- 0.25"
- 0.125"
- Sterilisation:
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) or gamma
- Autoclave or ethylene oxide (ETO)
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) or gamma
- Garantía:
- Out of box
- Out of box
- Out of box
- Out of box
- Out of box
- Out of box
- Out of box
- Out of box
- Out of box
- Out of box
- Out of box
- Out of box
- Out of box
- Out of box
- Out of box
- Out of box
- Out of box