Masterflex® Union Fittings, Hose Barb, Elbow, Stainless Steel, Avantor®
Quick and easy assembly without tools.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimize downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Create 90° bends in flexible tubing—without the kinks!
- Environmentally Preferable:
- Modelo:
- 30620-13
- 30620-14
- 30620-16
- 30620-15
- 30620-18
- 30620-17
- 30620-20
- Orifice dimensions:
- 0.09375"
- 0.09375"
- 0.1875"
- 0.125"
- 0.375"
- 0.3125"
- 0.6875"
- Sterilisation:
- Autoclave, ethylene oxide (ETO), or gamma
- Autoclave, ethylene oxide (ETO), or gamma
- Autoclave, ethylene oxide (ETO), or gamma
- Autoclave, ethylene oxide (ETO), or gamma
- Autoclave, ethylene oxide (ETO), or gamma
- Autoclave, ethylene oxide (ETO), or gamma
- Autoclave, ethylene oxide (ETO), or gamma
- Garantía:
- 1 Month
- 1 Month
- 1 Month
- 1 Month
- 1 Month
- 1 Month
- 1 Month
Frequently Bought Together