ContactUsLinkComponent
Masterflex® Union Fittings, Sanitary Clamp, 45° Elbow, Avantor®
Masterflex® Union Fittings, Sanitary Clamp, 45° Elbow, Avantor®
Supplier:  VWR International
Certificates
undefined
Masterflex® Union Fittings, Sanitary Clamp, 45° Elbow, Avantor®
Supplier:  VWR International
Certificates
Specifications
  • Chemical resistance:
  • Excellent
  • Max. operating pressure:
  • 150 psi in ambient conditions, using plastic true union style clamp
  • Max. temperature:
  • 138 °C (280 °F)
  • Min. temperature:
  • −40 °C (−40 °F)
  • Sterilisation:
  • Autoclave or ethylene oxide (ETO)
Specifications
Related Information
Frequently Bought Together