ContactUsLinkComponent
Masterflex® Single-Use Ultrasonic Flow Sensors, Avantor®
Masterflex® Single-Use Ultrasonic Flow Sensors, Avantor®
Proveedor:  VWR International
Certificados
undefined
Masterflex® Single-Use Ultrasonic Flow Sensors, Avantor®
Proveedor:  VWR International
Especificaciones
  • Precisión:
  • 2 to 4% after using calibration software
  • 2 to 4% after using calibration software
  • 2 to 4% after using calibration software
  • 2 to 4% after using calibration software
  • 2 to 4% after using calibration software
  • 2 to 4% after using calibration software
  • 2 to 4% after using calibration software
  • 2 to 4% after using calibration software
  • 2 to 4% after using calibration software
  • Temperatura máximo de operación:
  • 60 °C (140 °F)
  • 60 °C (140 °F)
  • 60 °C (140 °F)
  • 60 °C (140 °F)
  • 60 °C (140 °F)
  • 60 °C (140 °F)
  • 60 °C (140 °F)
  • 60 °C (140 °F)
  • 60 °C (140 °F)
  • Clasificación del gabinete:
  • IP 65
  • IP 65
  • IP 65
  • IP 65
  • IP 65
  • IP 65
  • IP 65
  • IP 65
  • IP 65
  • Garantía:
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year