Masterflex® Flowmeter and Flow Sensor Accessories, Avantor®
Supplier: AALBORG INSTRUMENTS & CONTROL
Masterflex® Flowmeter and Flow Sensor Accessories, Avantor®
Supplier: AALBORG INSTRUMENTS & CONTROL
Certificates
Acerca de este artículo
Make the most of your Masterflex flowmeters with these replacement parts and other accessories.
- Optimize performance of your flowmeter with these accessories
These power supplies, adapters, and other accessories are suitable for use with your Masterflex flowmeter and flow sensors.
4 Options Available Below
Product Details & Documents
Make the most of your Masterflex flowmeters with these replacement parts and other accessories.
- Optimize performance of your flowmeter with these accessories
These power supplies, adapters, and other accessories are suitable for use with your Masterflex flowmeter and flow sensors.
Specifications for Product Family
Especificaciones
- Model:
- 32498-40
- 32498-97
- 32775-50
- 32775-51
- 32775-52
- 32916-50
- 33501-96
- Connections:
- One USB connects to computor for power and data, 2nd USB for addtional power, M12 attaches to Masterflex Flow Sensor
- Warranty:
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
Especificaciones
Product Family Options
Información del producto
- Compatible withAvailabilityPrice
- MFLX32498-40Davis 110Vac/12VDC For Mass Flow ControllerEspecificaciones:
More
Specifications
Cat. No.MFLX32498-40Model32498-40ConnectionsWarranty1 year - MFLX32498-97Davis 110Vac/24VDC For Mass Flowmeter (Power Supply)Especificaciones:
More
Specifications
Cat. No.MFLX32498-97Model32498-97ConnectionsWarranty1 year - MFLX32775-50Adapter, 9/16"-18 UNF x 1/8" NPT(F)Adapter, 9/16"-18 UNF x 1/8" NPT(F)Discontinued-Especificaciones:
More
Specifications
Cat. No.MFLX32775-50Model32775-50ConnectionsWarranty1 year - MFLX32775-51Adapter, 9/16"-18 UNF x 1/4" NPT(F)Adapter, 9/16"-18 UNF x 1/4" NPT(F)Discontinued-Especificaciones:
More
Specifications
Cat. No.MFLX32775-51Model32775-51ConnectionsWarranty1 year - MFLX32775-52Adapter, 9/16"-18 UNF x 1/2" hose barbAdapter, 9/16"-18 UNF x 1/2" hose barbDiscontinued-Especificaciones:
More
Specifications
Cat. No.MFLX32775-52Model32775-52ConnectionsWarranty1 year - MFLX32916-50Power Adapter for Pressure Control Valves, 120/240 VAC, 50/60HzPower Adapter for Pressure Control Valves, 120/240 VAC, 50/60HzLog in Para disponibilidadEach (1UN)$2,560.00Especificaciones:CE Compliant
More
Specifications
Cat. No.MFLX32916-50Model32916-50ConnectionsWarranty1 yearCE Compliant - MFLX33501-96Masterflex Ultrasonic flow sensor communications packageMasterflex Ultrasonic flow sensor communications package33501-13, 33501-14, 33501-15, 33501-16, 33501-17, 33501-18, 33501-24, 33501-25, 33501-26, 33501-35, 33501-36, 33501-73Log in Para disponibilidadEach (1UN)$30,420.00Especificaciones:
More
Specifications
Cat. No.MFLX33501-96Model33501-96ConnectionsOne USB connects to computor for power and data, 2nd USB for addtional power, M12 attaches to Masterflex Flow SensorWarranty1 year