Orders LinkContactUsLinkComponent
Masterflex® Analog Gear Pump Systems, Avantor®
Masterflex® Analog Gear Pump Systems, Avantor®
Masterflex® Analog Gear Pump Systems, Avantor®
Masterflex® Analog Gear Pump Systems, Avantor®
Masterflex® Analog Gear Pump Systems, Avantor®
Simple variable speed control handles many fluid handling applications.
  • 100:1 speed-control range controls flow over a wide range
  • Simple operation with single-turn speed control
  • Separate power switch enables you to maintain speed setting when turning drive on/off; front-panel LED indicates power on
  • Remote control of start/stop via DB9 connector on back of drive
Compact, cost-effective drive offers simple variable speed control for many fluid handling applications from general transfer to filtration to chemical injection. Conveniently coupled with an A-mount gear pump head for pulseless fluid flow.
  • Body material:
  • 316 Stainless Steel
  • 316 Stainless Steel
  • 316 Stainless Steel
  • 316 Stainless Steel
  • 316 Stainless Steel
  • 316 Stainless Steel
  • 316 Stainless Steel
  • 316 Stainless Steel
  • 316 Stainless Steel
  • 316 Stainless Steel
  • 316 Stainless Steel
  • 316 Stainless Steel
  • 316 Stainless Steel
  • 316 Stainless Steel
  • 316 Stainless Steel
  • 316 Stainless Steel
  • Duty cycle continuous:
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Enclosure IP rating:
  • IP 22
  • IP 22
  • IP 22
  • IP 22
  • IP 22
  • IP 22
  • IP 22
  • IP 22
  • IP 22
  • IP 22
  • IP 22
  • IP 22
  • IP 22
  • IP 22
  • IP 22
  • IP 22
  • Motor hp:
  • 1/10
  • 1/10
  • 1/10
  • 1/10
  • 1/10
  • 1/10
  • 1/10
  • 1/10
  • 1/10
  • 1/10
  • 1/10
  • 1/10
  • 1/10
  • 1/10
  • 1/10
  • 1/10
  • Min. motor RPM:
  • 50 rpm
  • 50 rpm
  • 50 rpm
  • 50 rpm
  • 50 rpm
  • 50 rpm
  • 50 rpm
  • 50 rpm
  • 50 rpm
  • 50 rpm
  • 50 rpm
  • 50 rpm
  • 50 rpm
  • 50 rpm
  • 50 rpm
  • 50 rpm
  • Remote control input:
  • Start/Stop via Contact Closure
  • Start/Stop via Contact Closure
  • Start/Stop via Contact Closure
  • Start/Stop via Contact Closure
  • Start/Stop via Contact Closure
  • Start/Stop via Contact Closure
  • Start/Stop via Contact Closure
  • Start/Stop via Contact Closure
  • Start/Stop via Contact Closure
  • Start/Stop via Contact Closure
  • Start/Stop via Contact Closure
  • Start/Stop via Contact Closure
  • Start/Stop via Contact Closure
  • Start/Stop via Contact Closure
  • Start/Stop via Contact Closure
  • Start/Stop via Contact Closure
  • Warranty:
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
Product Family Options
  • Plug Type
    Flow rate
    Maximum Flow Rate
    Max. Motor RPM
    Power
    Width
    Length
    Height
  • 0.092 ml/rev
    460 ml/min
    5000 rpm
    115 VAC; 50/60 Hz
    18.3 cm (7.2")
    23 cm (23")
    13.4 cm (5.3")
    Each (1UN)
    Each (1UN) - $50,120.00
    $50,120.00
    Each (1UN)
       $50,120.00
    1 year warranty
    Specifications
    Cat. No.
    MFLX74013-02
    Body material
    316 Stainless Steel
    Duty cycle continuous
    Yes
    Enclosure IP rating
    IP 22
    Motor hp
    1/10
    Min. motor RPM
    50 rpm
    Remote control input
    Start/Stop via Contact Closure
    Warranty
    1 year
    1 year warranty
  • 0.092 ml/rev
    460 ml/min
    5000 rpm
    230 VAC; 50/60 Hz
    18.3 cm (7.2")
    23 cm (23")
    13.4 cm (5.3")
    Each (1UN)
    Each (1UN) - $52,260.00
    $52,260.00
    Each (1UN)
       $52,260.00
    1 year warranty
    Specifications
    Cat. No.
    MFLX74013-07
    Body material
    316 Stainless Steel
    Duty cycle continuous
    Yes
    Enclosure IP rating
    IP 22
    Motor hp
    1/10
    Min. motor RPM
    50 rpm
    Remote control input
    Start/Stop via Contact Closure
    Warranty
    1 year
    1 year warranty
  • 0.316 ml/rev
    1580 ml/min
    5000 rpm
    115 VAC; 50/60 Hz
    18.3 cm (7.2")
    23 cm (23")
    13.4 cm (5.3")
    Each (1UN)
    Each (1UN) - $49,920.00
    $49,920.00
    Each (1UN)
       $49,920.00
    1 year warranty
    Specifications
    Cat. No.
    MFLX74013-12
    Body material
    316 Stainless Steel
    Duty cycle continuous
    Yes
    Enclosure IP rating
    IP 22
    Motor hp
    1/10
    Min. motor RPM
    50 rpm
    Remote control input
    Start/Stop via Contact Closure
    Warranty
    1 year
    1 year warranty
  • 0.316 ml/rev
    1580 ml/min
    5000 rpm
    230 VAC; 50/60 Hz
    18.3 cm (7.2")
    23 cm (23")
    13.4 cm (5.3")
    Each (1UN)
    Each (1UN) - $49,920.00
    $49,920.00
    Each (1UN)
       $49,920.00
    1 year warranty
    Specifications
    Cat. No.
    MFLX74013-17
    Body material
    316 Stainless Steel
    Duty cycle continuous
    Yes
    Enclosure IP rating
    IP 22
    Motor hp
    1/10
    Min. motor RPM
    50 rpm
    Remote control input
    Start/Stop via Contact Closure
    Warranty
    1 year
    1 year warranty
  • 0.64 ml/rev
    3200 ml/min
    5000 rpm
    115 VAC; 50/60 Hz
    18.3 cm (7.2")
    23 cm (23")
    13.4 cm (5.3")
    Each (1UN)
    Each (1UN) - $51,480.00
    $51,480.00
    Each (1UN)
       $51,480.00
    1 year warranty
    Specifications
    Cat. No.
    MFLX74013-22
    Body material
    316 Stainless Steel
    Duty cycle continuous
    Yes
    Enclosure IP rating
    IP 22
    Motor hp
    1/10
    Min. motor RPM
    50 rpm
    Remote control input
    Start/Stop via Contact Closure
    Warranty
    1 year
    1 year warranty
  • 0.64 ml/rev
    3200 ml/min
    5000 rpm
    230 VAC; 50/60 Hz
    18.3 cm (7.2")
    23 cm (23")
    13.4 cm (5.3")
    Each (1UN)
    Each (1UN) - $51,480.00
    $51,480.00
    Each (1UN)
       $51,480.00
    1 year warranty
    Specifications
    Cat. No.
    MFLX74013-27
    Body material
    316 Stainless Steel
    Duty cycle continuous
    Yes
    Enclosure IP rating
    IP 22
    Motor hp
    1/10
    Min. motor RPM
    50 rpm
    Remote control input
    Start/Stop via Contact Closure
    Warranty
    1 year
    1 year warranty
  • 0.91 ml/rev
    4550 ml/min
    5000 rpm
    115 VAC; 50/60 Hz
    18.3 cm (7.2")
    23 cm (23")
    13.4 cm (5.3")
    Each (1UN)
    Each (1UN) - $58,310.00
    $58,310.00
    Each (1UN)
       $58,310.00
    1 year warranty
    Specifications
    Cat. No.
    MFLX74013-32
    Body material
    316 Stainless Steel
    Duty cycle continuous
    Yes
    Enclosure IP rating
    IP 22
    Motor hp
    1/10
    Min. motor RPM
    50 rpm
    Remote control input
    Start/Stop via Contact Closure
    Warranty
    1 year
    1 year warranty
  • 0.91 ml/rev
    4550 ml/min
    5000 rpm
    230 VAC; 50/60 Hz
    18.3 cm (7.2")
    23 cm (23")
    13.4 cm (5.3")
    Each (1UN)
    Each (1UN) - $53,040.00
    $53,040.00
    Each (1UN)
       $53,040.00
    1 year warranty
    Specifications
    Cat. No.
    MFLX74013-37
    Body material
    316 Stainless Steel
    Duty cycle continuous
    Yes
    Enclosure IP rating
    IP 22
    Motor hp
    1/10
    Min. motor RPM
    50 rpm
    Remote control input
    Start/Stop via Contact Closure
    Warranty
    1 year
    1 year warranty
  • 1.17 ml/rev
    5850 ml/min
    5000 rpm
    115 VAC; 50/60 Hz
    18.3 cm (7.2")
    23 cm (23")
    13.4 cm (5.3")
    Each (1UN)
    Each (1UN) - $50,900.00
    $50,900.00
    Each (1UN)
       $50,900.00
    1 year warranty
    Specifications
    Cat. No.
    MFLX74013-42
    Body material
    316 Stainless Steel
    Duty cycle continuous
    Yes
    Enclosure IP rating
    IP 22
    Motor hp
    1/10
    Min. motor RPM
    50 rpm
    Remote control input
    Start/Stop via Contact Closure
    Warranty
    1 year
    1 year warranty
  • 1.17 ml/rev
    5850 ml/min
    5000 rpm
    230 VAC; 50/60 Hz
    18.3 cm (7.2")
    23 cm (23")
    13.4 cm (5.3")
    Each (1UN)
    Each (1UN) - $54,020.00
    $54,020.00
    Each (1UN)
       $54,020.00
    1 year warranty
    Specifications
    Cat. No.
    MFLX74013-47
    Body material
    316 Stainless Steel
    Duty cycle continuous
    Yes
    Enclosure IP rating
    IP 22
    Motor hp
    1/10
    Min. motor RPM
    50 rpm
    Remote control input
    Start/Stop via Contact Closure
    Warranty
    1 year
    1 year warranty
  • 0.261 ml/rev
    1310 ml/min
    5000 rpm
    115 VAC; 50/60 Hz
    18.3 cm (7.2")
    23 cm (23")
    13.4 cm (5.3")
    Each (1UN)
    Each (1UN) - $49,340.00
    $49,340.00
    Each (1UN)
       $49,340.00
    1 year warranty
    Specifications
    Cat. No.
    MFLX74013-52
    Body material
    316 Stainless Steel
    Duty cycle continuous
    Yes
    Enclosure IP rating
    IP 22
    Motor hp
    1/10
    Min. motor RPM
    50 rpm
    Remote control input
    Start/Stop via Contact Closure
    Warranty
    1 year
    1 year warranty
  • 0.261 ml/rev
    1310 ml/min
    5000 rpm
    230 VAC; 50/60 Hz
    18.3 cm (7.2")
    23 cm (23")
    13.4 cm (5.3")
    Each (1UN)
    Each (1UN) - $51,680.00
    $51,680.00
    Each (1UN)
       $51,680.00
    1 year warranty
    Specifications
    Cat. No.
    MFLX74013-57
    Body material
    316 Stainless Steel
    Duty cycle continuous
    Yes
    Enclosure IP rating
    IP 22
    Motor hp
    1/10
    Min. motor RPM
    50 rpm
    Remote control input
    Start/Stop via Contact Closure
    Warranty
    1 year
    1 year warranty
  • 0.017 ml/rev
    85 ml/min
    5000 rpm
    115 VAC; 50/60 Hz
    18.3 cm (7.2")
    23 cm (23")
    13.4 cm (5.3")
    Each (1UN)
    Each (1UN) - $47,780.00
    $47,780.00
    Each (1UN)
       $47,780.00
    1 year warranty
    Specifications
    Cat. No.
    MFLX74013-72
    Body material
    316 Stainless Steel
    Duty cycle continuous
    Yes
    Enclosure IP rating
    IP 22
    Motor hp
    1/10
    Min. motor RPM
    50 rpm
    Remote control input
    Start/Stop via Contact Closure
    Warranty
    1 year
    1 year warranty
  • No
    0.017 ml/rev
    85 ml/min
    5000 rpm
    230 VAC; 50/60 Hz
    18.3 cm (7.2")
    23 cm (23")
    13.4 cm (5.3")
    Each (1UN)
    Each (1UN) - $63,380.00
    $63,380.00
    Each (1UN)
       $63,380.00
    1 year warranty
    Irregular Voltage
    Specifications
    Cat. No.
    MFLX74013-77
    Body material
    316 Stainless Steel
    Duty cycle continuous
    Yes
    Enclosure IP rating
    IP 22
    Motor hp
    1/10
    Min. motor RPM
    50 rpm
    Remote control input
    Start/Stop via Contact Closure
    Warranty
    1 year
    1 year warranty
    Irregular Voltage
  • USN
    0.017 ml/rev
    85 ml/min
    5000 rpm
    230 VAC; 50/60 Hz
    18.3 cm (7.2")
    23 cm (23")
    13.4 cm (5.3")
    1 year warranty
    Irregular Voltage
    Specifications
    Cat. No.
    MFLX74013-77-USN
    Body material
    316 Stainless Steel
    Duty cycle continuous
    Yes
    Enclosure IP rating
    IP 22
    Motor hp
    1/10
    Min. motor RPM
    50 rpm
    Remote control input
    Start/Stop via Contact Closure
    Warranty
    1 year
    1 year warranty
    Irregular Voltage
  • USS
    0.017 ml/rev
    85 ml/min
    5000 rpm
    230 VAC; 50/60 Hz
    18.3 cm (7.2")
    23 cm (23")
    13.4 cm (5.3")
    1 year warranty
    Specifications
    Cat. No.
    MFLX74013-77-USS
    Body material
    316 Stainless Steel
    Duty cycle continuous
    Yes
    Enclosure IP rating
    IP 22
    Motor hp
    1/10
    Min. motor RPM
    50 rpm
    Remote control input
    Start/Stop via Contact Closure
    Warranty
    1 year
    1 year warranty