Orders LinkContactUsLinkComponent
Masterflex® Digital Benchtop Gear Pump Systems, Avantor®
Masterflex® Digital Benchtop Gear Pump Systems, Avantor®
Masterflex® Digital Benchtop Gear Pump Systems, Avantor®
Certificates
Masterflex® Digital Benchtop Gear Pump Systems, Avantor®
8 Options Available Below
Product Details & Documents
Order the complete pump drive and head system.
  • Graphical LCD shows four operating modes
  • IP33 rating ensures splash and dust resistance
  • Systems deliver pulse-free flow in combination with Micropump® A-mount head
  • Remote control via DB25 female connector on drive
Calibrate, copy, and dispense features produce precise, pulseless batching and dispensing. View pump performance continually. Retrieve displacement and flow values by pump head size.
These precision systems are easily operated from the intuitive, multilanguage LCD front panel or optional foot switch 07523-92 for hands-free on/off control. The IP33-rated enclosure protects internal components from damage.
Service kits 07144-42 and 74012-52 include: gears, suction shoe, shaft seals, and springs
Service kit 74012-22 includes: gears, bushings, and seals
Delivery information: Includes 6-ft (1.8-m) line cord with IEC 320/CEE22 socket. These models are shipped with country specific plug/cord set; please specify ultimate destination when ordering.
Specifications for Product Family
Specifications
  • Model:
  • 74014-72
  • 74014-02
  • 74014-62
  • 74014-52
  • 74014-12
  • 74014-22
  • 74014-32
  • 74014-42
  • Differential pressure:
  • 1.38 bar (20 psi)
  • 5.17 bar (75 psi)
  • 5.17 bar (75 psi)
  • 8.61 bar (125 psi)
  • 5.17 bar (75 psi)
  • 5.17 bar (75 psi)
  • 5.17 bar (75 psi)
  • 4.14 bar (60 psi)
  • Wetted materials:
  • 316 SS, Graphite, PTFE
  • 316 SS, PPS, PTFE
  • 316 SS, PPS, PPS
  • 316 SS, PEEK, Viton
  • 316 SS, PPS, PTFE
  • 316 SS, PPS, PTFE
  • 316 SS, PPS, PTFE
  • 316 SS, PPS, Viton
  • Warranty:
  • See message
  • See message
  • See message
  • See message
  • See message
  • See message
  • See message
  • See message
Specifications
Product Family Options
US plug models
Product Information
  • Plug Type
    Flow rate
    Maximum Flow Rate
    Power
    Availability
    Price
  • MFLX74014-72
    Digital Gear Pump System, 0,017 ml/rev, 90-260 VAC
    Digital Gear Pump System, 0,017 ml/rev, 90-260 VAC
    US
    0.017 ml/rev
    61 ml/min
    90 - 260 VAC
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $77,030.00
    $77,030.00
    Each (1UN)
       $77,030.00
    Supplier Number: 74014-72
    Specifications:
    2 year warranty
    CE Compliant
    ETL listed
    More
    Specifications
    Cat. No.
    MFLX74014-72
    Model
    74014-72
    Differential pressure
    1.38 bar (20 psi)
    Wetted materials
    316 SS, Graphite, PTFE
    Warranty
    See message
    2 year warranty
    CE Compliant
    ETL listed
  • MFLX74014-02
    Digital Gear Pump System, 0,092 ml/rev, 90-260 VAC
    Digital Gear Pump System, 0,092 ml/rev, 90-260 VAC
    US
    0.092 ml/rev
    331.2 ml/min
    90 - 260 VAC
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $78,980.00
    $78,980.00
    Each (1UN)
       $78,980.00
    Supplier Number: 74014-02
    Specifications:
    2 year warranty
    CE Compliant
    ETL listed
    More
    Specifications
    Cat. No.
    MFLX74014-02
    Model
    74014-02
    Differential pressure
    5.17 bar (75 psi)
    Wetted materials
    316 SS, PPS, PTFE
    Warranty
    See message
    2 year warranty
    CE Compliant
    ETL listed
  • MFLX74014-62
    Digital Gear Pump System, 0,092 ml/rev, 90-260 VAC SKU 7401462
    Digital Gear Pump System, 0,092 ml/rev, 90-260 VAC SKU 7401462
    US
    0.092 ml/rev
    331.2 ml/min
    90 - 260 VAC
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $75,470.00
    $75,470.00
    Each (1UN)
       $75,470.00
    Supplier Number: 74014-62
    Specifications:
    2 year warranty
    CE Compliant
    ETL listed
    More
    Specifications
    Cat. No.
    MFLX74014-62
    Model
    74014-62
    Differential pressure
    5.17 bar (75 psi)
    Wetted materials
    316 SS, PPS, PPS
    Warranty
    See message
    2 year warranty
    CE Compliant
    ETL listed
  • MFLX74014-52
    Digital Gear Pump System, 0,261 ml/rev, 90-260 VAC
    Digital Gear Pump System, 0,261 ml/rev, 90-260 VAC
    US
    0.261 ml/rev
    939.6 ml/min
    90 - 260 VAC
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $78,390.00
    $78,390.00
    Each (1UN)
       $78,390.00
    Supplier Number: 74014-52
    Specifications:
    2 year warranty
    CE Compliant
    ETL listed
    More
    Specifications
    Cat. No.
    MFLX74014-52
    Model
    74014-52
    Differential pressure
    8.61 bar (125 psi)
    Wetted materials
    316 SS, PEEK, Viton
    Warranty
    See message
    2 year warranty
    CE Compliant
    ETL listed
  • MFLX74014-12
    Digital Gear Pump System, 0,316 ml/rev, 90-260 VAC
    Digital Gear Pump System, 0,316 ml/rev, 90-260 VAC
    US
    0.316 ml/rev
    1137.6 ml/min
    90 - 260 VAC
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $79,760.00
    $79,760.00
    Each (1UN)
       $79,760.00
    Supplier Number: 74014-12
    Specifications:
    2 year warranty
    CE Compliant
    ETL listed
    More
    Specifications
    Cat. No.
    MFLX74014-12
    Model
    74014-12
    Differential pressure
    5.17 bar (75 psi)
    Wetted materials
    316 SS, PPS, PTFE
    Warranty
    See message
    2 year warranty
    CE Compliant
    ETL listed
  • MFLX74014-22
    Digital Gear Pump System, 0,64 ml/rev, 90-260 VAC
    Digital Gear Pump System, 0,64 ml/rev, 90-260 VAC
    US
    0.64 ml/rev
    2304 ml/min
    90 - 260 VAC
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $82,100.00
    $82,100.00
    Each (1UN)
       $82,100.00
    Supplier Number: 74014-22
    Specifications:
    2 year warranty
    CE Compliant
    ETL listed
    More
    Specifications
    Cat. No.
    MFLX74014-22
    Model
    74014-22
    Differential pressure
    5.17 bar (75 psi)
    Wetted materials
    316 SS, PPS, PTFE
    Warranty
    See message
    2 year warranty
    CE Compliant
    ETL listed
  • MFLX74014-32
    Digital Gear Pump System, 0,91 ml/rev, 90-260 VAC
    Digital Gear Pump System, 0,91 ml/rev, 90-260 VAC
    US
    0.91 ml/rev
    3276 ml/min
    90 - 260 VAC
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $81,320.00
    $81,320.00
    Each (1UN)
       $81,320.00
    Supplier Number: 74014-32
    Specifications:
    2 year warranty
    CE Compliant
    ETL listed
    More
    Specifications
    Cat. No.
    MFLX74014-32
    Model
    74014-32
    Differential pressure
    5.17 bar (75 psi)
    Wetted materials
    316 SS, PPS, PTFE
    Warranty
    See message
    2 year warranty
    CE Compliant
    ETL listed
  • MFLX74014-42
    Digital Gear Pump System, 1,17 ml/rev, 90-260 VAC
    Digital Gear Pump System, 1,17 ml/rev, 90-260 VAC
    US
    1.17 ml/rev
    4212 ml/min
    90 - 260 VAC
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $78,590.00
    $78,590.00
    Each (1UN)
       $78,590.00
    Supplier Number: 74014-42
    Specifications:
    2 year warranty
    CE Compliant
    ETL listed
    More
    Specifications
    Cat. No.
    MFLX74014-42
    Model
    74014-42
    Differential pressure
    4.14 bar (60 psi)
    Wetted materials
    316 SS, PPS, Viton
    Warranty
    See message
    2 year warranty
    CE Compliant
    ETL listed
Accessories
Product Information
  • Catalog #
    Barb size/tubing int.Ø (port 2)
    Connection Type
    Availability
    Price
  • Fitting, HDPE, Straight, Hosebarb to Thread Adapter, ¼" ID x ⅛" NPT(M); 10/PK
    Fitting, HDPE, Straight, Hosebarb to Thread Adapter, ¼" ID x ⅛" NPT(M); 10/PK
    MFLX30621-31
    ¼"
    Thread to Barb
    Log in for availability
    Each (10UN) - $431.00
Product Information
  • Catalog #
    Barb size/tubing int.Ø (port 1)
    Barb size/tubing int.Ø (port 2)
    Availability
    Price
  • Fitting,polypropylene, straight, hose barb reducer, int.Ø ⅝×½";10/pack
    Fitting,polypropylene, straight, hose barb reducer, int.Ø ⅝×½";10/pack
    MFLX40610-16
    ⅝"
    ½"
    Log in for availability
    Each (10UN) - $539.00
Product Information
  • Catalog #
    Availability
    Price
  • Footswitch, momentary start/stop, DB25, 6' cable
    Footswitch, momentary start/stop, DB25, 6' cable
    MFLX07523-92
    Log in for availability
    Each (6Pie) - $7,730.00