Orders LinkContactUsLinkComponent
Masterflex® Drum Pump Replacement Motors, Avantor®
Supplier:   VWR International
Masterflex® Drum Pump Replacement Motors, Avantor®
Supplier:  VWR International
Certificates
Masterflex® Drum Pump Replacement Motors, Avantor®
3 Options Available Below
Product Details & Documents
Safety transfer a variety of liquids from drums, tanks, and small storage vessels.
  • Use with Masterflex® Complete Drum and Container Pumps
Drum and container pumps are engineered to safely transfer a variety of liquids from drums, tanks and small storage vessels. These Masterflex® drum pump replacement motors are one component of a system that includes a pump tube and motor (order pump tubes separately). Motors are equipped with variable-speed control for added safety and control.
Specifications for Product Family
Especificaciones
  • Warranty:
  • 3 years
  • 3 years
  • 3 years
Especificaciones
Product Family Options
Información del producto
  • Motor Type
    Power
    Availability
    Price
  • MFLX70604-03
    Replacement motor, ODP, 110 VAC, 60 Hz
    Replacement motor, ODP, 110 VAC, 60 Hz
    Open drip proof (ODP)
    110 VAC; 60 Hz
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $20,090.00
    $20,090.00
    Each (1UN)
       $20,090.00
    Especificaciones:
    ETL Canada listed
    More
    Specifications
    Cat. No.
    MFLX70604-03
    Warranty
    3 years
    ETL Canada listed
  • MFLX70604-13
    Replacement motor, TEFC, 110 VAC, 60 Hz
    Replacement motor, TEFC, 110 VAC, 60 Hz
    Totally enclosed, fan cooled (TEFC)
    110 VAC; 60 Hz
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $26,330.00
    $26,330.00
    Each (1UN)
       $26,330.00
    Especificaciones:
    CSA Certified
    ETL Canada listed
    More
    Specifications
    Cat. No.
    MFLX70604-13
    Warranty
    3 years
    CSA Certified
    ETL Canada listed
  • MFLX70604-16
    Replacement motor, TEFC, 230 VAC, 50 Hz
    Replacement motor, TEFC, 230 VAC, 50 Hz
    Totally enclosed fan cooled (TEFC)
    230 VAC; 50 Hz
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $28,080.00
    $28,080.00
    Each (1UN)
       $28,080.00
    Especificaciones:
    CE Compliant
    More
    Specifications
    Cat. No.
    MFLX70604-16
    Warranty
    3 years
    CE Compliant